Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96196803-96197064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLNK
Entrez Gene ID
29760
Gene Name
B-cell linker
Gene Aliases
AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca
UniGene
Hs.665244
Gene Chromosome Location
Chr.10: 96189171-96271576 on Build GRCh38
Transcripts
NM_001114094.1 NM_001258440.1 NM_001258441.1 NM_001258442.1 NM_013314.3 NR_047680.1 NR_047681.1 NR_047682.1 NR_047683.1 XM_011539728.2 XM_017016159.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4016966 c.1115G>A p.R372Q Chr.10: 96197044-96197044 on Build GRCh38
1638375 c.1204G>A p.E402K Chr.10: 96196955-96196955 on Build GRCh38
1638374 c.1228G>A p.G410S Chr.10: 96196931-96196931 on Build GRCh38
4914955 c.1181A>G p.Y394C Chr.10: 96196978-96196978 on Build GRCh38
6413338 c.1109T>C p.L370P Chr.10: 96197050-96197050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782319580 Chr.10: 96196962-96196962 on Build GRCh38
rs200470518 Chr.10: 96196868-96196868 on Build GRCh38
rs782657554 Chr.10: 96196858-96196858 on Build GRCh38
rs372532123 Chr.10: 96196984-96196984 on Build GRCh38
rs781853086 Chr.10: 96196869-96196869 on Build GRCh38
rs782174916 Chr.10: 96196958-96196958 on Build GRCh38
rs769746902 Chr.10: 96197007-96197007 on Build GRCh38
rs141103595 Chr.10: 96197019-96197019 on Build GRCh38
rs376755926 Chr.10: 96196880-96196880 on Build GRCh38
rs371617732 Chr.10: 96197028-96197028 on Build GRCh38
rs201039662 Chr.10: 96197021-96197021 on Build GRCh38
rs745631180 Chr.10: 96196872-96196875 on Build GRCh38
rs782179925 Chr.10: 96196895-96196895 on Build GRCh38
rs782323001 Chr.10: 96196896-96196896 on Build GRCh38
rs782088570 Chr.10: 96196967-96196967 on Build GRCh38
rs201384181 Chr.10: 96196957-96196957 on Build GRCh38
rs782481221 Chr.10: 96197031-96197031 on Build GRCh38
rs1042718588 Chr.10: 96196845-96196845 on Build GRCh38
rs782763305 Chr.10: 96196977-96196977 on Build GRCh38
rs781820500 Chr.10: 96197029-96197029 on Build GRCh38
rs782362176 Chr.10: 96196956-96196956 on Build GRCh38
rs782587540 Chr.10: 96196908-96196908 on Build GRCh38
rs587693697 Chr.10: 96196854-96196854 on Build GRCh38
rs782220932 Chr.10: 96196934-96196934 on Build GRCh38
rs782635269 Chr.10: 96197037-96197037 on Build GRCh38
rs782786523 Chr.10: 96196986-96196986 on Build GRCh38
rs782552788 Chr.10: 96196878-96196878 on Build GRCh38
rs782433581 Chr.10: 96197023-96197023 on Build GRCh38
rs267602649 Chr.10: 96196963-96196963 on Build GRCh38

View Full Product Details