Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9029238-9029499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM9B
Entrez Gene ID
171483
Gene Name
family with sequence similarity 9 member B
Gene Aliases
TEX39B
UniGene
Hs.733351
Gene Chromosome Location
Chr.X: 9024232-9197176 on Build GRCh38
Transcripts
NM_205849.3 XM_011545464.2

SNP Detail

SNP ID Chromosome Location
rs777746215 Chr.X: 9029335-9029335 on Build GRCh38
rs765938915 Chr.X: 9029412-9029412 on Build GRCh38
rs749220187 Chr.X: 9029354-9029354 on Build GRCh38
rs1020068816 Chr.X: 9029425-9029425 on Build GRCh38
rs777138780 Chr.X: 9029380-9029380 on Build GRCh38
rs369994703 Chr.X: 9029441-9029441 on Build GRCh38
rs140135828 Chr.X: 9029309-9029309 on Build GRCh38
rs200007521 Chr.X: 9029329-9029331 on Build GRCh38
rs773919417 Chr.X: 9029410-9029410 on Build GRCh38
rs750523826 Chr.X: 9029416-9029416 on Build GRCh38
rs767023855 Chr.X: 9029432-9029432 on Build GRCh38
rs766515216 Chr.X: 9029411-9029411 on Build GRCh38
rs190526660 Chr.X: 9029442-9029442 on Build GRCh38
rs1008145444 Chr.X: 9029417-9029417 on Build GRCh38
rs755962799 Chr.X: 9029328-9029329 on Build GRCh38
rs372330002 Chr.X: 9029314-9029314 on Build GRCh38
rs750996017 Chr.X: 9029415-9029415 on Build GRCh38
rs762206542 Chr.X: 9029385-9029385 on Build GRCh38
rs775407537 Chr.X: 9029363-9029363 on Build GRCh38
rs376783680 Chr.X: 9029449-9029449 on Build GRCh38
rs141078293 Chr.X: 9029399-9029399 on Build GRCh38
rs1044752211 Chr.X: 9029284-9029284 on Build GRCh38
rs12839241 Chr.X: 9029395-9029395 on Build GRCh38
rs753904744 Chr.X: 9029428-9029428 on Build GRCh38
rs144948393 Chr.X: 9029370-9029370 on Build GRCh38
rs781075289 Chr.X: 9029270-9029270 on Build GRCh38
rs150312431 Chr.X: 9029355-9029355 on Build GRCh38
rs749163144 Chr.X: 9029458-9029458 on Build GRCh38
rs780102149 Chr.X: 9029450-9029450 on Build GRCh38
rs768719015 Chr.X: 9029377-9029377 on Build GRCh38

View Full Product Details