Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 169518027-169518300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL41
Entrez Gene ID
10324
Gene Name
kelch like family member 41
Gene Aliases
KBTBD10, Krp1, SARCOSIN
UniGene
Hs.50550
Gene Chromosome Location
Chr.2: 169509702-169526262 on Build GRCh38
Transcripts
NM_006063.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3971711 c.1454G>A p.R485H Chr.2: 169518267-169518267 on Build GRCh38
3743310 c.1395G>T p.V465V Chr.2: 169518208-169518208 on Build GRCh38
6088405 c.1472C>T p.A491V Chr.2: 169518285-169518285 on Build GRCh38
374800 c.1455T>A p.R485R Chr.2: 169518268-169518268 on Build GRCh38
4448886 c.1407delC p.G473fs*9 Chr.2: 169518220-169518220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779897118 Chr.2: 169518222-169518222 on Build GRCh38
rs746840461 Chr.2: 169518224-169518224 on Build GRCh38
rs756855395 Chr.2: 169518174-169518174 on Build GRCh38
rs373391330 Chr.2: 169518270-169518270 on Build GRCh38
rs753321870 Chr.2: 169518168-169518168 on Build GRCh38
rs34623017 Chr.2: 169518254-169518254 on Build GRCh38
rs760904619 Chr.2: 169518248-169518248 on Build GRCh38
rs774465298 Chr.2: 169518256-169518256 on Build GRCh38
rs550173333 Chr.2: 169518067-169518067 on Build GRCh38
rs974135846 Chr.2: 169518266-169518266 on Build GRCh38
rs145810515 Chr.2: 169518234-169518234 on Build GRCh38
rs888742117 Chr.2: 169518056-169518056 on Build GRCh38
rs960093414 Chr.2: 169518128-169518128 on Build GRCh38
rs749464576 Chr.2: 169518223-169518224 on Build GRCh38
rs767846716 Chr.2: 169518145-169518145 on Build GRCh38
rs955616481 Chr.2: 169518149-169518149 on Build GRCh38
rs772173940 Chr.2: 169518218-169518218 on Build GRCh38
rs760007708 Chr.2: 169518267-169518267 on Build GRCh38
rs777303663 Chr.2: 169518164-169518164 on Build GRCh38
rs978825115 Chr.2: 169518261-169518261 on Build GRCh38
rs530124379 Chr.2: 169518057-169518057 on Build GRCh38
rs745775130 Chr.2: 169518205-169518205 on Build GRCh38
rs911390630 Chr.2: 169518245-169518245 on Build GRCh38
rs753046029 Chr.2: 169518273-169518273 on Build GRCh38
rs111476553 Chr.2: 169518189-169518189 on Build GRCh38
rs146806979 Chr.2: 169518225-169518225 on Build GRCh38
rs755775814 Chr.2: 169518163-169518163 on Build GRCh38
rs368737545 Chr.2: 169518156-169518156 on Build GRCh38
rs1004374979 Chr.2: 169518094-169518094 on Build GRCh38
rs778940575 Chr.2: 169518203-169518203 on Build GRCh38
rs762980013 Chr.2: 169518251-169518251 on Build GRCh38
rs13012306 Chr.2: 169518097-169518097 on Build GRCh38
rs755289061 Chr.2: 169518140-169518140 on Build GRCh38

View Full Product Details