Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157055300-157055567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAVCR1
Entrez Gene ID
26762
Gene Name
hepatitis A virus cellular receptor 1
Gene Aliases
CD365, HAVCR, HAVCR-1, KIM-1, KIM1, TIM, TIM-1, TIM1, TIMD-1, TIMD1
UniGene
Hs.129711
Gene Chromosome Location
Chr.5: 156979443-157059119 on Build GRCh38
Transcripts
NM_001173393.2 NM_001308156.1 NM_012206.3 XM_017009339.1 XM_017009340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6677772 c.245A>G p.D82G Chr.5: 157055335-157055335 on Build GRCh38
1683629 c.144_145insAG p.G50fs*42 Chr.5: 157055435-157055436 on Build GRCh38
3827524 c.87T>G p.G29G Chr.5: 157055493-157055493 on Build GRCh38
4689546 c.259G>T p.D87Y Chr.5: 157055321-157055321 on Build GRCh38
5811136 c.126C>A p.V42V Chr.5: 157055454-157055454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780221406 Chr.5: 157055343-157055343 on Build GRCh38
rs756075534 Chr.5: 157055350-157055350 on Build GRCh38
rs750200575 Chr.5: 157055521-157055521 on Build GRCh38
rs199873509 Chr.5: 157055335-157055335 on Build GRCh38
rs541541567 Chr.5: 157055366-157055366 on Build GRCh38
rs111358310 Chr.5: 157055346-157055346 on Build GRCh38
rs191117891 Chr.5: 157055376-157055376 on Build GRCh38
rs745465901 Chr.5: 157055371-157055371 on Build GRCh38
rs764402194 Chr.5: 157055328-157055328 on Build GRCh38
rs753653893 Chr.5: 157055440-157055440 on Build GRCh38
rs985045245 Chr.5: 157055497-157055497 on Build GRCh38
rs749086471 Chr.5: 157055355-157055355 on Build GRCh38
rs868143717 Chr.5: 157055487-157055493 on Build GRCh38
rs763215122 Chr.5: 157055415-157055415 on Build GRCh38
rs778524415 Chr.5: 157055442-157055442 on Build GRCh38
rs774628607 Chr.5: 157055488-157055488 on Build GRCh38
rs759667556 Chr.5: 157055490-157055490 on Build GRCh38
rs141472249 Chr.5: 157055493-157055493 on Build GRCh38
rs527766311 Chr.5: 157055364-157055364 on Build GRCh38
rs756746984 Chr.5: 157055431-157055432 on Build GRCh38
rs778345185 Chr.5: 157055448-157055454 on Build GRCh38
rs201914430 Chr.5: 157055447-157055447 on Build GRCh38
rs769107283 Chr.5: 157055475-157055475 on Build GRCh38
rs750683624 Chr.5: 157055410-157055410 on Build GRCh38
rs141773914 Chr.5: 157055435-157055435 on Build GRCh38
rs755237996 Chr.5: 157055362-157055362 on Build GRCh38
rs770213305 Chr.5: 157055480-157055480 on Build GRCh38
rs776921169 Chr.5: 157055396-157055396 on Build GRCh38
rs2270922 Chr.5: 157055428-157055428 on Build GRCh38
rs1015613765 Chr.5: 157055522-157055522 on Build GRCh38
rs948562287 Chr.5: 157055468-157055468 on Build GRCh38
rs762231462 Chr.5: 157055397-157055397 on Build GRCh38
rs767438942 Chr.5: 157055492-157055492 on Build GRCh38
rs771686148 Chr.5: 157055444-157055444 on Build GRCh38
rs1809941 Chr.5: 157055448-157055448 on Build GRCh38
rs998707188 Chr.5: 157055345-157055345 on Build GRCh38
rs376760776 Chr.5: 157055511-157055511 on Build GRCh38
rs757103136 Chr.5: 157055494-157055494 on Build GRCh38
rs765450007 Chr.5: 157055401-157055401 on Build GRCh38
rs202050012 Chr.5: 157055478-157055478 on Build GRCh38
rs750595548 Chr.5: 157055342-157055342 on Build GRCh38
rs368474218 Chr.5: 157055441-157055441 on Build GRCh38
rs889977270 Chr.5: 157055353-157055353 on Build GRCh38
rs772263173 Chr.5: 157055445-157055446 on Build GRCh38
rs766684661 Chr.5: 157055486-157055486 on Build GRCh38
rs748532028 Chr.5: 157055444-157055444 on Build GRCh38
rs200407035 Chr.5: 157055332-157055332 on Build GRCh38
rs777468511 Chr.5: 157055354-157055354 on Build GRCh38
rs771593977 Chr.5: 157055372-157055372 on Build GRCh38
rs773671804 Chr.5: 157055481-157055481 on Build GRCh38
rs373839023 Chr.5: 157055375-157055375 on Build GRCh38
rs61734029 Chr.5: 157055430-157055430 on Build GRCh38
rs370454823 Chr.5: 157055374-157055374 on Build GRCh38
rs748542797 Chr.5: 157055479-157055479 on Build GRCh38
rs766596791 Chr.5: 157055422-157055422 on Build GRCh38
rs372134091 Chr.5: 157055454-157055454 on Build GRCh38
rs754202847 Chr.5: 157055334-157055334 on Build GRCh38
rs758107413 Chr.5: 157055541-157055541 on Build GRCh38
rs761993425 Chr.5: 157055336-157055336 on Build GRCh38
rs746510936 Chr.5: 157055463-157055463 on Build GRCh38

View Full Product Details