Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147658897-147659122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACP6
Entrez Gene ID
51205
Gene Name
acid phosphatase 6, lysophosphatidic
Gene Aliases
ACPL1, LPAP, PACPL1
UniGene
Hs.562154
Gene Chromosome Location
Chr.1: 147629658-147670543 on Build GRCh38
Transcripts
NM_001323625.1 NM_016361.4 NR_136633.1 NR_136634.1 NR_136635.1 NR_136636.1 XM_011509601.2 XM_017001422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592440 c.480-4delT p.? Chr.1: 147659043-147659043 on Build GRCh38
5900976 c.559+4C>T p.? Chr.1: 147658956-147658956 on Build GRCh38
6592442 c.520C>T p.R174C Chr.1: 147658999-147658999 on Build GRCh38
4762422 c.525T>A p.C175* Chr.1: 147658994-147658994 on Build GRCh38
2121801 c.503G>A p.R168Q Chr.1: 147659016-147659016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782213484 Chr.1: 147659035-147659035 on Build GRCh38
rs782663351 Chr.1: 147659026-147659026 on Build GRCh38
rs933662327 Chr.1: 147659095-147659095 on Build GRCh38
rs782289084 Chr.1: 147659029-147659029 on Build GRCh38
rs782054761 Chr.1: 147659049-147659049 on Build GRCh38
rs368669673 Chr.1: 147658956-147658956 on Build GRCh38
rs782089866 Chr.1: 147658979-147658979 on Build GRCh38
rs782151151 Chr.1: 147658989-147658989 on Build GRCh38
rs200902719 Chr.1: 147659065-147659065 on Build GRCh38
rs370462357 Chr.1: 147658922-147658922 on Build GRCh38
rs371625494 Chr.1: 147659090-147659090 on Build GRCh38
rs782268074 Chr.1: 147658930-147658930 on Build GRCh38
rs782488963 Chr.1: 147658920-147658920 on Build GRCh38
rs782516157 Chr.1: 147659024-147659024 on Build GRCh38
rs782728075 Chr.1: 147659064-147659064 on Build GRCh38
rs587624546 Chr.1: 147659039-147659039 on Build GRCh38
rs377700485 Chr.1: 147659016-147659016 on Build GRCh38
rs201459879 Chr.1: 147659034-147659034 on Build GRCh38
rs782051973 Chr.1: 147659058-147659058 on Build GRCh38
rs978886014 Chr.1: 147659017-147659017 on Build GRCh38
rs781988759 Chr.1: 147659043-147659043 on Build GRCh38
rs11800736 Chr.1: 147658988-147658988 on Build GRCh38
rs782034885 Chr.1: 147658974-147658975 on Build GRCh38
rs782479679 Chr.1: 147658999-147658999 on Build GRCh38
rs782410212 Chr.1: 147658942-147658942 on Build GRCh38
rs782758442 Chr.1: 147658982-147658982 on Build GRCh38
rs782107178 Chr.1: 147659081-147659081 on Build GRCh38
rs782429206 Chr.1: 147659031-147659031 on Build GRCh38
rs782580992 Chr.1: 147659042-147659043 on Build GRCh38
rs112983319 Chr.1: 147659060-147659060 on Build GRCh38
rs782262771 Chr.1: 147659046-147659046 on Build GRCh38
rs782042720 Chr.1: 147658954-147658954 on Build GRCh38
rs782042918 Chr.1: 147658955-147658955 on Build GRCh38
rs782037463 Chr.1: 147659051-147659051 on Build GRCh38
rs781831832 Chr.1: 147658952-147658952 on Build GRCh38
rs781949098 Chr.1: 147658964-147658964 on Build GRCh38

View Full Product Details