Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 65595652-65595921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VWA9
Entrez Gene ID
81556
Gene Name
von Willebrand factor A domain containing 9
Gene Aliases
C15orf44
UniGene
Hs.6686
Gene Chromosome Location
Chr.15: 65578757-65611289 on Build GRCh38
Transcripts
NM_001136043.2 NM_001207058.1 NM_001207059.1 NR_045104.1 NR_045105.2 NR_045106.2 NR_045901.1 XM_006720708.3 XM_006720709.3 XM_006720711.2 XM_011522101.2 XM_017022644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6439836 c.949+1G>A p.? Chr.15: 65595732-65595732 on Build GRCh38
6439834 c.841+1G>A p.? Chr.15: 65595732-65595732 on Build GRCh38
4840183 c.876T>G p.F292L Chr.15: 65595806-65595806 on Build GRCh38
6439835 c.670+1G>A p.? Chr.15: 65595732-65595732 on Build GRCh38
1708317 c.622C>T p.P208S Chr.15: 65595781-65595781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79757744 Chr.15: 65595885-65595885 on Build GRCh38
rs956319298 Chr.15: 65595742-65595742 on Build GRCh38
rs977029350 Chr.15: 65595746-65595746 on Build GRCh38
rs754048264 Chr.15: 65595770-65595770 on Build GRCh38
rs758402314 Chr.15: 65595712-65595712 on Build GRCh38
rs374858128 Chr.15: 65595853-65595853 on Build GRCh38
rs201859068 Chr.15: 65595700-65595700 on Build GRCh38
rs761091594 Chr.15: 65595713-65595713 on Build GRCh38
rs919711376 Chr.15: 65595883-65595883 on Build GRCh38
rs768393721 Chr.15: 65595689-65595689 on Build GRCh38
rs780991986 Chr.15: 65595874-65595874 on Build GRCh38
rs747799896 Chr.15: 65595694-65595694 on Build GRCh38
rs762422732 Chr.15: 65595709-65595709 on Build GRCh38
rs746812910 Chr.15: 65595687-65595687 on Build GRCh38
rs757586749 Chr.15: 65595797-65595797 on Build GRCh38
rs1037398913 Chr.15: 65595895-65595895 on Build GRCh38
rs539073146 Chr.15: 65595730-65595730 on Build GRCh38
rs762251644 Chr.15: 65595864-65595864 on Build GRCh38
rs764567176 Chr.15: 65595757-65595757 on Build GRCh38
rs922795540 Chr.15: 65595814-65595814 on Build GRCh38
rs561251013 Chr.15: 65595878-65595878 on Build GRCh38
rs750484598 Chr.15: 65595846-65595846 on Build GRCh38
rs182490064 Chr.15: 65595851-65595851 on Build GRCh38
rs558803935 Chr.15: 65595821-65595821 on Build GRCh38
rs369042994 Chr.15: 65595693-65595693 on Build GRCh38
rs371744375 Chr.15: 65595710-65595710 on Build GRCh38
rs772727774 Chr.15: 65595701-65595701 on Build GRCh38

View Full Product Details