Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1455371-1455604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APC2
Entrez Gene ID
10297
Gene Name
APC2, WNT signaling pathway regulator
Gene Aliases
APCL
UniGene
Hs.446376
Gene Chromosome Location
Chr.19: 1446268-1473244 on Build GRCh38
Transcripts
NM_005883.2 XM_005259475.2 XM_006722607.2 XM_006722608.3 XM_006722609.3 XM_006722610.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5149763 c.608G>A p.G203D Chr.19: 1455469-1455469 on Build GRCh38
5966681 c.567_596del30 p.F189_E199>L Chr.19: 1455428-1455457 on Build GRCh38
6311429 c.597G>T p.E199D Chr.19: 1455458-1455458 on Build GRCh38
4074809 c.602G>A p.R201H Chr.19: 1455463-1455463 on Build GRCh38
3668186 c.564G>A p.E188E Chr.19: 1455425-1455425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999076071 Chr.19: 1455526-1455526 on Build GRCh38
rs1041822776 Chr.19: 1455459-1455459 on Build GRCh38
rs532831165 Chr.19: 1455522-1455522 on Build GRCh38
rs776575180 Chr.19: 1455407-1455407 on Build GRCh38
rs748343600 Chr.19: 1455420-1455420 on Build GRCh38
rs767507812 Chr.19: 1455501-1455501 on Build GRCh38
rs565322444 Chr.19: 1455500-1455500 on Build GRCh38
rs750200449 Chr.19: 1455480-1455480 on Build GRCh38
rs1054103388 Chr.19: 1455477-1455477 on Build GRCh38
rs889188449 Chr.19: 1455406-1455406 on Build GRCh38
rs868809340 Chr.19: 1455476-1455476 on Build GRCh38
rs1022692947 Chr.19: 1455577-1455577 on Build GRCh38
rs924312521 Chr.19: 1455475-1455475 on Build GRCh38
rs764839509 Chr.19: 1455479-1455479 on Build GRCh38
rs754881860 Chr.19: 1455508-1455508 on Build GRCh38
rs760108808 Chr.19: 1455539-1455539 on Build GRCh38
rs371007058 Chr.19: 1455541-1455541 on Build GRCh38
rs756934450 Chr.19: 1455468-1455468 on Build GRCh38
rs747578381 Chr.19: 1455512-1455531 on Build GRCh38
rs748242653 Chr.19: 1455514-1455514 on Build GRCh38
rs766873657 Chr.19: 1455448-1455448 on Build GRCh38
rs774869820 Chr.19: 1455533-1455533 on Build GRCh38
rs768678791 Chr.19: 1455391-1455391 on Build GRCh38
rs894757775 Chr.19: 1455486-1455486 on Build GRCh38
rs374545598 Chr.19: 1455429-1455429 on Build GRCh38
rs753350102 Chr.19: 1455467-1455467 on Build GRCh38
rs781318098 Chr.19: 1455511-1455511 on Build GRCh38
rs774961503 Chr.19: 1455452-1455452 on Build GRCh38
rs984205559 Chr.19: 1455580-1455580 on Build GRCh38
rs149886452 Chr.19: 1455425-1455425 on Build GRCh38
rs760125235 Chr.19: 1455453-1455453 on Build GRCh38
rs911708482 Chr.19: 1455397-1455397 on Build GRCh38
rs761282035 Chr.19: 1455549-1455549 on Build GRCh38
rs770025331 Chr.19: 1455518-1455518 on Build GRCh38
rs551157653 Chr.19: 1455575-1455575 on Build GRCh38
rs969755090 Chr.19: 1455579-1455579 on Build GRCh38
rs776179137 Chr.19: 1455546-1455546 on Build GRCh38
rs746968102 Chr.19: 1455389-1455389 on Build GRCh38
rs771356731 Chr.19: 1455531-1455531 on Build GRCh38
rs751428377 Chr.19: 1455505-1455505 on Build GRCh38
rs777904669 Chr.19: 1455523-1455523 on Build GRCh38
rs368519913 Chr.19: 1455492-1455492 on Build GRCh38
rs763602620 Chr.19: 1455458-1455458 on Build GRCh38
rs992942666 Chr.19: 1455545-1455545 on Build GRCh38
rs749554280 Chr.19: 1455528-1455528 on Build GRCh38
rs771658842 Chr.19: 1455514-1455515 on Build GRCh38

View Full Product Details