Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22098398-22098669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM160B2
Entrez Gene ID
64760
Gene Name
family with sequence similarity 160 member B2
Gene Aliases
RAI16
UniGene
Hs.491223
Gene Chromosome Location
Chr.8: 22089150-22104380 on Build GRCh38
Transcripts
NM_022749.5 XM_005273602.3 XM_006716381.2 XM_006716385.3 XM_017013732.1 XM_017013733.1 XM_017013734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658733 c.885G>A p.R295R Chr.8: 22098539-22098539 on Build GRCh38
454296 c.943A>G p.T315A Chr.8: 22098597-22098597 on Build GRCh38
6658735 c.875C>T p.A292V Chr.8: 22098529-22098529 on Build GRCh38
5011886 c.769A>G p.K257E Chr.8: 22098423-22098423 on Build GRCh38
6658736 c.56C>T p.A19V Chr.8: 22098529-22098529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35497596 Chr.8: 22098597-22098597 on Build GRCh38
rs746585142 Chr.8: 22098626-22098626 on Build GRCh38
rs755238365 Chr.8: 22098530-22098530 on Build GRCh38
rs770258006 Chr.8: 22098459-22098459 on Build GRCh38
rs775182401 Chr.8: 22098539-22098539 on Build GRCh38
rs776227768 Chr.8: 22098634-22098634 on Build GRCh38
rs371326435 Chr.8: 22098542-22098542 on Build GRCh38
rs1046784013 Chr.8: 22098508-22098508 on Build GRCh38
rs375925165 Chr.8: 22098565-22098565 on Build GRCh38
rs979854830 Chr.8: 22098592-22098592 on Build GRCh38
rs907759990 Chr.8: 22098452-22098452 on Build GRCh38
rs370872823 Chr.8: 22098494-22098494 on Build GRCh38
rs778228732 Chr.8: 22098596-22098596 on Build GRCh38
rs763809183 Chr.8: 22098512-22098512 on Build GRCh38
rs199670542 Chr.8: 22098635-22098635 on Build GRCh38
rs762986197 Chr.8: 22098560-22098560 on Build GRCh38
rs755041962 Chr.8: 22098575-22098575 on Build GRCh38
rs1002325765 Chr.8: 22098619-22098619 on Build GRCh38
rs750381771 Chr.8: 22098517-22098517 on Build GRCh38
rs758214577 Chr.8: 22098521-22098521 on Build GRCh38
rs780038902 Chr.8: 22098557-22098557 on Build GRCh38
rs767518854 Chr.8: 22098584-22098584 on Build GRCh38
rs375499512 Chr.8: 22098528-22098528 on Build GRCh38
rs140882816 Chr.8: 22098434-22098434 on Build GRCh38
rs200628638 Chr.8: 22098544-22098544 on Build GRCh38
rs375312941 Chr.8: 22098504-22098504 on Build GRCh38
rs779550433 Chr.8: 22098607-22098607 on Build GRCh38
rs200724432 Chr.8: 22098537-22098537 on Build GRCh38
rs150124585 Chr.8: 22098516-22098516 on Build GRCh38
rs974430189 Chr.8: 22098467-22098467 on Build GRCh38
rs369101347 Chr.8: 22098643-22098643 on Build GRCh38
rs180833096 Chr.8: 22098507-22098507 on Build GRCh38
rs751692783 Chr.8: 22098568-22098568 on Build GRCh38
rs201631142 Chr.8: 22098448-22098448 on Build GRCh38
rs751604937 Chr.8: 22098529-22098529 on Build GRCh38
rs919587882 Chr.8: 22098496-22098496 on Build GRCh38
rs761308865 Chr.8: 22098551-22098551 on Build GRCh38
rs386412283 Chr.8: 22098543-22098544 on Build GRCh38
rs756241025 Chr.8: 22098590-22098590 on Build GRCh38
rs200751092 Chr.8: 22098430-22098430 on Build GRCh38
rs373414985 Chr.8: 22098543-22098543 on Build GRCh38
rs368951510 Chr.8: 22098534-22098534 on Build GRCh38
rs372291105 Chr.8: 22098547-22098547 on Build GRCh38
rs577524598 Chr.8: 22098644-22098644 on Build GRCh38
rs1039846082 Chr.8: 22098549-22098549 on Build GRCh38
rs145516741 Chr.8: 22098585-22098585 on Build GRCh38
rs746497348 Chr.8: 22098542-22098542 on Build GRCh38
rs367902857 Chr.8: 22098510-22098510 on Build GRCh38
rs771623816 Chr.8: 22098499-22098499 on Build GRCh38
rs781574018 Chr.8: 22098436-22098436 on Build GRCh38
rs536224294 Chr.8: 22098559-22098559 on Build GRCh38
rs1014438064 Chr.8: 22098576-22098576 on Build GRCh38
rs538304467 Chr.8: 22098503-22098503 on Build GRCh38
rs757832979 Chr.8: 22098601-22098601 on Build GRCh38
rs781375678 Chr.8: 22098533-22098533 on Build GRCh38
rs771300488 Chr.8: 22098538-22098538 on Build GRCh38
rs773764902 Chr.8: 22098491-22098491 on Build GRCh38
rs776617525 Chr.8: 22098515-22098515 on Build GRCh38
rs776419111 Chr.8: 22098546-22098546 on Build GRCh38
rs200199022 Chr.8: 22098543-22098543 on Build GRCh38
rs768290456 Chr.8: 22098632-22098632 on Build GRCh38

View Full Product Details