Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43292517-43292754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGCS1
Entrez Gene ID
3157
Gene Name
3-hydroxy-3-methylglutaryl-CoA synthase 1
Gene Aliases
HMGCS
UniGene
Hs.397729
Gene Chromosome Location
Chr.5: 43287470-43313512 on Build GRCh38
Transcripts
NM_001098272.2 NM_001324219.1 NM_001324220.1 NM_001324222.1 NM_001324223.1 NM_001324224.1 NM_002130.7 XM_011514036.2 XM_017009412.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1671627 c.1343T>C p.L448P Chr.5: 43292604-43292604 on Build GRCh38
4464208 c.1322C>T p.P441L Chr.5: 43292625-43292625 on Build GRCh38
5525055 c.1329T>C p.G443G Chr.5: 43292618-43292618 on Build GRCh38
7134125 c.1388G>A p.R463K Chr.5: 43292559-43292559 on Build GRCh38
1068402 c.1355C>T p.T452M Chr.5: 43292592-43292592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758162734 Chr.5: 43292541-43292541 on Build GRCh38
rs774791910 Chr.5: 43292621-43292621 on Build GRCh38
rs767448406 Chr.5: 43292679-43292679 on Build GRCh38
rs764841594 Chr.5: 43292652-43292652 on Build GRCh38
rs748966555 Chr.5: 43292640-43292644 on Build GRCh38
rs968424099 Chr.5: 43292644-43292644 on Build GRCh38
rs376783581 Chr.5: 43292643-43292643 on Build GRCh38
rs757117917 Chr.5: 43292549-43292549 on Build GRCh38
rs370467427 Chr.5: 43292544-43292544 on Build GRCh38
rs768391982 Chr.5: 43292700-43292700 on Build GRCh38
rs987978854 Chr.5: 43292639-43292643 on Build GRCh38
rs781144120 Chr.5: 43292578-43292578 on Build GRCh38
rs9292869 Chr.5: 43292706-43292706 on Build GRCh38
rs759059766 Chr.5: 43292650-43292650 on Build GRCh38
rs762425498 Chr.5: 43292634-43292634 on Build GRCh38
rs757207769 Chr.5: 43292675-43292675 on Build GRCh38
rs776039052 Chr.5: 43292648-43292648 on Build GRCh38
rs141436337 Chr.5: 43292614-43292614 on Build GRCh38
rs371014387 Chr.5: 43292665-43292665 on Build GRCh38
rs751491785 Chr.5: 43292548-43292548 on Build GRCh38
rs769940444 Chr.5: 43292599-43292599 on Build GRCh38
rs751295681 Chr.5: 43292673-43292673 on Build GRCh38
rs369661164 Chr.5: 43292680-43292680 on Build GRCh38
rs780292296 Chr.5: 43292601-43292601 on Build GRCh38
rs763878991 Chr.5: 43292666-43292666 on Build GRCh38
rs749729014 Chr.5: 43292610-43292610 on Build GRCh38
rs769138555 Chr.5: 43292613-43292613 on Build GRCh38
rs752377585 Chr.5: 43292654-43292654 on Build GRCh38
rs956543626 Chr.5: 43292607-43292607 on Build GRCh38
rs745911220 Chr.5: 43292586-43292586 on Build GRCh38
rs770427434 Chr.5: 43292678-43292678 on Build GRCh38

View Full Product Details