Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54430202-54430469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNIH1
Entrez Gene ID
10175
Gene Name
cornichon family AMPA receptor auxiliary protein 1
Gene Aliases
CNIH, CNIH-1, CNIL, TGAM77
UniGene
Hs.294603
Gene Chromosome Location
Chr.14: 54426929-54441430 on Build GRCh38
Transcripts
NM_005776.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
433140 c.407+2T>G p.? Chr.14: 54430259-54430259 on Build GRCh38
1201536 c.390T>G p.F130L Chr.14: 54430278-54430278 on Build GRCh38
6076086 c.407+1G>C p.? Chr.14: 54430260-54430260 on Build GRCh38
1370133 c.394delT p.Y132fs*6 Chr.14: 54430274-54430274 on Build GRCh38
1370132 c.400C>A p.L134I Chr.14: 54430268-54430268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889135113 Chr.14: 54430383-54430383 on Build GRCh38
rs778660936 Chr.14: 54430226-54430226 on Build GRCh38
rs771960166 Chr.14: 54430324-54430324 on Build GRCh38
rs775504716 Chr.14: 54430334-54430334 on Build GRCh38
rs756633702 Chr.14: 54430440-54430440 on Build GRCh38
rs999675806 Chr.14: 54430266-54430266 on Build GRCh38
rs768199347 Chr.14: 54430350-54430350 on Build GRCh38
rs753969883 Chr.14: 54430251-54430260 on Build GRCh38
rs1040379372 Chr.14: 54430439-54430439 on Build GRCh38
rs1051082918 Chr.14: 54430247-54430247 on Build GRCh38
rs774806273 Chr.14: 54430411-54430411 on Build GRCh38
rs761526019 Chr.14: 54430354-54430354 on Build GRCh38
rs890251946 Chr.14: 54430234-54430234 on Build GRCh38
rs11552805 Chr.14: 54430361-54430361 on Build GRCh38
rs11552803 Chr.14: 54430382-54430382 on Build GRCh38
rs776071416 Chr.14: 54430352-54430352 on Build GRCh38
rs753256646 Chr.14: 54430425-54430425 on Build GRCh38
rs765028966 Chr.14: 54430355-54430355 on Build GRCh38
rs1051464074 Chr.14: 54430233-54430233 on Build GRCh38
rs746446099 Chr.14: 54430340-54430340 on Build GRCh38
rs368293767 Chr.14: 54430245-54430245 on Build GRCh38
rs762093840 Chr.14: 54430337-54430337 on Build GRCh38
rs537064406 Chr.14: 54430419-54430419 on Build GRCh38
rs768067762 Chr.14: 54430421-54430421 on Build GRCh38
rs763188121 Chr.14: 54430391-54430391 on Build GRCh38
rs1030696897 Chr.14: 54430405-54430405 on Build GRCh38
rs915390079 Chr.14: 54430240-54430240 on Build GRCh38
rs537569161 Chr.14: 54430286-54430286 on Build GRCh38

View Full Product Details