Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145454494-145454245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1B
Entrez Gene ID
774
Gene Name
calcium voltage-gated channel subunit alpha1 B
Gene Aliases
BIII, CACNL1A5, CACNN, Cav2.2, DYT23
UniGene
Hs.495522
Gene Chromosome Location
Chr.9: 137877789-138124624 on Build GRCh38
Transcripts
NM_000718.3 NM_001243812.1 XM_011518991.2 XM_011518992.2 XM_011518993.2 XM_011518994.2 XM_017015115.1 XM_017015116.1 XM_017015117.1 XM_017015118.1 XM_017015119.1 XM_017015120.1 NR_121583.1

Gene Symbol
LOC100133077
Entrez Gene ID
100133077
Gene Name
uncharacterized LOC100133077
Gene Aliases
-
UniGene
Hs.640081
Gene Chromosome Location
Chr.9: 137867925-137892570 on Build GRCh38
Transcripts
NM_000718.3 NM_001243812.1 XM_011518991.2 XM_011518992.2 XM_011518993.2 XM_011518994.2 XM_017015115.1 XM_017015116.1 XM_017015117.1 XM_017015118.1 XM_017015119.1 XM_017015120.1 NR_121583.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
404111 c.60G>T p.R20R Chr.9: 137877993-137877993 on Build GRCh38
4163655 c.49G>A p.G17S Chr.9: 137877982-137877982 on Build GRCh38
6028082 c.103delG p.L37fs*64 Chr.9: 137878036-137878036 on Build GRCh38
753009 c.143C>A p.S48* Chr.9: 137878076-137878076 on Build GRCh38
404629 c.59G>T p.R20L Chr.9: 137877992-137877992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201645191 Chr.9: 137878149-137878149 on Build GRCh38
rs200843832 Chr.9: 137878143-137878143 on Build GRCh38
rs199833027 Chr.9: 137878111-137878111 on Build GRCh38
rs760728160 Chr.9: 137878171-137878171 on Build GRCh38
rs759633664 Chr.9: 137878050-137878051 on Build GRCh38
rs775276054 Chr.9: 137878041-137878041 on Build GRCh38
rs746177646 Chr.9: 137878032-137878032 on Build GRCh38
rs376286287 Chr.9: 137878144-137878144 on Build GRCh38
rs201338669 Chr.9: 137878100-137878100 on Build GRCh38
rs760479029 Chr.9: 137878082-137878082 on Build GRCh38
rs753538003 Chr.9: 137878130-137878130 on Build GRCh38
rs139719558 Chr.9: 137878161-137878161 on Build GRCh38
rs769445298 Chr.9: 137878033-137878033 on Build GRCh38
rs762496265 Chr.9: 137878050-137878050 on Build GRCh38
rs199588193 Chr.9: 137878122-137878122 on Build GRCh38
rs200515483 Chr.9: 137878125-137878125 on Build GRCh38
rs771028331 Chr.9: 137878014-137878015 on Build GRCh38
rs377621007 Chr.9: 137878112-137878112 on Build GRCh38
rs750256540 Chr.9: 137878081-137878081 on Build GRCh38
rs201813576 Chr.9: 137878094-137878094 on Build GRCh38
rs200276036 Chr.9: 137878095-137878095 on Build GRCh38
rs772818413 Chr.9: 137878162-137878162 on Build GRCh38
rs763438014 Chr.9: 137878175-137878176 on Build GRCh38
rs757778139 Chr.9: 137878104-137878104 on Build GRCh38
rs780495002 Chr.9: 137878157-137878157 on Build GRCh38
rs771390024 Chr.9: 137878026-137878026 on Build GRCh38
rs763706875 Chr.9: 137878059-137878059 on Build GRCh38
rs201591503 Chr.9: 137878116-137878116 on Build GRCh38
rs769799716 Chr.9: 137878097-137878098 on Build GRCh38
rs749565808 Chr.9: 137878158-137878158 on Build GRCh38
rs768150608 Chr.9: 137878121-137878121 on Build GRCh38
rs201870839 Chr.9: 137878128-137878128 on Build GRCh38
rs772139423 Chr.9: 137878123-137878123 on Build GRCh38
rs776819631 Chr.9: 137878019-137878020 on Build GRCh38
rs781231804 Chr.9: 137878152-137878152 on Build GRCh38
rs773341028 Chr.9: 137878127-137878127 on Build GRCh38
rs780255192 Chr.9: 137878113-137878113 on Build GRCh38
rs761871500 Chr.9: 137878074-137878074 on Build GRCh38
rs201253748 Chr.9: 137878025-137878025 on Build GRCh38
rs191962352 Chr.9: 137877990-137877990 on Build GRCh38
rs199731221 Chr.9: 137878132-137878132 on Build GRCh38
rs746373724 Chr.9: 137878110-137878110 on Build GRCh38
rs199860587 Chr.9: 137878155-137878155 on Build GRCh38
rs749007898 Chr.9: 137878120-137878120 on Build GRCh38
rs775992694 Chr.9: 137878154-137878154 on Build GRCh38
rs765081336 Chr.9: 137878179-137878179 on Build GRCh38
rs766238445 Chr.9: 137878129-137878129 on Build GRCh38
rs777695397 Chr.9: 137878020-137878020 on Build GRCh38
rs187204220 Chr.9: 137877982-137877982 on Build GRCh38
rs773700895 Chr.9: 137878063-137878063 on Build GRCh38
rs758519804 Chr.9: 137878098-137878098 on Build GRCh38
rs748315878 Chr.9: 137877996-137877996 on Build GRCh38
rs758563303 Chr.9: 137878012-137878012 on Build GRCh38
rs757397831 Chr.9: 137878151-137878151 on Build GRCh38
rs753013232 Chr.9: 137878093-137878093 on Build GRCh38
rs781617028 Chr.9: 137878105-137878105 on Build GRCh38
rs768366699 Chr.9: 137878159-137878159 on Build GRCh38
rs777004745 Chr.9: 137878028-137878028 on Build GRCh38
rs949782913 Chr.9: 137877997-137877997 on Build GRCh38
rs766075787 Chr.9: 137878083-137878083 on Build GRCh38
rs767511286 Chr.9: 137878076-137878076 on Build GRCh38
rs750201374 Chr.9: 137877981-137877981 on Build GRCh38
rs759189262 Chr.9: 137878177-137878177 on Build GRCh38
rs201529170 Chr.9: 137878174-137878174 on Build GRCh38
rs370377726 Chr.9: 137878156-137878156 on Build GRCh38
rs200803427 Chr.9: 137878115-137878115 on Build GRCh38

View Full Product Details