Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70787596-70787851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIGLA
Entrez Gene ID
344018
Gene Name
folliculogenesis specific bHLH transcription factor
Gene Aliases
BHLHC8, FIGALPHA, POF6
UniGene
Hs.407636
Gene Chromosome Location
Chr.2: 70777310-70790643 on Build GRCh38
Transcripts
NM_001004311.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5002361 c.245A>G p.N82S Chr.2: 70787788-70787788 on Build GRCh38
3839918 c.371C>A p.A124D Chr.2: 70787662-70787662 on Build GRCh38
5650352 c.334G>A p.E112K Chr.2: 70787699-70787699 on Build GRCh38
1153847 c.305G>A p.S102N Chr.2: 70787728-70787728 on Build GRCh38
6509448 c.283C>T p.L95F Chr.2: 70787750-70787750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779683613 Chr.2: 70787703-70787703 on Build GRCh38
rs919463905 Chr.2: 70787687-70787687 on Build GRCh38
rs782356078 Chr.2: 70787740-70787740 on Build GRCh38
rs782103382 Chr.2: 70787671-70787671 on Build GRCh38
rs782504042 Chr.2: 70787796-70787796 on Build GRCh38
rs886056271 Chr.2: 70787638-70787638 on Build GRCh38
rs782450302 Chr.2: 70787690-70787690 on Build GRCh38
rs782723662 Chr.2: 70787630-70787630 on Build GRCh38
rs34451883 Chr.2: 70787646-70787647 on Build GRCh38
rs781845895 Chr.2: 70787696-70787696 on Build GRCh38
rs782196840 Chr.2: 70787722-70787722 on Build GRCh38
rs782030456 Chr.2: 70787761-70787761 on Build GRCh38
rs782288538 Chr.2: 70787643-70787643 on Build GRCh38
rs199650129 Chr.2: 70787799-70787799 on Build GRCh38
rs782422846 Chr.2: 70787705-70787705 on Build GRCh38
rs199702150 Chr.2: 70787785-70787785 on Build GRCh38
rs782471267 Chr.2: 70787817-70787817 on Build GRCh38
rs555588922 Chr.2: 70787774-70787774 on Build GRCh38
rs981670974 Chr.2: 70787682-70787682 on Build GRCh38
rs782106232 Chr.2: 70787783-70787783 on Build GRCh38
rs781950015 Chr.2: 70787750-70787750 on Build GRCh38
rs781892008 Chr.2: 70787814-70787814 on Build GRCh38
rs782565660 Chr.2: 70787822-70787822 on Build GRCh38
rs781915110 Chr.2: 70787778-70787778 on Build GRCh38
rs200490298 Chr.2: 70787734-70787734 on Build GRCh38
rs782704018 Chr.2: 70787775-70787775 on Build GRCh38
rs575734669 Chr.2: 70787704-70787704 on Build GRCh38
rs782564869 Chr.2: 70787693-70787693 on Build GRCh38
rs782818820 Chr.2: 70787677-70787677 on Build GRCh38
rs782680717 Chr.2: 70787715-70787715 on Build GRCh38
rs200072751 Chr.2: 70787688-70787688 on Build GRCh38

View Full Product Details