Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 36681385-36681654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHOD3
Entrez Gene ID
80206
Gene Name
formin homology 2 domain containing 3
Gene Aliases
FHOS2, Formactin2
UniGene
Hs.436636
Gene Chromosome Location
Chr.18: 36297696-36780220 on Build GRCh38
Transcripts
NM_001281739.2 NM_001281740.2 NM_025135.4 XM_005258349.1 XM_005258354.1 XM_005258355.1 XM_011526189.1 XM_011526190.1 XM_011526192.1 XM_011526193.2 XM_011526195.1 XM_011526196.1 XM_011526197.1 XM_017026006.1 XM_017026007.1 XM_017026008.1 XM_017026009.1 XM_017026010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
325352 c.1354C>A p.Q452K Chr.18: 36681479-36681479 on Build GRCh38
5916658 c.1330C>T p.L444F Chr.18: 36681455-36681455 on Build GRCh38
6663998 c.1429G>T p.E477* Chr.18: 36681554-36681554 on Build GRCh38
3701361 c.1345A>T p.R449W Chr.18: 36681470-36681470 on Build GRCh38
7152992 c.1337C>T p.S446L Chr.18: 36681462-36681462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141148037 Chr.18: 36681512-36681512 on Build GRCh38
rs768377913 Chr.18: 36681528-36681528 on Build GRCh38
rs780516974 Chr.18: 36681513-36681513 on Build GRCh38
rs965612992 Chr.18: 36681600-36681600 on Build GRCh38
rs774149210 Chr.18: 36681532-36681532 on Build GRCh38
rs79108771 Chr.18: 36681619-36681619 on Build GRCh38
rs200278646 Chr.18: 36681546-36681546 on Build GRCh38
rs752959012 Chr.18: 36681560-36681560 on Build GRCh38
rs370344363 Chr.18: 36681523-36681523 on Build GRCh38
rs201418593 Chr.18: 36681610-36681610 on Build GRCh38
rs202238556 Chr.18: 36681570-36681570 on Build GRCh38
rs151313792 Chr.18: 36681510-36681510 on Build GRCh38
rs778642416 Chr.18: 36681612-36681612 on Build GRCh38
rs144993045 Chr.18: 36681543-36681543 on Build GRCh38
rs765891878 Chr.18: 36681601-36681604 on Build GRCh38
rs781710376 Chr.18: 36681590-36681590 on Build GRCh38
rs772659898 Chr.18: 36681585-36681585 on Build GRCh38
rs372938962 Chr.18: 36681458-36681458 on Build GRCh38
rs750951318 Chr.18: 36681595-36681595 on Build GRCh38
rs763417828 Chr.18: 36681415-36681416 on Build GRCh38
rs781419029 Chr.18: 36681549-36681549 on Build GRCh38
rs568370110 Chr.18: 36681579-36681579 on Build GRCh38
rs758818742 Chr.18: 36681509-36681509 on Build GRCh38
rs759635243 Chr.18: 36681485-36681485 on Build GRCh38
rs142462042 Chr.18: 36681469-36681469 on Build GRCh38
rs780754499 Chr.18: 36681430-36681430 on Build GRCh38
rs772079874 Chr.18: 36681547-36681547 on Build GRCh38
rs765465359 Chr.18: 36681492-36681492 on Build GRCh38
rs533511296 Chr.18: 36681466-36681466 on Build GRCh38
rs9964535 Chr.18: 36681548-36681548 on Build GRCh38
rs542241 Chr.18: 36681596-36681596 on Build GRCh38
rs370680795 Chr.18: 36681412-36681412 on Build GRCh38
rs769238069 Chr.18: 36681517-36681517 on Build GRCh38
rs142042172 Chr.18: 36681558-36681558 on Build GRCh38
rs770557225 Chr.18: 36681428-36681428 on Build GRCh38
rs779743857 Chr.18: 36681520-36681520 on Build GRCh38
rs956451393 Chr.18: 36681588-36681589 on Build GRCh38
rs751992485 Chr.18: 36681563-36681563 on Build GRCh38
rs745353976 Chr.18: 36681514-36681514 on Build GRCh38
rs368544178 Chr.18: 36681562-36681562 on Build GRCh38
rs34100890 Chr.18: 36681619-36681619 on Build GRCh38
rs779434946 Chr.18: 36681603-36681603 on Build GRCh38
rs771809219 Chr.18: 36681616-36681616 on Build GRCh38
rs144071785 Chr.18: 36681496-36681497 on Build GRCh38
rs112727295 Chr.18: 36681614-36681614 on Build GRCh38
rs148807316 Chr.18: 36681445-36681445 on Build GRCh38
rs201006691 Chr.18: 36681483-36681483 on Build GRCh38
rs753014109 Chr.18: 36681495-36681495 on Build GRCh38
rs763272073 Chr.18: 36681561-36681561 on Build GRCh38
rs550295372 Chr.18: 36681467-36681467 on Build GRCh38
rs199579476 Chr.18: 36681444-36681444 on Build GRCh38
rs374168306 Chr.18: 36681589-36681589 on Build GRCh38
rs775047283 Chr.18: 36681450-36681450 on Build GRCh38
rs748885882 Chr.18: 36681609-36681609 on Build GRCh38
rs774271898 Chr.18: 36681460-36681460 on Build GRCh38
rs909335271 Chr.18: 36681465-36681465 on Build GRCh38

View Full Product Details