Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 118843121-118843346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EN1
Entrez Gene ID
2019
Gene Name
engrailed homeobox 1
Gene Aliases
-
UniGene
Hs.271977
Gene Chromosome Location
Chr.2: 118842171-118848183 on Build GRCh38
Transcripts
NM_001426.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4803280 c.921G>A p.R307R Chr.2: 118843196-118843196 on Build GRCh38
3041081 c.925G>A p.A309T Chr.2: 118843192-118843192 on Build GRCh38
4976792 c.866C>T p.P289L Chr.2: 118843251-118843251 on Build GRCh38
7235707 c.968A>C p.Q323P Chr.2: 118843149-118843149 on Build GRCh38
6153594 c.907G>C p.D303H Chr.2: 118843210-118843210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150977527 Chr.2: 118843205-118843205 on Build GRCh38
rs538621981 Chr.2: 118843314-118843314 on Build GRCh38
rs796778743 Chr.2: 118843270-118843270 on Build GRCh38
rs767480596 Chr.2: 118843217-118843217 on Build GRCh38
rs972943886 Chr.2: 118843278-118843278 on Build GRCh38
rs80156392 Chr.2: 118843280-118843280 on Build GRCh38
rs538761075 Chr.2: 118843288-118843288 on Build GRCh38
rs758612305 Chr.2: 118843202-118843202 on Build GRCh38
rs750658689 Chr.2: 118843275-118843275 on Build GRCh38
rs770636668 Chr.2: 118843184-118843184 on Build GRCh38
rs959285828 Chr.2: 118843279-118843279 on Build GRCh38
rs1032594414 Chr.2: 118843274-118843274 on Build GRCh38
rs751701618 Chr.2: 118843221-118843223 on Build GRCh38
rs758603688 Chr.2: 118843287-118843287 on Build GRCh38
rs866644300 Chr.2: 118843319-118843319 on Build GRCh38
rs949014270 Chr.2: 118843311-118843311 on Build GRCh38
rs764066167 Chr.2: 118843258-118843258 on Build GRCh38
rs990749062 Chr.2: 118843284-118843284 on Build GRCh38
rs1008649030 Chr.2: 118843236-118843236 on Build GRCh38
rs186026467 Chr.2: 118843313-118843313 on Build GRCh38
rs947622399 Chr.2: 118843303-118843303 on Build GRCh38
rs398088817 Chr.2: 118843281-118843282 on Build GRCh38
rs928735852 Chr.2: 118843275-118843275 on Build GRCh38
rs918804318 Chr.2: 118843324-118843324 on Build GRCh38
rs962054009 Chr.2: 118843220-118843221 on Build GRCh38
rs748684153 Chr.2: 118843175-118843175 on Build GRCh38
rs181999906 Chr.2: 118843312-118843312 on Build GRCh38
rs747307197 Chr.2: 118843298-118843298 on Build GRCh38
rs901600851 Chr.2: 118843317-118843317 on Build GRCh38
rs552946276 Chr.2: 118843292-118843292 on Build GRCh38
rs571122176 Chr.2: 118843268-118843268 on Build GRCh38
rs780372731 Chr.2: 118843148-118843148 on Build GRCh38
rs139728151 Chr.2: 118843193-118843193 on Build GRCh38
rs879079590 Chr.2: 118843199-118843199 on Build GRCh38
rs370594131 Chr.2: 118843154-118843154 on Build GRCh38
rs375974616 Chr.2: 118843237-118843237 on Build GRCh38
rs757246228 Chr.2: 118843266-118843266 on Build GRCh38
rs546644099 Chr.2: 118843261-118843261 on Build GRCh38
rs145326362 Chr.2: 118843167-118843167 on Build GRCh38
rs369801550 Chr.2: 118843220-118843220 on Build GRCh38
rs769146878 Chr.2: 118843161-118843161 on Build GRCh38
rs917565466 Chr.2: 118843290-118843290 on Build GRCh38
rs904460049 Chr.2: 118843157-118843157 on Build GRCh38
rs760696157 Chr.2: 118843250-118843250 on Build GRCh38

View Full Product Details