Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 580236-580463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSG
Entrez Gene ID
682
Gene Name
basigin (Ok blood group)
Gene Aliases
5F7, CD147, EMMPRIN, OK, TCSF
UniGene
Hs.501293
Gene Chromosome Location
Chr.19: 571277-583493 on Build GRCh38
Transcripts
NM_001322243.1 NM_001728.3 NM_198589.2 NM_198590.2 NM_198591.2 XM_017027173.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7216305 c.42C>G p.S14S Chr.19: 580388-580388 on Build GRCh38
1246270 c.114C>T p.H38H Chr.19: 580460-580460 on Build GRCh38
7174400 c.112C>T p.H38Y Chr.19: 580458-580458 on Build GRCh38
5390969 c.75C>T p.F25F Chr.19: 580421-580421 on Build GRCh38
4391151 c.645C>T p.I215I Chr.19: 580451-580451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757662028 Chr.19: 580439-580439 on Build GRCh38
rs963123686 Chr.19: 580304-580304 on Build GRCh38
rs2108832 Chr.19: 580268-580268 on Build GRCh38
rs967163989 Chr.19: 580435-580435 on Build GRCh38
rs764012546 Chr.19: 580332-580332 on Build GRCh38
rs765327178 Chr.19: 580356-580357 on Build GRCh38
rs567629506 Chr.19: 580441-580441 on Build GRCh38
rs767247412 Chr.19: 580404-580404 on Build GRCh38
rs373209745 Chr.19: 580354-580354 on Build GRCh38
rs541295754 Chr.19: 580317-580317 on Build GRCh38
rs954434934 Chr.19: 580266-580266 on Build GRCh38
rs758686821 Chr.19: 580352-580352 on Build GRCh38
rs11879069 Chr.19: 580333-580333 on Build GRCh38
rs140722203 Chr.19: 580397-580397 on Build GRCh38
rs189460104 Chr.19: 580353-580353 on Build GRCh38
rs1055768994 Chr.19: 580319-580319 on Build GRCh38
rs762638776 Chr.19: 580331-580331 on Build GRCh38
rs554979181 Chr.19: 580298-580298 on Build GRCh38
rs11551909 Chr.19: 580445-580445 on Build GRCh38
rs113465218 Chr.19: 580324-580324 on Build GRCh38
rs566889863 Chr.19: 580373-580373 on Build GRCh38
rs879266841 Chr.19: 580316-580316 on Build GRCh38
rs753080156 Chr.19: 580424-580424 on Build GRCh38
rs767084325 Chr.19: 580346-580346 on Build GRCh38
rs1013953044 Chr.19: 580262-580262 on Build GRCh38
rs199867545 Chr.19: 580335-580335 on Build GRCh38
rs41306874 Chr.19: 580295-580295 on Build GRCh38
rs202113312 Chr.19: 580389-580389 on Build GRCh38
rs369795007 Chr.19: 580350-580350 on Build GRCh38
rs138600800 Chr.19: 580299-580299 on Build GRCh38
rs954383088 Chr.19: 580405-580405 on Build GRCh38
rs28992473 Chr.19: 580349-580349 on Build GRCh38
rs769915952 Chr.19: 580372-580372 on Build GRCh38
rs113194627 Chr.19: 580378-580378 on Build GRCh38
rs969288382 Chr.19: 580412-580412 on Build GRCh38
rs750148301 Chr.19: 580411-580411 on Build GRCh38
rs758959711 Chr.19: 580427-580427 on Build GRCh38
rs201531347 Chr.19: 580351-580351 on Build GRCh38
rs765981796 Chr.19: 580416-580416 on Build GRCh38
rs945915945 Chr.19: 580296-580297 on Build GRCh38
rs145916692 Chr.19: 580437-580437 on Build GRCh38
rs201396156 Chr.19: 580363-580363 on Build GRCh38
rs913029337 Chr.19: 580272-580272 on Build GRCh38
rs972257647 Chr.19: 580312-580312 on Build GRCh38
rs963092708 Chr.19: 580432-580432 on Build GRCh38
rs774217472 Chr.19: 580392-580392 on Build GRCh38
rs376453857 Chr.19: 580391-580391 on Build GRCh38
rs771167010 Chr.19: 580355-580355 on Build GRCh38
rs543090783 Chr.19: 580269-580269 on Build GRCh38
rs968984575 Chr.19: 580287-580287 on Build GRCh38
rs2229664 Chr.19: 580388-580388 on Build GRCh38
rs746151202 Chr.19: 580442-580442 on Build GRCh38
rs760248167 Chr.19: 580415-580415 on Build GRCh38
rs199983087 Chr.19: 580369-580369 on Build GRCh38
rs104894669 Chr.19: 580428-580428 on Build GRCh38

View Full Product Details