Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122847936-122848205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIP1R
Entrez Gene ID
9026
Gene Name
huntingtin interacting protein 1 related
Gene Aliases
HIP12, HIP3, ILWEQ
UniGene
Hs.524815
Gene Chromosome Location
Chr.12: 122834498-122862961 on Build GRCh38
Transcripts
NM_001303097.1 NM_001303099.1 NM_003959.2 XM_011538963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6679976 c.151G>A p.A51T Chr.12: 122848088-122848088 on Build GRCh38
4641638 c.101G>A p.S34N Chr.12: 122848038-122848038 on Build GRCh38
6567870 c.111A>C p.K37N Chr.12: 122848048-122848048 on Build GRCh38
5432056 c.105C>T p.I35I Chr.12: 122848042-122848042 on Build GRCh38
6679989 c.118A>G p.N40D Chr.12: 122848055-122848055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964009217 Chr.12: 122847962-122847962 on Build GRCh38
rs770513477 Chr.12: 122848089-122848089 on Build GRCh38
rs563418222 Chr.12: 122848181-122848181 on Build GRCh38
rs765296536 Chr.12: 122847997-122847997 on Build GRCh38
rs371860720 Chr.12: 122848107-122848107 on Build GRCh38
rs746265607 Chr.12: 122848064-122848064 on Build GRCh38
rs765243628 Chr.12: 122848094-122848094 on Build GRCh38
rs759442153 Chr.12: 122848092-122848092 on Build GRCh38
rs907283688 Chr.12: 122848079-122848079 on Build GRCh38
rs749728728 Chr.12: 122848145-122848145 on Build GRCh38
rs768244770 Chr.12: 122848019-122848020 on Build GRCh38
rs776525480 Chr.12: 122848091-122848091 on Build GRCh38
rs369412426 Chr.12: 122848010-122848010 on Build GRCh38
rs760319031 Chr.12: 122848088-122848088 on Build GRCh38
rs147888758 Chr.12: 122848078-122848078 on Build GRCh38
rs919966750 Chr.12: 122847991-122847991 on Build GRCh38
rs1026886402 Chr.12: 122848150-122848150 on Build GRCh38
rs753589603 Chr.12: 122847982-122847982 on Build GRCh38
rs754913172 Chr.12: 122847983-122847983 on Build GRCh38
rs995364811 Chr.12: 122848147-122848147 on Build GRCh38
rs376982590 Chr.12: 122848134-122848134 on Build GRCh38
rs781312823 Chr.12: 122848122-122848122 on Build GRCh38
rs767469611 Chr.12: 122848004-122848004 on Build GRCh38
rs780538202 Chr.12: 122848144-122848144 on Build GRCh38
rs750766051 Chr.12: 122848128-122848128 on Build GRCh38
rs747346997 Chr.12: 122847975-122847975 on Build GRCh38
rs112655537 Chr.12: 122848072-122848072 on Build GRCh38
rs757448297 Chr.12: 122848118-122848118 on Build GRCh38
rs375011579 Chr.12: 122847981-122847981 on Build GRCh38
rs201026318 Chr.12: 122848099-122848099 on Build GRCh38
rs372704092 Chr.12: 122848015-122848015 on Build GRCh38
rs1055391040 Chr.12: 122848157-122848157 on Build GRCh38
rs564034019 Chr.12: 122848106-122848106 on Build GRCh38
rs892843877 Chr.12: 122848168-122848168 on Build GRCh38
rs752738501 Chr.12: 122848095-122848095 on Build GRCh38
rs934921212 Chr.12: 122848002-122848002 on Build GRCh38
rs781559195 Chr.12: 122848028-122848028 on Build GRCh38
rs572100848 Chr.12: 122848087-122848087 on Build GRCh38
rs747647256 Chr.12: 122848073-122848073 on Build GRCh38
rs975416802 Chr.12: 122847972-122847972 on Build GRCh38

View Full Product Details