Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54391080-54391574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102724050
Entrez Gene ID
102724050
Gene Name
uncharacterized LOC102724050
Gene Aliases
-
UniGene
Hs.670941
Gene Chromosome Location
Chr.12: 54353691-54467030 on Build GRCh38
Transcripts
NR_120486.1 NR_120487.1 NM_001130967.2 NM_001130968.2 XM_011538168.2 XM_011538169.2 XM_011538170.1 XM_011538171.2

Gene Symbol
ZNF385A
Entrez Gene ID
25946
Gene Name
zinc finger protein 385A
Gene Aliases
HZF, RZF, ZFP385, ZNF385
UniGene
Hs.505653
Gene Chromosome Location
Chr.12: 54369129-54391609 on Build GRCh38
Transcripts
NR_120486.1 NR_120487.1 NM_001130967.2 NM_001130968.2 XM_011538168.2 XM_011538169.2 XM_011538170.1 XM_011538171.2

SNP Detail

SNP ID Chromosome Location
rs957659420 Chr.12: 54391254-54391254 on Build GRCh38
rs976937883 Chr.12: 54391397-54391397 on Build GRCh38
rs185149959 Chr.12: 54391206-54391206 on Build GRCh38
rs373926796 Chr.12: 54391117-54391117 on Build GRCh38
rs1018399006 Chr.12: 54391314-54391314 on Build GRCh38
rs894831080 Chr.12: 54391267-54391267 on Build GRCh38
rs902257121 Chr.12: 54391401-54391401 on Build GRCh38
rs909460428 Chr.12: 54391517-54391517 on Build GRCh38
rs181150313 Chr.12: 54391118-54391118 on Build GRCh38
rs552753508 Chr.12: 54391483-54391483 on Build GRCh38
rs965138905 Chr.12: 54391318-54391318 on Build GRCh38
rs1006555306 Chr.12: 54391264-54391264 on Build GRCh38
rs902356517 Chr.12: 54391167-54391167 on Build GRCh38
rs999063723 Chr.12: 54391457-54391457 on Build GRCh38
rs1055425574 Chr.12: 54391228-54391228 on Build GRCh38
rs548138220 Chr.12: 54391195-54391195 on Build GRCh38
rs537316629 Chr.12: 54391396-54391396 on Build GRCh38
rs906485961 Chr.12: 54391136-54391136 on Build GRCh38
rs923435093 Chr.12: 54391457-54391459 on Build GRCh38
rs983633801 Chr.12: 54391485-54391485 on Build GRCh38
rs1010950984 Chr.12: 54391271-54391271 on Build GRCh38
rs7301131 Chr.12: 54391478-54391478 on Build GRCh38
rs1013175578 Chr.12: 54391315-54391315 on Build GRCh38

View Full Product Details