Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57823820-57824316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSP2
Entrez Gene ID
10106
Gene Name
CTD small phosphatase 2
Gene Aliases
OS4, PSR2, SCP2
UniGene
Hs.524530
Gene Chromosome Location
Chr.12: 57819927-57846964 on Build GRCh38
Transcripts
NM_005730.3 XM_005268556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4147352 c.672A>C p.I224I Chr.12: 57823922-57823922 on Build GRCh38
5765519 c.639C>T p.T213T Chr.12: 57823955-57823955 on Build GRCh38
5765533 c.603G>T p.K201N Chr.12: 57823991-57823991 on Build GRCh38
1238616 c.577G>A p.V193M Chr.12: 57824017-57824017 on Build GRCh38
4575634 c.596A>G p.Y199C Chr.12: 57823998-57823998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774272222 Chr.12: 57824287-57824287 on Build GRCh38
rs78073833 Chr.12: 57824144-57824144 on Build GRCh38
rs528788948 Chr.12: 57824222-57824222 on Build GRCh38
rs761403823 Chr.12: 57824083-57824083 on Build GRCh38
rs74568739 Chr.12: 57823958-57823958 on Build GRCh38
rs1049782028 Chr.12: 57824011-57824011 on Build GRCh38
rs774936151 Chr.12: 57823921-57823921 on Build GRCh38
rs758269345 Chr.12: 57823981-57823981 on Build GRCh38
rs746850735 Chr.12: 57823887-57823887 on Build GRCh38
rs1043612511 Chr.12: 57824012-57824012 on Build GRCh38
rs766226008 Chr.12: 57824122-57824122 on Build GRCh38
rs951822634 Chr.12: 57824167-57824167 on Build GRCh38
rs769661563 Chr.12: 57823919-57823919 on Build GRCh38
rs763692586 Chr.12: 57823933-57823933 on Build GRCh38
rs932834632 Chr.12: 57823912-57823912 on Build GRCh38
rs372871136 Chr.12: 57824099-57824099 on Build GRCh38
rs367578724 Chr.12: 57823892-57823892 on Build GRCh38
rs769447012 Chr.12: 57824259-57824259 on Build GRCh38
rs371776245 Chr.12: 57824193-57824193 on Build GRCh38
rs536556645 Chr.12: 57824176-57824176 on Build GRCh38
rs781385318 Chr.12: 57824008-57824008 on Build GRCh38
rs371747999 Chr.12: 57823878-57823878 on Build GRCh38
rs796175724 Chr.12: 57823915-57823915 on Build GRCh38
rs779382330 Chr.12: 57824192-57824192 on Build GRCh38
rs111421573 Chr.12: 57823997-57823997 on Build GRCh38
rs777781101 Chr.12: 57824184-57824184 on Build GRCh38
rs77410204 Chr.12: 57823924-57823924 on Build GRCh38
rs879069221 Chr.12: 57824175-57824175 on Build GRCh38
rs762714721 Chr.12: 57824038-57824038 on Build GRCh38
rs757392254 Chr.12: 57824190-57824190 on Build GRCh38
rs139687109 Chr.12: 57824290-57824290 on Build GRCh38
rs78662830 Chr.12: 57823922-57823922 on Build GRCh38
rs868454867 Chr.12: 57824142-57824142 on Build GRCh38
rs375983786 Chr.12: 57823879-57823879 on Build GRCh38
rs375138358 Chr.12: 57823946-57823946 on Build GRCh38
rs970187589 Chr.12: 57824135-57824135 on Build GRCh38
rs756200799 Chr.12: 57824181-57824181 on Build GRCh38
rs759720451 Chr.12: 57824103-57824104 on Build GRCh38
rs781080931 Chr.12: 57823889-57823889 on Build GRCh38
rs904567774 Chr.12: 57824017-57824017 on Build GRCh38
rs766026719 Chr.12: 57823982-57823982 on Build GRCh38
rs370609789 Chr.12: 57824233-57824233 on Build GRCh38
rs563637364 Chr.12: 57823907-57823907 on Build GRCh38
rs200561450 Chr.12: 57824179-57824179 on Build GRCh38
rs995231664 Chr.12: 57824138-57824138 on Build GRCh38
rs764969768 Chr.12: 57823949-57823949 on Build GRCh38
rs762703799 Chr.12: 57824268-57824268 on Build GRCh38
rs369014414 Chr.12: 57824078-57824078 on Build GRCh38
rs772806247 Chr.12: 57824266-57824266 on Build GRCh38
rs749454719 Chr.12: 57824036-57824036 on Build GRCh38
rs754828681 Chr.12: 57824006-57824006 on Build GRCh38
rs751292723 Chr.12: 57824129-57824129 on Build GRCh38
rs775971558 Chr.12: 57823943-57823943 on Build GRCh38
rs373350059 Chr.12: 57824043-57824043 on Build GRCh38
rs1029081352 Chr.12: 57824020-57824020 on Build GRCh38
rs12822897 Chr.12: 57823925-57823925 on Build GRCh38
rs748413501 Chr.12: 57823891-57823891 on Build GRCh38
rs778548979 Chr.12: 57823852-57823852 on Build GRCh38
rs767907387 Chr.12: 57824190-57824191 on Build GRCh38
rs202073979 Chr.12: 57823876-57823876 on Build GRCh38
rs184941879 Chr.12: 57823913-57823913 on Build GRCh38
rs774273456 Chr.12: 57823853-57823854 on Build GRCh38
rs772702512 Chr.12: 57824205-57824205 on Build GRCh38
rs780710349 Chr.12: 57824216-57824216 on Build GRCh38
rs369938855 Chr.12: 57823996-57823996 on Build GRCh38
rs761335371 Chr.12: 57824084-57824084 on Build GRCh38
rs566402043 Chr.12: 57824151-57824151 on Build GRCh38
rs200920287 Chr.12: 57824095-57824095 on Build GRCh38
rs547470011 Chr.12: 57824147-57824147 on Build GRCh38
rs375402815 Chr.12: 57824255-57824255 on Build GRCh38
rs1040559286 Chr.12: 57823881-57823881 on Build GRCh38
rs765161871 Chr.12: 57823875-57823875 on Build GRCh38
rs990644760 Chr.12: 57823845-57823845 on Build GRCh38
rs79510860 Chr.12: 57823961-57823961 on Build GRCh38
rs1012306141 Chr.12: 57824159-57824159 on Build GRCh38
rs754092066 Chr.12: 57824186-57824186 on Build GRCh38
rs779108955 Chr.12: 57824044-57824044 on Build GRCh38
rs773628562 Chr.12: 57823894-57823894 on Build GRCh38
rs1036858847 Chr.12: 57824118-57824118 on Build GRCh38
rs796106620 Chr.12: 57823987-57823987 on Build GRCh38
rs563483951 Chr.12: 57824018-57824018 on Build GRCh38
rs868709875 Chr.12: 57823938-57823938 on Build GRCh38
rs113792624 Chr.12: 57823991-57823991 on Build GRCh38
rs376232288 Chr.12: 57824058-57824058 on Build GRCh38
rs796729342 Chr.12: 57823979-57823979 on Build GRCh38
rs770480549 Chr.12: 57824281-57824281 on Build GRCh38
rs796989809 Chr.12: 57823980-57823980 on Build GRCh38
rs752568029 Chr.12: 57824180-57824180 on Build GRCh38
rs756931156 Chr.12: 57823853-57823853 on Build GRCh38
rs879171354 Chr.12: 57824175-57824175 on Build GRCh38
rs369224307 Chr.12: 57823886-57823886 on Build GRCh38
rs944354427 Chr.12: 57823877-57823877 on Build GRCh38
rs772434435 Chr.12: 57824059-57824059 on Build GRCh38
rs192597051 Chr.12: 57823955-57823955 on Build GRCh38
rs77145290 Chr.12: 57823955-57823956 on Build GRCh38

View Full Product Details