Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110452902-110453404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPN3
Entrez Gene ID
51184
Gene Name
GPN-loop GTPase 3
Gene Aliases
ATPBD1C
UniGene
Hs.634680
Gene Chromosome Location
Chr.12: 110452486-110468835 on Build GRCh38
Transcripts
NM_001164372.1 NM_001164373.1 NM_016301.3 XM_005253896.4 XM_017019394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4574997 c.934A>G p.M312V Chr.12: 110453072-110453072 on Build GRCh38
4574998 c.817A>G p.M273V Chr.12: 110453072-110453072 on Build GRCh38
1208565 c.952C>T p.Q318* Chr.12: 110453054-110453054 on Build GRCh38
5739797 c.922G>T p.E308* Chr.12: 110453084-110453084 on Build GRCh38
5739798 c.805G>T p.E269* Chr.12: 110453084-110453084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149713209 Chr.12: 110452959-110452959 on Build GRCh38
rs779030739 Chr.12: 110453109-110453109 on Build GRCh38
rs376361210 Chr.12: 110453042-110453042 on Build GRCh38
rs1032696320 Chr.12: 110453277-110453277 on Build GRCh38
rs895827354 Chr.12: 110453304-110453304 on Build GRCh38
rs541477114 Chr.12: 110452933-110452933 on Build GRCh38
rs974812685 Chr.12: 110452972-110452972 on Build GRCh38
rs978404871 Chr.12: 110453268-110453268 on Build GRCh38
rs921600530 Chr.12: 110453010-110453010 on Build GRCh38
rs529270245 Chr.12: 110453243-110453243 on Build GRCh38
rs373419093 Chr.12: 110453041-110453041 on Build GRCh38
rs1028067846 Chr.12: 110453203-110453203 on Build GRCh38
rs776451770 Chr.12: 110453002-110453002 on Build GRCh38
rs200226170 Chr.12: 110453063-110453063 on Build GRCh38
rs568239573 Chr.12: 110453237-110453237 on Build GRCh38
rs181742521 Chr.12: 110453165-110453165 on Build GRCh38
rs751404912 Chr.12: 110453064-110453064 on Build GRCh38
rs764900257 Chr.12: 110453033-110453033 on Build GRCh38
rs937311579 Chr.12: 110453283-110453283 on Build GRCh38
rs569927764 Chr.12: 110453346-110453346 on Build GRCh38
rs190429303 Chr.12: 110452958-110452958 on Build GRCh38
rs1014264289 Chr.12: 110453325-110453325 on Build GRCh38
rs138166782 Chr.12: 110453093-110453093 on Build GRCh38
rs1041802656 Chr.12: 110453343-110453343 on Build GRCh38
rs372672794 Chr.12: 110452969-110452969 on Build GRCh38
rs186824410 Chr.12: 110453245-110453245 on Build GRCh38
rs144309333 Chr.12: 110453188-110453188 on Build GRCh38
rs370414908 Chr.12: 110453144-110453144 on Build GRCh38
rs951869909 Chr.12: 110453160-110453176 on Build GRCh38
rs998066615 Chr.12: 110453195-110453195 on Build GRCh38
rs35343339 Chr.12: 110453201-110453202 on Build GRCh38
rs761487347 Chr.12: 110453019-110453019 on Build GRCh38
rs757456653 Chr.12: 110453084-110453084 on Build GRCh38
rs1033659805 Chr.12: 110452944-110452944 on Build GRCh38
rs749948144 Chr.12: 110453035-110453035 on Build GRCh38
rs754833965 Chr.12: 110453066-110453066 on Build GRCh38
rs956248918 Chr.12: 110453363-110453363 on Build GRCh38
rs754564161 Chr.12: 110452978-110452978 on Build GRCh38
rs367859550 Chr.12: 110453072-110453072 on Build GRCh38
rs769089314 Chr.12: 110452990-110452990 on Build GRCh38
rs777439084 Chr.12: 110453066-110453069 on Build GRCh38
rs1044878304 Chr.12: 110452992-110452992 on Build GRCh38
rs954411615 Chr.12: 110453031-110453031 on Build GRCh38
rs539841313 Chr.12: 110453021-110453021 on Build GRCh38
rs1055680330 Chr.12: 110453302-110453302 on Build GRCh38
rs373767119 Chr.12: 110453137-110453137 on Build GRCh38
rs767234920 Chr.12: 110453071-110453071 on Build GRCh38
rs747863460 Chr.12: 110453380-110453380 on Build GRCh38

View Full Product Details