Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 108232481-108232988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD13
Entrez Gene ID
84945
Gene Name
abhydrolase domain containing 13
Gene Aliases
BEM46L1, C13orf6, bA153I24.2
UniGene
Hs.183528
Gene Chromosome Location
Chr.13: 108218415-108235440 on Build GRCh38
Transcripts
NM_032859.2 XM_011521128.2

SNP Detail

SNP ID Chromosome Location
rs892056884 Chr.13: 108232871-108232871 on Build GRCh38
rs55665720 Chr.13: 108232808-108232808 on Build GRCh38
rs976603469 Chr.13: 108232589-108232589 on Build GRCh38
rs745786535 Chr.13: 108232949-108232949 on Build GRCh38
rs986655934 Chr.13: 108232725-108232725 on Build GRCh38
rs998096571 Chr.13: 108232867-108232867 on Build GRCh38
rs1055383603 Chr.13: 108232742-108232742 on Build GRCh38
rs771745624 Chr.13: 108232613-108232613 on Build GRCh38
rs939619378 Chr.13: 108232715-108232715 on Build GRCh38
rs1022371210 Chr.13: 108232537-108232537 on Build GRCh38
rs1018953336 Chr.13: 108232794-108232794 on Build GRCh38
rs550680250 Chr.13: 108232704-108232704 on Build GRCh38
rs575424079 Chr.13: 108232955-108232955 on Build GRCh38
rs763503027 Chr.13: 108232951-108232955 on Build GRCh38
rs909618002 Chr.13: 108232659-108232659 on Build GRCh38
rs186149567 Chr.13: 108232611-108232611 on Build GRCh38
rs1010780886 Chr.13: 108232890-108232890 on Build GRCh38
rs983930168 Chr.13: 108232905-108232909 on Build GRCh38
rs566433238 Chr.13: 108232855-108232855 on Build GRCh38
rs1001560857 Chr.13: 108232948-108232948 on Build GRCh38
rs983858040 Chr.13: 108232640-108232640 on Build GRCh38
rs79447654 Chr.13: 108232630-108232630 on Build GRCh38
rs967143079 Chr.13: 108232552-108232552 on Build GRCh38
rs182838039 Chr.13: 108232747-108232747 on Build GRCh38
rs554051601 Chr.13: 108232787-108232787 on Build GRCh38
rs999978923 Chr.13: 108232788-108232788 on Build GRCh38
rs776149998 Chr.13: 108232949-108232952 on Build GRCh38
rs918548674 Chr.13: 108232675-108232675 on Build GRCh38
rs902615902 Chr.13: 108232787-108232787 on Build GRCh38
rs561791648 Chr.13: 108232612-108232612 on Build GRCh38
rs1034326012 Chr.13: 108232586-108232586 on Build GRCh38
rs1024445984 Chr.13: 108232896-108232896 on Build GRCh38
rs772805493 Chr.13: 108232713-108232713 on Build GRCh38
rs555294343 Chr.13: 108232938-108232938 on Build GRCh38
rs962769153 Chr.13: 108232624-108232624 on Build GRCh38
rs980048934 Chr.13: 108232560-108232560 on Build GRCh38
rs942441977 Chr.13: 108232789-108232789 on Build GRCh38
rs953908116 Chr.13: 108232897-108232897 on Build GRCh38
rs190625520 Chr.13: 108232739-108232739 on Build GRCh38
rs1039505249 Chr.13: 108232795-108232795 on Build GRCh38
rs1011066562 Chr.13: 108232783-108232783 on Build GRCh38
rs895341761 Chr.13: 108232753-108232753 on Build GRCh38
rs912292281 Chr.13: 108232730-108232730 on Build GRCh38
rs907268241 Chr.13: 108232939-108232939 on Build GRCh38
rs1055109627 Chr.13: 108232827-108232827 on Build GRCh38
rs1019339472 Chr.13: 108232578-108232578 on Build GRCh38
rs546488209 Chr.13: 108232828-108232828 on Build GRCh38
rs774203919 Chr.13: 108232856-108232856 on Build GRCh38
rs761250667 Chr.13: 108232573-108232573 on Build GRCh38
rs961164574 Chr.13: 108232933-108232933 on Build GRCh38
rs903502065 Chr.13: 108232800-108232800 on Build GRCh38
rs773867823 Chr.13: 108232547-108232547 on Build GRCh38
rs187384181 Chr.13: 108232911-108232911 on Build GRCh38
rs528637379 Chr.13: 108232579-108232579 on Build GRCh38
rs1011035521 Chr.13: 108232512-108232512 on Build GRCh38
rs971455818 Chr.13: 108232557-108232557 on Build GRCh38
rs1036706064 Chr.13: 108232690-108232690 on Build GRCh38
rs927692841 Chr.13: 108232941-108232941 on Build GRCh38
rs909681101 Chr.13: 108232914-108232915 on Build GRCh38

View Full Product Details