Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 44927549-44928059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL28
Entrez Gene ID
54813
Gene Name
kelch like family member 28
Gene Aliases
BTBD5
UniGene
Hs.550906
Gene Chromosome Location
Chr.14: 44924310-44961978 on Build GRCh38
Transcripts
NM_001308112.1 NM_017658.4 XM_005267770.3 XM_011536847.2 XM_011536849.2

SNP Detail

SNP ID Chromosome Location
rs1023368783 Chr.14: 44927586-44927586 on Build GRCh38
rs6572290 Chr.14: 44927962-44927962 on Build GRCh38
rs191707081 Chr.14: 44927722-44927722 on Build GRCh38
rs906278750 Chr.14: 44927860-44927860 on Build GRCh38
rs188992033 Chr.14: 44927620-44927620 on Build GRCh38
rs933448590 Chr.14: 44927729-44927729 on Build GRCh38
rs888380838 Chr.14: 44927644-44927644 on Build GRCh38
rs79628027 Chr.14: 44927706-44927706 on Build GRCh38
rs183032983 Chr.14: 44927776-44927776 on Build GRCh38
rs1003253714 Chr.14: 44927869-44927869 on Build GRCh38
rs1035902014 Chr.14: 44927619-44927619 on Build GRCh38
rs999672181 Chr.14: 44927745-44927745 on Build GRCh38
rs1016503024 Chr.14: 44927956-44927956 on Build GRCh38
rs958296340 Chr.14: 44927755-44927755 on Build GRCh38
rs763456062 Chr.14: 44927862-44927862 on Build GRCh38
rs760437512 Chr.14: 44927957-44927957 on Build GRCh38
rs199601468 Chr.14: 44927997-44927997 on Build GRCh38
rs750944430 Chr.14: 44927895-44927895 on Build GRCh38
rs576627822 Chr.14: 44927677-44927677 on Build GRCh38
rs559518939 Chr.14: 44927808-44927808 on Build GRCh38
rs58836600 Chr.14: 44927768-44927768 on Build GRCh38
rs918698369 Chr.14: 44927832-44927832 on Build GRCh38
rs369559749 Chr.14: 44928010-44928010 on Build GRCh38
rs970492303 Chr.14: 44927591-44927591 on Build GRCh38
rs937256404 Chr.14: 44927911-44927911 on Build GRCh38
rs1023339451 Chr.14: 44927831-44927831 on Build GRCh38
rs59360063 Chr.14: 44927746-44927746 on Build GRCh38
rs942019378 Chr.14: 44927712-44927712 on Build GRCh38
rs370424922 Chr.14: 44927993-44927993 on Build GRCh38
rs935171794 Chr.14: 44927777-44927777 on Build GRCh38
rs977143735 Chr.14: 44927824-44927824 on Build GRCh38
rs1005485835 Chr.14: 44927935-44927935 on Build GRCh38
rs1011905113 Chr.14: 44927797-44927797 on Build GRCh38
rs1055755876 Chr.14: 44927927-44927927 on Build GRCh38
rs950889968 Chr.14: 44927905-44927905 on Build GRCh38
rs987546157 Chr.14: 44928030-44928030 on Build GRCh38
rs893735834 Chr.14: 44927793-44927793 on Build GRCh38
rs950950440 Chr.14: 44927666-44927666 on Build GRCh38
rs983687084 Chr.14: 44927675-44927675 on Build GRCh38
rs954719953 Chr.14: 44928002-44928002 on Build GRCh38
rs772740211 Chr.14: 44927783-44927783 on Build GRCh38
rs921900081 Chr.14: 44927985-44927985 on Build GRCh38
rs902930806 Chr.14: 44927728-44927728 on Build GRCh38
rs930019268 Chr.14: 44927885-44927885 on Build GRCh38
rs538841685 Chr.14: 44927597-44927597 on Build GRCh38
rs974744405 Chr.14: 44927719-44927719 on Build GRCh38

View Full Product Details