Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49622909-49623411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGAT2
Entrez Gene ID
4247
Gene Name
mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase
Gene Aliases
CDG2A, CDGS2, GLCNACTII, GNT-II, GNT2
UniGene
Hs.93338
Gene Chromosome Location
Chr.14: 49620771-49623481 on Build GRCh38
Transcripts
NM_002408.3

SNP Detail

SNP ID Chromosome Location
rs919869818 Chr.14: 49623270-49623271 on Build GRCh38
rs189705265 Chr.14: 49623233-49623233 on Build GRCh38
rs1039899808 Chr.14: 49623070-49623070 on Build GRCh38
rs906999716 Chr.14: 49623213-49623213 on Build GRCh38
rs1046955157 Chr.14: 49623271-49623273 on Build GRCh38
rs992884093 Chr.14: 49623044-49623044 on Build GRCh38
rs779203353 Chr.14: 49623144-49623144 on Build GRCh38
rs1040835491 Chr.14: 49623320-49623320 on Build GRCh38
rs960214333 Chr.14: 49623365-49623365 on Build GRCh38
rs1011373 Chr.14: 49622978-49622978 on Build GRCh38
rs149722843 Chr.14: 49623112-49623112 on Build GRCh38
rs943741793 Chr.14: 49623272-49623272 on Build GRCh38
rs989953454 Chr.14: 49622966-49622966 on Build GRCh38
rs757674496 Chr.14: 49622984-49622984 on Build GRCh38
rs1034676616 Chr.14: 49623286-49623286 on Build GRCh38
rs1013652206 Chr.14: 49623178-49623178 on Build GRCh38
rs912952904 Chr.14: 49622975-49622975 on Build GRCh38
rs957252920 Chr.14: 49622938-49622938 on Build GRCh38
rs559336604 Chr.14: 49622967-49622967 on Build GRCh38
rs569688212 Chr.14: 49623179-49623179 on Build GRCh38
rs980678940 Chr.14: 49623216-49623216 on Build GRCh38
rs886050523 Chr.14: 49623278-49623281 on Build GRCh38
rs573436485 Chr.14: 49623271-49623274 on Build GRCh38
rs960141094 Chr.14: 49623039-49623039 on Build GRCh38
rs548966102 Chr.14: 49623191-49623191 on Build GRCh38
rs992831653 Chr.14: 49623372-49623372 on Build GRCh38
rs879797900 Chr.14: 49623141-49623141 on Build GRCh38
rs536689512 Chr.14: 49623238-49623238 on Build GRCh38
rs537612516 Chr.14: 49623381-49623381 on Build GRCh38
rs886050522 Chr.14: 49623281-49623281 on Build GRCh38
rs549718708 Chr.14: 49623133-49623133 on Build GRCh38
rs750394836 Chr.14: 49623211-49623214 on Build GRCh38
rs879604712 Chr.14: 49623271-49623271 on Build GRCh38
rs535317346 Chr.14: 49623326-49623326 on Build GRCh38
rs1031769834 Chr.14: 49623350-49623350 on Build GRCh38
rs529002828 Chr.14: 49623185-49623185 on Build GRCh38
rs756139772 Chr.14: 49623268-49623270 on Build GRCh38
rs558537786 Chr.14: 49623331-49623331 on Build GRCh38
rs571740661 Chr.14: 49623334-49623334 on Build GRCh38
rs946263071 Chr.14: 49622976-49622976 on Build GRCh38
rs185441515 Chr.14: 49623128-49623128 on Build GRCh38
rs200202235 Chr.14: 49623282-49623282 on Build GRCh38
rs139830740 Chr.14: 49622982-49622982 on Build GRCh38
rs535682855 Chr.14: 49623088-49623088 on Build GRCh38
rs887204343 Chr.14: 49623160-49623160 on Build GRCh38
rs886050521 Chr.14: 49623216-49623219 on Build GRCh38
rs139553752 Chr.14: 49623031-49623031 on Build GRCh38
rs978028360 Chr.14: 49622987-49622987 on Build GRCh38
rs986475641 Chr.14: 49623370-49623370 on Build GRCh38
rs957081723 Chr.14: 49623305-49623305 on Build GRCh38

View Full Product Details