Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 67646840-67647340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARG2
Entrez Gene ID
384
Gene Name
arginase 2
Gene Aliases
-
UniGene
Hs.226007
Gene Chromosome Location
Chr.14: 67619862-67651720 on Build GRCh38
Transcripts
NM_001172.3 XM_017020925.1

Gene Symbol
GPHN
Entrez Gene ID
10243
Gene Name
gephyrin
Gene Aliases
GEPH, GPH, GPHRYN, HKPX1, MOCODC
UniGene
Hs.208765
Gene Chromosome Location
Chr.14: 66507407-67735831 on Build GRCh38
Transcripts
NM_001172.3 XM_017020925.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5670322 c.663T>C p.D221D Chr.14: 67646966-67646966 on Build GRCh38
6458239 c.664delA p.I222fs*17 Chr.14: 67646967-67646967 on Build GRCh38
3983655 c.618-10C>T p.? Chr.14: 67646911-67646911 on Build GRCh38
5478735 c.630G>T p.K210N Chr.14: 67646933-67646933 on Build GRCh38
7203499 c.716T>C p.I239T Chr.14: 67647019-67647019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535965075 Chr.14: 67646987-67646987 on Build GRCh38
rs747904249 Chr.14: 67646974-67646974 on Build GRCh38
rs199907124 Chr.14: 67647063-67647063 on Build GRCh38
rs980474730 Chr.14: 67647248-67647248 on Build GRCh38
rs573030314 Chr.14: 67647035-67647035 on Build GRCh38
rs774197237 Chr.14: 67646979-67646979 on Build GRCh38
rs753174084 Chr.14: 67646891-67646891 on Build GRCh38
rs61990126 Chr.14: 67647204-67647204 on Build GRCh38
rs749268136 Chr.14: 67646885-67646889 on Build GRCh38
rs145867978 Chr.14: 67647000-67647000 on Build GRCh38
rs533251976 Chr.14: 67647111-67647111 on Build GRCh38
rs527930454 Chr.14: 67647296-67647296 on Build GRCh38
rs750708129 Chr.14: 67647012-67647012 on Build GRCh38
rs779248770 Chr.14: 67646937-67646937 on Build GRCh38
rs190496290 Chr.14: 67647162-67647162 on Build GRCh38
rs781163951 Chr.14: 67647041-67647041 on Build GRCh38
rs902058986 Chr.14: 67647091-67647091 on Build GRCh38
rs770589105 Chr.14: 67646916-67646917 on Build GRCh38
rs141792961 Chr.14: 67646986-67646986 on Build GRCh38
rs568480532 Chr.14: 67646947-67646947 on Build GRCh38
rs889476314 Chr.14: 67647067-67647067 on Build GRCh38
rs746928269 Chr.14: 67646964-67646964 on Build GRCh38
rs768622010 Chr.14: 67646968-67646968 on Build GRCh38
rs918359026 Chr.14: 67647019-67647019 on Build GRCh38
rs769094848 Chr.14: 67647076-67647076 on Build GRCh38
rs182319313 Chr.14: 67647259-67647259 on Build GRCh38
rs761017358 Chr.14: 67646916-67646916 on Build GRCh38
rs754272721 Chr.14: 67647044-67647044 on Build GRCh38
rs780015756 Chr.14: 67647020-67647020 on Build GRCh38
rs17104534 Chr.14: 67647021-67647021 on Build GRCh38
rs574863109 Chr.14: 67647181-67647181 on Build GRCh38
rs765825080 Chr.14: 67646875-67646875 on Build GRCh38
rs560257193 Chr.14: 67647093-67647093 on Build GRCh38
rs775112979 Chr.14: 67647024-67647027 on Build GRCh38
rs761177455 Chr.14: 67646898-67646898 on Build GRCh38
rs767333263 Chr.14: 67646997-67647000 on Build GRCh38
rs3208526 Chr.14: 67646921-67646921 on Build GRCh38
rs914604842 Chr.14: 67646863-67646863 on Build GRCh38
rs867654375 Chr.14: 67647140-67647140 on Build GRCh38
rs759202982 Chr.14: 67646986-67646988 on Build GRCh38
rs893018647 Chr.14: 67647135-67647135 on Build GRCh38
rs780145505 Chr.14: 67646950-67646950 on Build GRCh38
rs1039321871 Chr.14: 67647079-67647079 on Build GRCh38
rs368547332 Chr.14: 67647051-67647051 on Build GRCh38
rs970080122 Chr.14: 67647185-67647185 on Build GRCh38
rs758630775 Chr.14: 67647014-67647014 on Build GRCh38
rs1058584 Chr.14: 67647150-67647150 on Build GRCh38
rs764379736 Chr.14: 67646912-67646912 on Build GRCh38
rs554279681 Chr.14: 67647082-67647083 on Build GRCh38
rs773801177 Chr.14: 67647065-67647065 on Build GRCh38
rs754217370 Chr.14: 67646915-67646915 on Build GRCh38
rs568264895 Chr.14: 67647001-67647001 on Build GRCh38
rs766799685 Chr.14: 67646957-67646957 on Build GRCh38
rs372873309 Chr.14: 67647071-67647071 on Build GRCh38
rs373436709 Chr.14: 67647103-67647103 on Build GRCh38
rs748027189 Chr.14: 67647048-67647048 on Build GRCh38
rs756715543 Chr.14: 67646995-67646995 on Build GRCh38
rs780911383 Chr.14: 67646969-67646969 on Build GRCh38
rs769482292 Chr.14: 67646979-67646979 on Build GRCh38
rs1049524273 Chr.14: 67647149-67647149 on Build GRCh38
rs372974736 Chr.14: 67646938-67646938 on Build GRCh38
rs542343361 Chr.14: 67647238-67647238 on Build GRCh38
rs773070918 Chr.14: 67646872-67646873 on Build GRCh38
rs777476327 Chr.14: 67647059-67647059 on Build GRCh38
rs577213999 Chr.14: 67647105-67647105 on Build GRCh38
rs773510157 Chr.14: 67646892-67646892 on Build GRCh38
rs764657573 Chr.14: 67646990-67646990 on Build GRCh38
rs928268505 Chr.14: 67647089-67647089 on Build GRCh38
rs760139055 Chr.14: 67647145-67647145 on Build GRCh38
rs762550966 Chr.14: 67646980-67646980 on Build GRCh38
rs748953219 Chr.14: 67647062-67647062 on Build GRCh38

View Full Product Details