Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57883714-57884215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGB1
Entrez Gene ID
1258
Gene Name
cyclic nucleotide gated channel beta 1
Gene Aliases
CNCG2, CNCG3L, CNCG4, CNG4, CNGB1B, GAR1, GARP, GARP2, RCNC2, RCNCb, RCNCbeta, RP45
UniGene
Hs.147062
Gene Chromosome Location
Chr.16: 57882340-57971116 on Build GRCh38
Transcripts
NM_001286130.1 NM_001297.4 XM_011522870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4061474 c.3747G>T p.K1249N Chr.16: 57884173-57884173 on Build GRCh38
3510562 c.3738G>A p.R1246R Chr.16: 57884182-57884182 on Build GRCh38
4061475 c.3737G>T p.R1246M Chr.16: 57884183-57884183 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1024097250 Chr.16: 57883906-57883906 on Build GRCh38
rs376930411 Chr.16: 57883968-57883968 on Build GRCh38
rs750832506 Chr.16: 57884133-57884133 on Build GRCh38
rs1004688962 Chr.16: 57883788-57883788 on Build GRCh38
rs750725220 Chr.16: 57884183-57884183 on Build GRCh38
rs182224027 Chr.16: 57884048-57884048 on Build GRCh38
rs553830954 Chr.16: 57884170-57884170 on Build GRCh38
rs756653795 Chr.16: 57884146-57884146 on Build GRCh38
rs753833037 Chr.16: 57884115-57884126 on Build GRCh38
rs748268051 Chr.16: 57884179-57884179 on Build GRCh38
rs769708952 Chr.16: 57884122-57884122 on Build GRCh38
rs915400837 Chr.16: 57883958-57883958 on Build GRCh38
rs547487989 Chr.16: 57883973-57883973 on Build GRCh38
rs766703655 Chr.16: 57884137-57884137 on Build GRCh38
rs890172879 Chr.16: 57883761-57883761 on Build GRCh38
rs995041853 Chr.16: 57884049-57884049 on Build GRCh38
rs751720550 Chr.16: 57884030-57884030 on Build GRCh38
rs527847594 Chr.16: 57883826-57883826 on Build GRCh38
rs763247427 Chr.16: 57884125-57884125 on Build GRCh38
rs761081124 Chr.16: 57884134-57884134 on Build GRCh38
rs13338071 Chr.16: 57884131-57884131 on Build GRCh38
rs778876989 Chr.16: 57884138-57884139 on Build GRCh38
rs77128930 Chr.16: 57883843-57883843 on Build GRCh38
rs886052190 Chr.16: 57883929-57883929 on Build GRCh38
rs956752961 Chr.16: 57883939-57883939 on Build GRCh38
rs745680361 Chr.16: 57884113-57884113 on Build GRCh38
rs988331943 Chr.16: 57883940-57883940 on Build GRCh38
rs116564376 Chr.16: 57883883-57883883 on Build GRCh38
rs982076657 Chr.16: 57883929-57883929 on Build GRCh38
rs779965828 Chr.16: 57884165-57884165 on Build GRCh38
rs921480478 Chr.16: 57883978-57883978 on Build GRCh38
rs1003564801 Chr.16: 57884097-57884097 on Build GRCh38
rs57309302 Chr.16: 57883854-57883854 on Build GRCh38
rs565898281 Chr.16: 57883977-57883977 on Build GRCh38
rs199896896 Chr.16: 57884152-57884152 on Build GRCh38
rs1051347174 Chr.16: 57884014-57884014 on Build GRCh38
rs572543809 Chr.16: 57884110-57884110 on Build GRCh38
rs534779665 Chr.16: 57884191-57884191 on Build GRCh38
rs538760668 Chr.16: 57884163-57884163 on Build GRCh38
rs774523347 Chr.16: 57884130-57884130 on Build GRCh38
rs372109962 Chr.16: 57884192-57884192 on Build GRCh38
rs536435864 Chr.16: 57883982-57883982 on Build GRCh38
rs745787278 Chr.16: 57884154-57884154 on Build GRCh38
rs890225271 Chr.16: 57884015-57884015 on Build GRCh38
rs775914460 Chr.16: 57884123-57884123 on Build GRCh38
rs931662996 Chr.16: 57883994-57883994 on Build GRCh38
rs371395789 Chr.16: 57884140-57884140 on Build GRCh38
rs905079597 Chr.16: 57884066-57884066 on Build GRCh38
rs757224951 Chr.16: 57884128-57884129 on Build GRCh38
rs760024413 Chr.16: 57884043-57884043 on Build GRCh38
rs112033454 Chr.16: 57883920-57883920 on Build GRCh38
rs751092081 Chr.16: 57884141-57884141 on Build GRCh38
rs772044451 Chr.16: 57884190-57884190 on Build GRCh38
rs768899200 Chr.16: 57884126-57884126 on Build GRCh38
rs375526865 Chr.16: 57884139-57884139 on Build GRCh38
rs879485949 Chr.16: 57884116-57884116 on Build GRCh38
rs548548999 Chr.16: 57883993-57883993 on Build GRCh38
rs189293298 Chr.16: 57883942-57883942 on Build GRCh38
rs34407910 Chr.16: 57884108-57884108 on Build GRCh38
rs1036637298 Chr.16: 57884037-57884037 on Build GRCh38
rs561430118 Chr.16: 57884181-57884181 on Build GRCh38
rs556523294 Chr.16: 57884053-57884053 on Build GRCh38
rs183796279 Chr.16: 57883936-57883936 on Build GRCh38
rs768947322 Chr.16: 57884169-57884169 on Build GRCh38
rs940429648 Chr.16: 57884035-57884035 on Build GRCh38
rs569923676 Chr.16: 57883995-57883995 on Build GRCh38
rs749054291 Chr.16: 57884166-57884166 on Build GRCh38

View Full Product Details