Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49994982-49995485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLX3
Entrez Gene ID
1747
Gene Name
distal-less homeobox 3
Gene Aliases
AI4, TDO
UniGene
Hs.134194
Gene Chromosome Location
Chr.17: 49990005-49995224 on Build GRCh38
Transcripts
NM_005220.2

SNP Detail

SNP ID Chromosome Location
rs369031536 Chr.17: 49995049-49995049 on Build GRCh38
rs1043956513 Chr.17: 49995346-49995346 on Build GRCh38
rs1008995558 Chr.17: 49995403-49995404 on Build GRCh38
rs375929276 Chr.17: 49995035-49995035 on Build GRCh38
rs377216020 Chr.17: 49995342-49995342 on Build GRCh38
rs554800240 Chr.17: 49995068-49995068 on Build GRCh38
rs776884515 Chr.17: 49995462-49995463 on Build GRCh38
rs886053136 Chr.17: 49995188-49995188 on Build GRCh38
rs965028666 Chr.17: 49995143-49995143 on Build GRCh38
rs1046809562 Chr.17: 49995373-49995373 on Build GRCh38
rs543871008 Chr.17: 49995129-49995129 on Build GRCh38
rs1004228387 Chr.17: 49995079-49995079 on Build GRCh38
rs72034312 Chr.17: 49995356-49995357 on Build GRCh38
rs867171170 Chr.17: 49995433-49995433 on Build GRCh38
rs954695166 Chr.17: 49995402-49995402 on Build GRCh38
rs931090957 Chr.17: 49995367-49995367 on Build GRCh38
rs761819584 Chr.17: 49995021-49995021 on Build GRCh38
rs1018009161 Chr.17: 49995362-49995362 on Build GRCh38
rs62059977 Chr.17: 49995370-49995370 on Build GRCh38
rs889268885 Chr.17: 49995352-49995352 on Build GRCh38
rs576319593 Chr.17: 49995094-49995094 on Build GRCh38
rs939341009 Chr.17: 49995293-49995293 on Build GRCh38
rs908044828 Chr.17: 49995154-49995154 on Build GRCh38
rs973666468 Chr.17: 49995180-49995180 on Build GRCh38
rs918488139 Chr.17: 49995183-49995183 on Build GRCh38
rs796585153 Chr.17: 49995355-49995356 on Build GRCh38
rs34783534 Chr.17: 49995187-49995187 on Build GRCh38
rs34970236 Chr.17: 49995102-49995103 on Build GRCh38
rs949805501 Chr.17: 49995236-49995236 on Build GRCh38
rs796947150 Chr.17: 49995367-49995368 on Build GRCh38
rs1006673069 Chr.17: 49995360-49995360 on Build GRCh38
rs751977901 Chr.17: 49995050-49995050 on Build GRCh38
rs753696997 Chr.17: 49995036-49995036 on Build GRCh38
rs954993315 Chr.17: 49995145-49995145 on Build GRCh38
rs150591955 Chr.17: 49995130-49995130 on Build GRCh38
rs1029449246 Chr.17: 49995401-49995402 on Build GRCh38
rs10459948 Chr.17: 49995132-49995132 on Build GRCh38
rs546133632 Chr.17: 49995223-49995223 on Build GRCh38
rs977088019 Chr.17: 49995178-49995178 on Build GRCh38
rs916917114 Chr.17: 49995224-49995224 on Build GRCh38
rs190434497 Chr.17: 49995462-49995462 on Build GRCh38
rs946950692 Chr.17: 49995343-49995343 on Build GRCh38
rs10671429 Chr.17: 49995433-49995434 on Build GRCh38
rs10585123 Chr.17: 49995357-49995358 on Build GRCh38
rs983938947 Chr.17: 49995188-49995188 on Build GRCh38
rs758755309 Chr.17: 49995040-49995040 on Build GRCh38
rs773407489 Chr.17: 49995356-49995356 on Build GRCh38
rs1041412653 Chr.17: 49995240-49995240 on Build GRCh38
rs796225845 Chr.17: 49995372-49995373 on Build GRCh38
rs764775985 Chr.17: 49995367-49995368 on Build GRCh38
rs780565504 Chr.17: 49995041-49995041 on Build GRCh38
rs1025990920 Chr.17: 49995392-49995392 on Build GRCh38
rs765400612 Chr.17: 49995028-49995028 on Build GRCh38
rs985090075 Chr.17: 49995003-49995003 on Build GRCh38
rs376831875 Chr.17: 49995038-49995038 on Build GRCh38
rs561223399 Chr.17: 49995371-49995372 on Build GRCh38
rs559276921 Chr.17: 49995415-49995415 on Build GRCh38
rs962172166 Chr.17: 49995172-49995172 on Build GRCh38
rs1017977546 Chr.17: 49995140-49995140 on Build GRCh38
rs769426660 Chr.17: 49995004-49995004 on Build GRCh38
rs756722786 Chr.17: 49995127-49995127 on Build GRCh38
rs866556249 Chr.17: 49995464-49995464 on Build GRCh38
rs909712654 Chr.17: 49995200-49995200 on Build GRCh38
rs796259046 Chr.17: 49995357-49995358 on Build GRCh38
rs987551931 Chr.17: 49995153-49995153 on Build GRCh38
rs892643725 Chr.17: 49995128-49995128 on Build GRCh38
rs1028981560 Chr.17: 49995136-49995136 on Build GRCh38
rs112448353 Chr.17: 49995371-49995371 on Build GRCh38
rs951365307 Chr.17: 49995430-49995430 on Build GRCh38
rs962225958 Chr.17: 49995455-49995456 on Build GRCh38
rs1014989235 Chr.17: 49995441-49995441 on Build GRCh38
rs903037139 Chr.17: 49995330-49995330 on Build GRCh38
rs2278163 Chr.17: 49995062-49995062 on Build GRCh38
rs976677497 Chr.17: 49995034-49995034 on Build GRCh38
rs559617688 Chr.17: 49995250-49995250 on Build GRCh38
rs112507296 Chr.17: 49995009-49995009 on Build GRCh38
rs1053117931 Chr.17: 49995303-49995303 on Build GRCh38
rs532523216 Chr.17: 49995133-49995133 on Build GRCh38
rs927979771 Chr.17: 49995275-49995275 on Build GRCh38
rs753895089 Chr.17: 49995020-49995020 on Build GRCh38
rs756924776 Chr.17: 49995119-49995119 on Build GRCh38
rs576352687 Chr.17: 49995410-49995410 on Build GRCh38
rs764020124 Chr.17: 49995017-49995017 on Build GRCh38
rs892682756 Chr.17: 49995312-49995312 on Build GRCh38
rs111865260 Chr.17: 49995355-49995356 on Build GRCh38
rs796424166 Chr.17: 49995371-49995372 on Build GRCh38

View Full Product Details