Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 76079610-76080099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZACN
Entrez Gene ID
353174
Gene Name
zinc activated ion channel
Gene Aliases
L2, LGICZ, LGICZ1, ZAC, ZAC1
UniGene
Hs.714919
Gene Chromosome Location
Chr.17: 76079182-76082804 on Build GRCh38
Transcripts
NM_180990.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5031956 c.232C>G p.R78G Chr.17: 76079711-76079711 on Build GRCh38
7318155 c.360C>G p.L120L Chr.17: 76079980-76079980 on Build GRCh38
1710818 c.347G>A p.W116* Chr.17: 76079967-76079967 on Build GRCh38
3522469 c.248C>T p.S83F Chr.17: 76079727-76079727 on Build GRCh38
5387368 c.332C>T p.P111L Chr.17: 76079952-76079952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139684245 Chr.17: 76079934-76079934 on Build GRCh38
rs756471163 Chr.17: 76080011-76080011 on Build GRCh38
rs373901649 Chr.17: 76079650-76079650 on Build GRCh38
rs376718726 Chr.17: 76079893-76079893 on Build GRCh38
rs772452404 Chr.17: 76079946-76079946 on Build GRCh38
rs759113634 Chr.17: 76079764-76079764 on Build GRCh38
rs772392608 Chr.17: 76079654-76079654 on Build GRCh38
rs776065369 Chr.17: 76079973-76079973 on Build GRCh38
rs540383105 Chr.17: 76079954-76079954 on Build GRCh38
rs370157774 Chr.17: 76079666-76079666 on Build GRCh38
rs746255877 Chr.17: 76079943-76079943 on Build GRCh38
rs765210418 Chr.17: 76079906-76079906 on Build GRCh38
rs550251960 Chr.17: 76079721-76079721 on Build GRCh38
rs774719253 Chr.17: 76079757-76079757 on Build GRCh38
rs999147289 Chr.17: 76079675-76079675 on Build GRCh38
rs770422618 Chr.17: 76079678-76079678 on Build GRCh38
rs763477841 Chr.17: 76079744-76079744 on Build GRCh38
rs561686196 Chr.17: 76080037-76080037 on Build GRCh38
rs750232134 Chr.17: 76079844-76079844 on Build GRCh38
rs200146097 Chr.17: 76079858-76079858 on Build GRCh38
rs567890838 Chr.17: 76079694-76079694 on Build GRCh38
rs536879025 Chr.17: 76079706-76079706 on Build GRCh38
rs749381325 Chr.17: 76079936-76079936 on Build GRCh38
rs746589365 Chr.17: 76079876-76079877 on Build GRCh38
rs1038845061 Chr.17: 76080077-76080077 on Build GRCh38
rs765128190 Chr.17: 76079986-76079986 on Build GRCh38
rs780466837 Chr.17: 76080045-76080045 on Build GRCh38
rs768296457 Chr.17: 76079957-76079957 on Build GRCh38
rs778816513 Chr.17: 76080026-76080026 on Build GRCh38
rs369660049 Chr.17: 76079651-76079651 on Build GRCh38
rs777951932 Chr.17: 76079865-76079865 on Build GRCh38
rs777300001 Chr.17: 76079695-76079695 on Build GRCh38
rs372079220 Chr.17: 76079859-76079859 on Build GRCh38
rs373419607 Chr.17: 76079663-76079663 on Build GRCh38
rs892028711 Chr.17: 76079792-76079792 on Build GRCh38
rs903492496 Chr.17: 76079997-76079997 on Build GRCh38
rs200269075 Chr.17: 76080047-76080047 on Build GRCh38
rs752991699 Chr.17: 76079932-76079932 on Build GRCh38
rs762934534 Chr.17: 76079994-76079994 on Build GRCh38
rs893001065 Chr.17: 76079697-76079697 on Build GRCh38
rs148442220 Chr.17: 76079947-76079947 on Build GRCh38
rs771757981 Chr.17: 76079879-76079879 on Build GRCh38
rs757164319 Chr.17: 76079838-76079838 on Build GRCh38
rs746672363 Chr.17: 76079955-76079955 on Build GRCh38
rs1019453333 Chr.17: 76080055-76080055 on Build GRCh38
rs548207801 Chr.17: 76079667-76079667 on Build GRCh38
rs775550156 Chr.17: 76079660-76079660 on Build GRCh38
rs764362832 Chr.17: 76080014-76080014 on Build GRCh38
rs775089186 Chr.17: 76079883-76079883 on Build GRCh38
rs767150915 Chr.17: 76079766-76079766 on Build GRCh38
rs746392242 Chr.17: 76080027-76080027 on Build GRCh38
rs903605986 Chr.17: 76079826-76079826 on Build GRCh38
rs752134812 Chr.17: 76079699-76079699 on Build GRCh38
rs936429549 Chr.17: 76080000-76080000 on Build GRCh38
rs548448874 Chr.17: 76080021-76080021 on Build GRCh38
rs1000579419 Chr.17: 76079848-76079848 on Build GRCh38
rs756108910 Chr.17: 76079701-76079701 on Build GRCh38
rs200681257 Chr.17: 76079931-76079931 on Build GRCh38
rs779784382 Chr.17: 76079720-76079728 on Build GRCh38
rs981564472 Chr.17: 76080015-76080015 on Build GRCh38
rs373191028 Chr.17: 76079933-76079933 on Build GRCh38
rs758238721 Chr.17: 76079929-76079929 on Build GRCh38
rs997149808 Chr.17: 76079747-76079747 on Build GRCh38
rs113860382 Chr.17: 76079903-76079903 on Build GRCh38
rs549500563 Chr.17: 76079995-76079998 on Build GRCh38
rs749772531 Chr.17: 76079870-76079870 on Build GRCh38
rs34700740 Chr.17: 76080047-76080047 on Build GRCh38
rs377543076 Chr.17: 76079665-76079665 on Build GRCh38
rs773772239 Chr.17: 76079683-76079683 on Build GRCh38
rs201792263 Chr.17: 76079894-76079894 on Build GRCh38
rs140302218 Chr.17: 76079711-76079711 on Build GRCh38
rs776475018 Chr.17: 76080068-76080071 on Build GRCh38
rs776144412 Chr.17: 76079900-76079900 on Build GRCh38
rs746228334 Chr.17: 76079878-76079878 on Build GRCh38
rs770473701 Chr.17: 76079729-76079729 on Build GRCh38
rs529296856 Chr.17: 76079995-76079995 on Build GRCh38
rs376809476 Chr.17: 76079938-76079938 on Build GRCh38
rs199817656 Chr.17: 76080043-76080043 on Build GRCh38
rs878985790 Chr.17: 76079966-76079966 on Build GRCh38
rs781491291 Chr.17: 76079724-76079724 on Build GRCh38
rs760152946 Chr.17: 76079797-76079797 on Build GRCh38
rs114867401 Chr.17: 76079793-76079793 on Build GRCh38
rs764054967 Chr.17: 76079798-76079798 on Build GRCh38
rs773825209 Chr.17: 76079730-76079730 on Build GRCh38
rs778725073 Chr.17: 76079712-76079712 on Build GRCh38
rs1053481536 Chr.17: 76079904-76079904 on Build GRCh38
rs760047265 Chr.17: 76079802-76079802 on Build GRCh38
rs750236707 Chr.17: 76079911-76079911 on Build GRCh38
rs889524314 Chr.17: 76080031-76080031 on Build GRCh38
rs774644336 Chr.17: 76079872-76079872 on Build GRCh38
rs142551926 Chr.17: 76079974-76079974 on Build GRCh38
rs572773620 Chr.17: 76079854-76079854 on Build GRCh38
rs377430797 Chr.17: 76079690-76079690 on Build GRCh38
rs150992971 Chr.17: 76079990-76079990 on Build GRCh38
rs753746098 Chr.17: 76079837-76079837 on Build GRCh38
rs760277532 Chr.17: 76079890-76079890 on Build GRCh38
rs778581425 Chr.17: 76079841-76079841 on Build GRCh38
rs947288962 Chr.17: 76080050-76080050 on Build GRCh38
rs570259408 Chr.17: 76079726-76079726 on Build GRCh38
rs780904634 Chr.17: 76080004-76080005 on Build GRCh38
rs766096852 Chr.17: 76079689-76079689 on Build GRCh38
rs755216058 Chr.17: 76079845-76079845 on Build GRCh38
rs144683704 Chr.17: 76079939-76079939 on Build GRCh38
rs142902667 Chr.17: 76079751-76079751 on Build GRCh38
rs762608100 Chr.17: 76079843-76079843 on Build GRCh38
rs374179633 Chr.17: 76079687-76079687 on Build GRCh38

View Full Product Details