Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11549836-11550330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNN1
Entrez Gene ID
1264
Gene Name
calponin 1
Gene Aliases
HEL-S-14, SMCC, Sm-Calp
UniGene
Hs.465929
Gene Chromosome Location
Chr.19: 11538717-11550323 on Build GRCh38
Transcripts
NM_001299.5 NM_001308341.1 NM_001308342.1 XM_005259741.2 XM_017026289.1

SNP Detail

SNP ID Chromosome Location
rs150280468 Chr.19: 11549954-11549954 on Build GRCh38
rs867506039 Chr.19: 11550045-11550045 on Build GRCh38
rs892960276 Chr.19: 11550119-11550119 on Build GRCh38
rs542006621 Chr.19: 11549967-11549967 on Build GRCh38
rs1011510824 Chr.19: 11550157-11550157 on Build GRCh38
rs569825499 Chr.19: 11550166-11550166 on Build GRCh38
rs537222572 Chr.19: 11550044-11550044 on Build GRCh38
rs950545303 Chr.19: 11550277-11550278 on Build GRCh38
rs74759333 Chr.19: 11550056-11550056 on Build GRCh38
rs902471036 Chr.19: 11549871-11549871 on Build GRCh38
rs772804068 Chr.19: 11550051-11550051 on Build GRCh38
rs750129845 Chr.19: 11550114-11550116 on Build GRCh38
rs117822289 Chr.19: 11550211-11550211 on Build GRCh38
rs960450359 Chr.19: 11550246-11550246 on Build GRCh38
rs1055276372 Chr.19: 11549991-11549991 on Build GRCh38
rs537256906 Chr.19: 11550206-11550206 on Build GRCh38
rs190733600 Chr.19: 11549974-11549974 on Build GRCh38
rs754780320 Chr.19: 11550200-11550200 on Build GRCh38
rs761796263 Chr.19: 11550103-11550103 on Build GRCh38
rs778113659 Chr.19: 11549990-11549990 on Build GRCh38
rs1008710297 Chr.19: 11549960-11549960 on Build GRCh38
rs550989532 Chr.19: 11550110-11550110 on Build GRCh38
rs766897866 Chr.19: 11550288-11550288 on Build GRCh38
rs559771997 Chr.19: 11549916-11549916 on Build GRCh38
rs916714423 Chr.19: 11550034-11550034 on Build GRCh38
rs574563838 Chr.19: 11550032-11550032 on Build GRCh38
rs35986026 Chr.19: 11549964-11549964 on Build GRCh38
rs576728816 Chr.19: 11549888-11549888 on Build GRCh38
rs565826768 Chr.19: 11550073-11550073 on Build GRCh38
rs1043752879 Chr.19: 11549865-11549865 on Build GRCh38
rs1004972547 Chr.19: 11550229-11550229 on Build GRCh38
rs552220753 Chr.19: 11550023-11550023 on Build GRCh38
rs749880254 Chr.19: 11550000-11550000 on Build GRCh38
rs771411977 Chr.19: 11550012-11550012 on Build GRCh38
rs775588528 Chr.19: 11550055-11550055 on Build GRCh38
rs956254098 Chr.19: 11549978-11549978 on Build GRCh38
rs1128572 Chr.19: 11550301-11550301 on Build GRCh38
rs1034661421 Chr.19: 11550230-11550230 on Build GRCh38
rs916599514 Chr.19: 11550020-11550020 on Build GRCh38
rs935342005 Chr.19: 11549877-11549877 on Build GRCh38
rs534305314 Chr.19: 11550304-11550304 on Build GRCh38
rs994627263 Chr.19: 11550208-11550208 on Build GRCh38
rs531022778 Chr.19: 11549975-11549975 on Build GRCh38
rs72995011 Chr.19: 11550111-11550111 on Build GRCh38
rs79983076 Chr.19: 11550072-11550072 on Build GRCh38
rs138581507 Chr.19: 11549894-11549894 on Build GRCh38
rs891225716 Chr.19: 11550134-11550134 on Build GRCh38
rs576896675 Chr.19: 11550226-11550226 on Build GRCh38
rs924779301 Chr.19: 11549972-11549972 on Build GRCh38
rs758802518 Chr.19: 11550165-11550165 on Build GRCh38
rs1027325689 Chr.19: 11549985-11549985 on Build GRCh38
rs750249699 Chr.19: 11550201-11550201 on Build GRCh38
rs1035205471 Chr.19: 11550017-11550017 on Build GRCh38
rs902633421 Chr.19: 11550071-11550071 on Build GRCh38
rs1029990418 Chr.19: 11550251-11550251 on Build GRCh38
rs1051085234 Chr.19: 11550076-11550076 on Build GRCh38
rs983124871 Chr.19: 11549994-11549994 on Build GRCh38

View Full Product Details