Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41116376-41116891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2F1
Entrez Gene ID
1572
Gene Name
cytochrome P450 family 2 subfamily F member 1
Gene Aliases
C2F1, CYP2F, CYPIIF1
UniGene
Hs.558318
Gene Chromosome Location
Chr.19: 41114292-41128381 on Build GRCh38
Transcripts
NM_000774.4 NR_135528.1 XM_011526551.2 XM_011526552.2 XM_011526553.2 XM_011526554.2 XM_011526555.2 XM_017026384.1 XM_017026385.1 XM_017026386.1 XM_017026387.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6084927 c.173T>A p.L58Q Chr.19: 41116456-41116456 on Build GRCh38
4935280 c.228G>A p.V76V Chr.19: 41116511-41116511 on Build GRCh38
5529519 c.291C>T p.G97G Chr.19: 41116574-41116574 on Build GRCh38
233920 c.334+9C>T p.? Chr.19: 41116626-41116626 on Build GRCh38
4613487 c.310delT p.F105fs*20 Chr.19: 41116593-41116593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766915556 Chr.19: 41116413-41116413 on Build GRCh38
rs370948653 Chr.19: 41116665-41116665 on Build GRCh38
rs757071675 Chr.19: 41116433-41116433 on Build GRCh38
rs57670668 Chr.19: 41116576-41116576 on Build GRCh38
rs989878161 Chr.19: 41116434-41116434 on Build GRCh38
rs201282398 Chr.19: 41116659-41116659 on Build GRCh38
rs774876663 Chr.19: 41116628-41116628 on Build GRCh38
rs915311064 Chr.19: 41116437-41116437 on Build GRCh38
rs749302242 Chr.19: 41116828-41116828 on Build GRCh38
rs183407114 Chr.19: 41116754-41116754 on Build GRCh38
rs564557979 Chr.19: 41116630-41116630 on Build GRCh38
rs755721234 Chr.19: 41116418-41116418 on Build GRCh38
rs779982873 Chr.19: 41116578-41116578 on Build GRCh38
rs374038071 Chr.19: 41116647-41116647 on Build GRCh38
rs879587043 Chr.19: 41116830-41116830 on Build GRCh38
rs777504485 Chr.19: 41116420-41116420 on Build GRCh38
rs111817081 Chr.19: 41116627-41116627 on Build GRCh38
rs900707404 Chr.19: 41116555-41116555 on Build GRCh38
rs868422588 Chr.19: 41116587-41116587 on Build GRCh38
rs376200584 Chr.19: 41116648-41116648 on Build GRCh38
rs144909127 Chr.19: 41116506-41116506 on Build GRCh38
rs769759085 Chr.19: 41116730-41116730 on Build GRCh38
rs781435044 Chr.19: 41116610-41116610 on Build GRCh38
rs375742886 Chr.19: 41116641-41116641 on Build GRCh38
rs959223174 Chr.19: 41116679-41116679 on Build GRCh38
rs1039303112 Chr.19: 41116547-41116547 on Build GRCh38
rs1036458322 Chr.19: 41116823-41116823 on Build GRCh38
rs748348532 Chr.19: 41116615-41116615 on Build GRCh38
rs766257652 Chr.19: 41116667-41116667 on Build GRCh38
rs201870178 Chr.19: 41116642-41116642 on Build GRCh38
rs944684079 Chr.19: 41116766-41116766 on Build GRCh38
rs763863949 Chr.19: 41116554-41116554 on Build GRCh38
rs967556477 Chr.19: 41116678-41116678 on Build GRCh38
rs757994193 Chr.19: 41116465-41116466 on Build GRCh38
rs940744457 Chr.19: 41116793-41116793 on Build GRCh38
rs915851281 Chr.19: 41116717-41116717 on Build GRCh38
rs770202369 Chr.19: 41116617-41116617 on Build GRCh38
rs752465443 Chr.19: 41116429-41116429 on Build GRCh38
rs923229196 Chr.19: 41116863-41116863 on Build GRCh38
rs12975986 Chr.19: 41116710-41116710 on Build GRCh38
rs772249109 Chr.19: 41116458-41116458 on Build GRCh38
rs779989368 Chr.19: 41116460-41116460 on Build GRCh38
rs989250358 Chr.19: 41116690-41116690 on Build GRCh38
rs745767766 Chr.19: 41116450-41116450 on Build GRCh38
rs747044071 Chr.19: 41116464-41116464 on Build GRCh38
rs762195176 Chr.19: 41116503-41116503 on Build GRCh38
rs372152390 Chr.19: 41116698-41116698 on Build GRCh38
rs1039333788 Chr.19: 41116832-41116832 on Build GRCh38
rs1033543850 Chr.19: 41116675-41116675 on Build GRCh38
rs151222092 Chr.19: 41116577-41116577 on Build GRCh38
rs796628336 Chr.19: 41116668-41116668 on Build GRCh38
rs1004292236 Chr.19: 41116629-41116629 on Build GRCh38
rs776783483 Chr.19: 41116487-41116487 on Build GRCh38
rs749604339 Chr.19: 41116626-41116626 on Build GRCh38
rs760208996 Chr.19: 41116530-41116530 on Build GRCh38
rs186281561 Chr.19: 41116624-41116624 on Build GRCh38
rs977519814 Chr.19: 41116475-41116475 on Build GRCh38
rs768698647 Chr.19: 41116836-41116836 on Build GRCh38
rs930809524 Chr.19: 41116848-41116848 on Build GRCh38
rs753703363 Chr.19: 41116430-41116430 on Build GRCh38
rs199588250 Chr.19: 41116575-41116575 on Build GRCh38
rs928536385 Chr.19: 41116501-41116501 on Build GRCh38
rs908392247 Chr.19: 41116785-41116785 on Build GRCh38
rs777323567 Chr.19: 41116521-41116521 on Build GRCh38
rs753504147 Chr.19: 41116574-41116574 on Build GRCh38
rs984386916 Chr.19: 41116781-41116781 on Build GRCh38
rs922153442 Chr.19: 41116845-41116845 on Build GRCh38
rs76335535 Chr.19: 41116590-41116590 on Build GRCh38
rs763356097 Chr.19: 41116520-41116520 on Build GRCh38
rs191496638 Chr.19: 41116715-41116715 on Build GRCh38
rs534149886 Chr.19: 41116844-41116844 on Build GRCh38
rs145103345 Chr.19: 41116504-41116504 on Build GRCh38
rs138177478 Chr.19: 41116756-41116756 on Build GRCh38
rs1035289129 Chr.19: 41116709-41116709 on Build GRCh38
rs768879115 Chr.19: 41116484-41116484 on Build GRCh38
rs779032097 Chr.19: 41116449-41116449 on Build GRCh38

View Full Product Details