Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48473813-48474309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH2
Entrez Gene ID
9266
Gene Name
cytohesin 2
Gene Aliases
ARNO, CTS18, CTS18.1, PSCD2, PSCD2L, SEC7L, Sec7p-L, Sec7p-like
UniGene
Hs.144011
Gene Chromosome Location
Chr.19: 48469208-48482314 on Build GRCh38
Transcripts
NM_004228.6 NM_017457.5 XM_006723472.2 XM_006723473.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7333798 c.576C>T p.V192V Chr.19: 48474210-48474210 on Build GRCh38
4973656 c.664G>A p.E222K Chr.19: 48474298-48474298 on Build GRCh38
6191012 c.664G>A p.E222K Chr.19: 48474298-48474298 on Build GRCh38
7363588 c.671G>T p.G224V Chr.19: 48474305-48474305 on Build GRCh38
7113266 c.588C>T p.N196N Chr.19: 48474222-48474222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs28582663 Chr.19: 48473932-48473932 on Build GRCh38
rs61748773 Chr.19: 48474222-48474222 on Build GRCh38
rs755432500 Chr.19: 48474052-48474052 on Build GRCh38
rs28433711 Chr.19: 48474125-48474125 on Build GRCh38
rs765488390 Chr.19: 48474027-48474027 on Build GRCh38
rs746574144 Chr.19: 48473963-48473963 on Build GRCh38
rs542894323 Chr.19: 48473860-48473860 on Build GRCh38
rs753565456 Chr.19: 48474205-48474205 on Build GRCh38
rs907765893 Chr.19: 48474070-48474070 on Build GRCh38
rs771977589 Chr.19: 48474152-48474152 on Build GRCh38
rs769951654 Chr.19: 48474248-48474248 on Build GRCh38
rs538695510 Chr.19: 48474193-48474193 on Build GRCh38
rs368619143 Chr.19: 48473897-48473897 on Build GRCh38
rs760748383 Chr.19: 48473879-48473879 on Build GRCh38
rs202097541 Chr.19: 48474269-48474269 on Build GRCh38
rs774399359 Chr.19: 48474021-48474021 on Build GRCh38
rs975824182 Chr.19: 48474177-48474177 on Build GRCh38
rs562697189 Chr.19: 48474100-48474100 on Build GRCh38
rs766914095 Chr.19: 48474034-48474034 on Build GRCh38
rs373886599 Chr.19: 48473865-48473865 on Build GRCh38
rs967635752 Chr.19: 48473875-48473875 on Build GRCh38
rs978735449 Chr.19: 48473887-48473887 on Build GRCh38
rs780015719 Chr.19: 48474231-48474231 on Build GRCh38
rs372095603 Chr.19: 48474174-48474174 on Build GRCh38
rs895557450 Chr.19: 48474130-48474130 on Build GRCh38
rs200137114 Chr.19: 48474258-48474258 on Build GRCh38
rs1020483249 Chr.19: 48474140-48474140 on Build GRCh38
rs148173211 Chr.19: 48473989-48473989 on Build GRCh38
rs369066635 Chr.19: 48474026-48474026 on Build GRCh38
rs537835463 Chr.19: 48473979-48473979 on Build GRCh38
rs573120638 Chr.19: 48473892-48473892 on Build GRCh38
rs903418691 Chr.19: 48474155-48474155 on Build GRCh38
rs1033420646 Chr.19: 48474164-48474164 on Build GRCh38
rs766756536 Chr.19: 48474183-48474183 on Build GRCh38
rs377111289 Chr.19: 48473987-48473987 on Build GRCh38
rs780584269 Chr.19: 48474056-48474056 on Build GRCh38
rs973326493 Chr.19: 48474264-48474264 on Build GRCh38
rs373048304 Chr.19: 48474268-48474268 on Build GRCh38
rs372308279 Chr.19: 48473933-48473933 on Build GRCh38
rs557093699 Chr.19: 48474201-48474201 on Build GRCh38
rs751866749 Chr.19: 48474184-48474184 on Build GRCh38
rs757452084 Chr.19: 48473929-48473929 on Build GRCh38
rs1006139156 Chr.19: 48473967-48473967 on Build GRCh38
rs774765800 Chr.19: 48474265-48474265 on Build GRCh38
rs1020509403 Chr.19: 48473843-48473843 on Build GRCh38
rs532630301 Chr.19: 48474029-48474029 on Build GRCh38
rs1047982304 Chr.19: 48474245-48474245 on Build GRCh38
rs760826182 Chr.19: 48473925-48473925 on Build GRCh38
rs776128809 Chr.19: 48473870-48473870 on Build GRCh38
rs766467656 Chr.19: 48474003-48474003 on Build GRCh38
rs1010784436 Chr.19: 48474086-48474086 on Build GRCh38
rs937077634 Chr.19: 48474124-48474124 on Build GRCh38
rs556212908 Chr.19: 48474037-48474037 on Build GRCh38
rs755760100 Chr.19: 48473992-48473992 on Build GRCh38
rs755790296 Chr.19: 48474068-48474068 on Build GRCh38
rs567141309 Chr.19: 48474134-48474134 on Build GRCh38
rs955466370 Chr.19: 48474213-48474213 on Build GRCh38
rs768351607 Chr.19: 48473866-48473866 on Build GRCh38
rs775781718 Chr.19: 48473914-48473914 on Build GRCh38
rs940629100 Chr.19: 48474083-48474083 on Build GRCh38
rs776648598 Chr.19: 48474019-48474020 on Build GRCh38
rs769511918 Chr.19: 48473871-48473871 on Build GRCh38
rs868252335 Chr.19: 48474286-48474286 on Build GRCh38
rs759505371 Chr.19: 48474022-48474022 on Build GRCh38
rs138838239 Chr.19: 48474273-48474273 on Build GRCh38
rs758967193 Chr.19: 48473956-48473956 on Build GRCh38
rs764450200 Chr.19: 48473927-48473927 on Build GRCh38
rs752185512 Chr.19: 48474065-48474065 on Build GRCh38
rs752209282 Chr.19: 48474196-48474196 on Build GRCh38
rs752389593 Chr.19: 48473957-48473957 on Build GRCh38
rs548442097 Chr.19: 48474133-48474133 on Build GRCh38
rs745813686 Chr.19: 48474138-48474138 on Build GRCh38
rs759578321 Chr.19: 48474288-48474299 on Build GRCh38
rs771078990 Chr.19: 48474261-48474261 on Build GRCh38
rs748213849 Chr.19: 48474236-48474236 on Build GRCh38
rs1014398047 Chr.19: 48474137-48474137 on Build GRCh38
rs773229556 Chr.19: 48474257-48474257 on Build GRCh38
rs771051877 Chr.19: 48474009-48474009 on Build GRCh38
rs754455899 Chr.19: 48473971-48473971 on Build GRCh38
rs368995401 Chr.19: 48474211-48474211 on Build GRCh38
rs773389277 Chr.19: 48474170-48474170 on Build GRCh38
rs763958248 Chr.19: 48474280-48474280 on Build GRCh38
rs780717608 Chr.19: 48473976-48473976 on Build GRCh38
rs546573877 Chr.19: 48474166-48474166 on Build GRCh38
rs920251408 Chr.19: 48474266-48474266 on Build GRCh38
rs773014453 Chr.19: 48473885-48473885 on Build GRCh38
rs61497330 Chr.19: 48474186-48474186 on Build GRCh38
rs898858476 Chr.19: 48474114-48474114 on Build GRCh38
rs1804728 Chr.19: 48474244-48474244 on Build GRCh38
rs544309668 Chr.19: 48474094-48474094 on Build GRCh38
rs181597580 Chr.19: 48474156-48474156 on Build GRCh38
rs188841092 Chr.19: 48474023-48474023 on Build GRCh38
rs141113238 Chr.19: 48474208-48474208 on Build GRCh38
rs769956837 Chr.19: 48474169-48474169 on Build GRCh38
rs887658349 Chr.19: 48473935-48473935 on Build GRCh38
rs371423693 Chr.19: 48473864-48473864 on Build GRCh38
rs758840872 Chr.19: 48473862-48473862 on Build GRCh38
rs756912419 Chr.19: 48474207-48474207 on Build GRCh38
rs375723005 Chr.19: 48474204-48474204 on Build GRCh38
rs763279556 Chr.19: 48474030-48474030 on Build GRCh38
rs749092477 Chr.19: 48474006-48474006 on Build GRCh38

View Full Product Details