Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36730953-36731457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIM2
Entrez Gene ID
6493
Gene Name
single-minded family bHLH transcription factor 2
Gene Aliases
HMC13F06, HMC29C01, SIM, bHLHe15
UniGene
Hs.146186
Gene Chromosome Location
Chr.21: 36699117-36750219 on Build GRCh38
Transcripts
NM_005069.4 NM_009586.3 XM_011529694.1 XM_017028442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7414187 c.793C>A p.L265I Chr.21: 36731094-36731094 on Build GRCh38
7402625 c.791C>A p.T264N Chr.21: 36731092-36731092 on Build GRCh38
4391322 c.758C>T p.T253M Chr.21: 36731059-36731059 on Build GRCh38
7414186 c.793C>A p.L265I Chr.21: 36731094-36731094 on Build GRCh38
323333 c.829C>T p.L277F Chr.21: 36731130-36731130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770808205 Chr.21: 36731090-36731090 on Build GRCh38
rs781203530 Chr.21: 36731014-36731014 on Build GRCh38
rs756792453 Chr.21: 36731163-36731163 on Build GRCh38
rs747167310 Chr.21: 36731072-36731072 on Build GRCh38
rs1018669135 Chr.21: 36730979-36730979 on Build GRCh38
rs781638907 Chr.21: 36731171-36731171 on Build GRCh38
rs554853935 Chr.21: 36731342-36731342 on Build GRCh38
rs182926138 Chr.21: 36731280-36731280 on Build GRCh38
rs929035264 Chr.21: 36731330-36731330 on Build GRCh38
rs751583781 Chr.21: 36731002-36731002 on Build GRCh38
rs368492200 Chr.21: 36731177-36731177 on Build GRCh38
rs777141819 Chr.21: 36731117-36731117 on Build GRCh38
rs769091808 Chr.21: 36731112-36731112 on Build GRCh38
rs535175817 Chr.21: 36731015-36731016 on Build GRCh38
rs368622558 Chr.21: 36731133-36731133 on Build GRCh38
rs1025939786 Chr.21: 36731190-36731190 on Build GRCh38
rs773334944 Chr.21: 36731063-36731063 on Build GRCh38
rs762475183 Chr.21: 36731387-36731387 on Build GRCh38
rs1015671494 Chr.21: 36731106-36731106 on Build GRCh38
rs1005911514 Chr.21: 36731349-36731349 on Build GRCh38
rs748943381 Chr.21: 36731085-36731085 on Build GRCh38
rs376029267 Chr.21: 36731069-36731069 on Build GRCh38
rs765448691 Chr.21: 36731060-36731060 on Build GRCh38
rs116591818 Chr.21: 36731301-36731301 on Build GRCh38
rs979212526 Chr.21: 36731176-36731176 on Build GRCh38
rs1044330759 Chr.21: 36731354-36731354 on Build GRCh38
rs544770673 Chr.21: 36731340-36731341 on Build GRCh38
rs757931635 Chr.21: 36731167-36731167 on Build GRCh38
rs376055256 Chr.21: 36730994-36730994 on Build GRCh38
rs1022904739 Chr.21: 36731218-36731218 on Build GRCh38
rs773456534 Chr.21: 36731121-36731121 on Build GRCh38
rs558642509 Chr.21: 36731015-36731015 on Build GRCh38
rs776912680 Chr.21: 36731052-36731052 on Build GRCh38
rs777315620 Chr.21: 36731027-36731027 on Build GRCh38
rs1044527681 Chr.21: 36731316-36731316 on Build GRCh38
rs763155198 Chr.21: 36731126-36731126 on Build GRCh38
rs1007234724 Chr.21: 36730977-36730977 on Build GRCh38
rs763853956 Chr.21: 36731152-36731152 on Build GRCh38
rs139320055 Chr.21: 36731120-36731120 on Build GRCh38
rs150004951 Chr.21: 36731166-36731166 on Build GRCh38
rs114671311 Chr.21: 36731281-36731281 on Build GRCh38
rs867898450 Chr.21: 36731304-36731304 on Build GRCh38
rs771231823 Chr.21: 36731178-36731178 on Build GRCh38
rs372352643 Chr.21: 36731162-36731162 on Build GRCh38
rs755894692 Chr.21: 36731079-36731079 on Build GRCh38
rs569628867 Chr.21: 36730998-36730998 on Build GRCh38
rs531153366 Chr.21: 36731279-36731279 on Build GRCh38
rs377248274 Chr.21: 36731193-36731193 on Build GRCh38
rs370463435 Chr.21: 36731037-36731037 on Build GRCh38
rs528676401 Chr.21: 36731225-36731225 on Build GRCh38
rs748793311 Chr.21: 36731036-36731036 on Build GRCh38
rs149786959 Chr.21: 36731111-36731111 on Build GRCh38
rs775954129 Chr.21: 36731046-36731046 on Build GRCh38
rs150823127 Chr.21: 36731105-36731105 on Build GRCh38
rs370067255 Chr.21: 36731174-36731174 on Build GRCh38
rs779997341 Chr.21: 36731001-36731001 on Build GRCh38
rs561924014 Chr.21: 36731272-36731272 on Build GRCh38
rs146003844 Chr.21: 36731226-36731226 on Build GRCh38
rs1050982630 Chr.21: 36731243-36731243 on Build GRCh38
rs1002742961 Chr.21: 36731236-36731236 on Build GRCh38
rs139841287 Chr.21: 36731151-36731151 on Build GRCh38
rs574567165 Chr.21: 36731346-36731346 on Build GRCh38
rs774518103 Chr.21: 36731136-36731136 on Build GRCh38
rs558189874 Chr.21: 36731316-36731337 on Build GRCh38
rs113634109 Chr.21: 36731408-36731408 on Build GRCh38
rs777452609 Chr.21: 36731084-36731084 on Build GRCh38
rs572270415 Chr.21: 36731051-36731051 on Build GRCh38
rs763053811 Chr.21: 36731065-36731065 on Build GRCh38
rs774747134 Chr.21: 36731184-36731184 on Build GRCh38
rs748647360 Chr.21: 36731173-36731173 on Build GRCh38
rs554792457 Chr.21: 36731071-36731071 on Build GRCh38
rs772227381 Chr.21: 36731066-36731066 on Build GRCh38
rs147904157 Chr.21: 36731138-36731138 on Build GRCh38
rs778548369 Chr.21: 36731165-36731165 on Build GRCh38
rs1006111359 Chr.21: 36731203-36731203 on Build GRCh38
rs199502401 Chr.21: 36731134-36731134 on Build GRCh38
rs386818319 Chr.21: 36731316-36731343 on Build GRCh38
rs932374400 Chr.21: 36731219-36731219 on Build GRCh38
rs552319378 Chr.21: 36731303-36731303 on Build GRCh38
rs887417395 Chr.21: 36731348-36731348 on Build GRCh38
rs2250702 Chr.21: 36731329-36731329 on Build GRCh38
rs767523762 Chr.21: 36731147-36731147 on Build GRCh38
rs956080892 Chr.21: 36731433-36731433 on Build GRCh38
rs1023354909 Chr.21: 36731115-36731115 on Build GRCh38
rs755736760 Chr.21: 36731020-36731020 on Build GRCh38
rs369451752 Chr.21: 36731199-36731199 on Build GRCh38
rs960785689 Chr.21: 36731252-36731252 on Build GRCh38
rs943565343 Chr.21: 36731158-36731158 on Build GRCh38
rs534777844 Chr.21: 36731059-36731059 on Build GRCh38
rs189923799 Chr.21: 36731006-36731006 on Build GRCh38
rs905819383 Chr.21: 36731374-36731374 on Build GRCh38
rs188248879 Chr.21: 36731315-36731315 on Build GRCh38

View Full Product Details