Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36743296-36743802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIM2
Entrez Gene ID
6493
Gene Name
single-minded family bHLH transcription factor 2
Gene Aliases
HMC13F06, HMC29C01, SIM, bHLHe15
UniGene
Hs.146186
Gene Chromosome Location
Chr.21: 36699117-36750219 on Build GRCh38
Transcripts
NM_005069.4 NM_009586.3 XM_011529694.1 XM_017028442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7389934 c.1163C>T p.P388L Chr.21: 36743551-36743551 on Build GRCh38
284993 c.1167+8G>A p.? Chr.21: 36743563-36743563 on Build GRCh38
4101486 c.1044C>A p.S348S Chr.21: 36743432-36743432 on Build GRCh38
1751629 c.1098T>A p.T366T Chr.21: 36743486-36743486 on Build GRCh38
4101487 c.1044C>A p.S348S Chr.21: 36743432-36743432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376111637 Chr.21: 36743734-36743734 on Build GRCh38
rs888302224 Chr.21: 36743620-36743620 on Build GRCh38
rs374366409 Chr.21: 36743562-36743562 on Build GRCh38
rs550411332 Chr.21: 36743435-36743435 on Build GRCh38
rs1033167471 Chr.21: 36743744-36743744 on Build GRCh38
rs759299787 Chr.21: 36743381-36743381 on Build GRCh38
rs768274425 Chr.21: 36743567-36743567 on Build GRCh38
rs768329415 Chr.21: 36743482-36743482 on Build GRCh38
rs903429286 Chr.21: 36743738-36743738 on Build GRCh38
rs753867260 Chr.21: 36743553-36743553 on Build GRCh38
rs746714059 Chr.21: 36743565-36743565 on Build GRCh38
rs766797775 Chr.21: 36743501-36743501 on Build GRCh38
rs893870730 Chr.21: 36743463-36743463 on Build GRCh38
rs760284642 Chr.21: 36743389-36743389 on Build GRCh38
rs780104964 Chr.21: 36743339-36743339 on Build GRCh38
rs1000896539 Chr.21: 36743740-36743740 on Build GRCh38
rs570532752 Chr.21: 36743448-36743448 on Build GRCh38
rs943943649 Chr.21: 36743333-36743333 on Build GRCh38
rs775313694 Chr.21: 36743480-36743480 on Build GRCh38
rs377543172 Chr.21: 36743564-36743564 on Build GRCh38
rs1054270242 Chr.21: 36743768-36743768 on Build GRCh38
rs369823231 Chr.21: 36743366-36743366 on Build GRCh38
rs559218000 Chr.21: 36743338-36743338 on Build GRCh38
rs745663760 Chr.21: 36743469-36743469 on Build GRCh38
rs752968777 Chr.21: 36743540-36743540 on Build GRCh38
rs199980367 Chr.21: 36743375-36743375 on Build GRCh38
rs910910051 Chr.21: 36743319-36743319 on Build GRCh38
rs771431957 Chr.21: 36743580-36743580 on Build GRCh38
rs376777271 Chr.21: 36743378-36743378 on Build GRCh38
rs201838382 Chr.21: 36743377-36743377 on Build GRCh38
rs1041750082 Chr.21: 36743721-36743721 on Build GRCh38
rs755402134 Chr.21: 36743566-36743567 on Build GRCh38
rs376122916 Chr.21: 36743349-36743349 on Build GRCh38
rs138791805 Chr.21: 36743720-36743720 on Build GRCh38
rs766822804 Chr.21: 36743427-36743427 on Build GRCh38
rs778702815 Chr.21: 36743466-36743466 on Build GRCh38
rs1041692765 Chr.21: 36743392-36743392 on Build GRCh38
rs370355073 Chr.21: 36743551-36743551 on Build GRCh38
rs373442792 Chr.21: 36743348-36743348 on Build GRCh38
rs763202037 Chr.21: 36743422-36743422 on Build GRCh38
rs113607866 Chr.21: 36743560-36743560 on Build GRCh38
rs556036828 Chr.21: 36743581-36743581 on Build GRCh38
rs749055349 Chr.21: 36743369-36743369 on Build GRCh38
rs768470635 Chr.21: 36743608-36743608 on Build GRCh38
rs771705646 Chr.21: 36743476-36743476 on Build GRCh38
rs756174339 Chr.21: 36743549-36743549 on Build GRCh38
rs140977260 Chr.21: 36743526-36743526 on Build GRCh38
rs751005705 Chr.21: 36743421-36743421 on Build GRCh38
rs537345117 Chr.21: 36743777-36743777 on Build GRCh38
rs916896542 Chr.21: 36743703-36743703 on Build GRCh38
rs957792035 Chr.21: 36743508-36743508 on Build GRCh38
rs150148817 Chr.21: 36743563-36743563 on Build GRCh38
rs776209803 Chr.21: 36743572-36743572 on Build GRCh38
rs140462434 Chr.21: 36743367-36743367 on Build GRCh38
rs755157848 Chr.21: 36743434-36743434 on Build GRCh38
rs369519991 Chr.21: 36743402-36743402 on Build GRCh38
rs569427373 Chr.21: 36743607-36743607 on Build GRCh38
rs190470625 Chr.21: 36743737-36743737 on Build GRCh38
rs935920244 Chr.21: 36743741-36743741 on Build GRCh38
rs985203326 Chr.21: 36743542-36743542 on Build GRCh38
rs968039267 Chr.21: 36743441-36743441 on Build GRCh38
rs775686363 Chr.21: 36743590-36743590 on Build GRCh38
rs767082851 Chr.21: 36743387-36743387 on Build GRCh38
rs759924706 Chr.21: 36743583-36743583 on Build GRCh38
rs760821555 Chr.21: 36743602-36743602 on Build GRCh38
rs767930388 Chr.21: 36743585-36743585 on Build GRCh38
rs781395898 Chr.21: 36743359-36743359 on Build GRCh38
rs773839597 Chr.21: 36743486-36743486 on Build GRCh38
rs143650216 Chr.21: 36743388-36743388 on Build GRCh38
rs1055860136 Chr.21: 36743677-36743677 on Build GRCh38
rs895922646 Chr.21: 36743681-36743681 on Build GRCh38
rs763580102 Chr.21: 36743493-36743493 on Build GRCh38
rs750767448 Chr.21: 36743336-36743336 on Build GRCh38
rs949788690 Chr.21: 36743704-36743704 on Build GRCh38
rs538561116 Chr.21: 36743669-36743669 on Build GRCh38
rs558456257 Chr.21: 36743673-36743673 on Build GRCh38
rs760229622 Chr.21: 36743481-36743481 on Build GRCh38
rs752737842 Chr.21: 36743444-36743444 on Build GRCh38
rs897094093 Chr.21: 36743347-36743347 on Build GRCh38
rs111561480 Chr.21: 36743462-36743462 on Build GRCh38
rs756229321 Chr.21: 36743446-36743446 on Build GRCh38
rs767698678 Chr.21: 36743539-36743539 on Build GRCh38

View Full Product Details