Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58081502-58082026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNB
Entrez Gene ID
2317
Gene Name
filamin B
Gene Aliases
ABP-278, ABP-280, AOI, FH1, FLN-B, FLN1L, LRS1, SCT, TABP, TAP
UniGene
Hs.476448
Gene Chromosome Location
Chr.3: 58008400-58172255 on Build GRCh38
Transcripts
NM_001164317.1 NM_001164318.1 NM_001164319.1 NM_001457.3 XM_005264978.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5009952 c.731C>T p.P244L Chr.3: 58081720-58081720 on Build GRCh38
3916518 c.700C>T p.L234L Chr.3: 58081689-58081689 on Build GRCh38
3596735 c.768A>G p.K256K Chr.3: 58081757-58081757 on Build GRCh38
3596734 c.768A>G p.K256K Chr.3: 58081757-58081757 on Build GRCh38
172580 c.655G>T p.E219* Chr.3: 58081644-58081644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs80356508 Chr.3: 58081668-58081668 on Build GRCh38
rs371715057 Chr.3: 58081751-58081751 on Build GRCh38
rs375494750 Chr.3: 58081854-58081854 on Build GRCh38
rs953126780 Chr.3: 58081552-58081552 on Build GRCh38
rs761419793 Chr.3: 58081609-58081611 on Build GRCh38
rs776986249 Chr.3: 58081691-58081691 on Build GRCh38
rs1009155747 Chr.3: 58081895-58081895 on Build GRCh38
rs374601070 Chr.3: 58081577-58081577 on Build GRCh38
rs953178553 Chr.3: 58081866-58081866 on Build GRCh38
rs576985217 Chr.3: 58081727-58081727 on Build GRCh38
rs376619286 Chr.3: 58081765-58081765 on Build GRCh38
rs111433950 Chr.3: 58081709-58081709 on Build GRCh38
rs986194672 Chr.3: 58081776-58081776 on Build GRCh38
rs199540800 Chr.3: 58081802-58081802 on Build GRCh38
rs763710544 Chr.3: 58081719-58081719 on Build GRCh38
rs369055996 Chr.3: 58081798-58081798 on Build GRCh38
rs140815373 Chr.3: 58081658-58081658 on Build GRCh38
rs771976937 Chr.3: 58081784-58081784 on Build GRCh38
rs879475138 Chr.3: 58081918-58081918 on Build GRCh38
rs574643677 Chr.3: 58081657-58081657 on Build GRCh38
rs367648404 Chr.3: 58081632-58081632 on Build GRCh38
rs142650256 Chr.3: 58081576-58081576 on Build GRCh38
rs1000134283 Chr.3: 58081667-58081667 on Build GRCh38
rs755253275 Chr.3: 58081636-58081636 on Build GRCh38
rs1045781125 Chr.3: 58081788-58081788 on Build GRCh38
rs772165318 Chr.3: 58081672-58081672 on Build GRCh38
rs781656496 Chr.3: 58081803-58081803 on Build GRCh38
rs771387705 Chr.3: 58081688-58081688 on Build GRCh38
rs770162436 Chr.3: 58081816-58081816 on Build GRCh38
rs767031309 Chr.3: 58081721-58081721 on Build GRCh38
rs899075127 Chr.3: 58081838-58081838 on Build GRCh38
rs753697239 Chr.3: 58081745-58081745 on Build GRCh38
rs143743264 Chr.3: 58081676-58081676 on Build GRCh38
rs1018671479 Chr.3: 58081912-58081912 on Build GRCh38
rs375645067 Chr.3: 58081579-58081579 on Build GRCh38
rs140037128 Chr.3: 58081720-58081720 on Build GRCh38
rs151014421 Chr.3: 58081656-58081656 on Build GRCh38
rs377057780 Chr.3: 58081810-58081810 on Build GRCh38
rs778126745 Chr.3: 58081846-58081846 on Build GRCh38
rs1020524949 Chr.3: 58081624-58081624 on Build GRCh38
rs756148157 Chr.3: 58081743-58081743 on Build GRCh38
rs569503999 Chr.3: 58081758-58081758 on Build GRCh38
rs764825815 Chr.3: 58081649-58081655 on Build GRCh38
rs1055964862 Chr.3: 58081741-58081741 on Build GRCh38
rs972432246 Chr.3: 58081941-58081941 on Build GRCh38
rs749900657 Chr.3: 58081732-58081732 on Build GRCh38
rs984017977 Chr.3: 58081697-58081697 on Build GRCh38
rs752945898 Chr.3: 58081768-58081768 on Build GRCh38
rs780012995 Chr.3: 58081612-58081612 on Build GRCh38
rs747025323 Chr.3: 58081795-58081795 on Build GRCh38
rs1038800863 Chr.3: 58081814-58081814 on Build GRCh38
rs1040659635 Chr.3: 58081787-58081812 on Build GRCh38
rs750342917 Chr.3: 58081588-58081588 on Build GRCh38
rs758503616 Chr.3: 58081782-58081782 on Build GRCh38
rs749534602 Chr.3: 58081673-58081673 on Build GRCh38
rs121908896 Chr.3: 58081692-58081692 on Build GRCh38
rs967554798 Chr.3: 58081965-58081965 on Build GRCh38
rs937748371 Chr.3: 58081561-58081561 on Build GRCh38
rs977099823 Chr.3: 58081990-58081990 on Build GRCh38
rs919159429 Chr.3: 58081993-58081993 on Build GRCh38
rs756022296 Chr.3: 58081605-58081605 on Build GRCh38
rs80356507 Chr.3: 58081689-58081689 on Build GRCh38
rs373034402 Chr.3: 58081850-58081850 on Build GRCh38

View Full Product Details