Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 64539152-64539652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS9
Entrez Gene ID
56999
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 9
Gene Aliases
-
UniGene
Hs.656071
Gene Chromosome Location
Chr.3: 64515654-64688000 on Build GRCh38
Transcripts
NM_001318781.1 NM_182920.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1048043 c.5548A>G p.S1850G Chr.3: 64539268-64539268 on Build GRCh38
6295005 c.5601C>A p.A1867A Chr.3: 64539215-64539215 on Build GRCh38
4500119 c.5580C>T p.A1860A Chr.3: 64539236-64539236 on Build GRCh38
6999709 c.5596G>A p.A1866T Chr.3: 64539220-64539220 on Build GRCh38
5569353 c.5577A>G p.T1859T Chr.3: 64539239-64539239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773505105 Chr.3: 64539175-64539175 on Build GRCh38
rs981792208 Chr.3: 64539397-64539397 on Build GRCh38
rs754393700 Chr.3: 64539219-64539219 on Build GRCh38
rs369176747 Chr.3: 64539260-64539260 on Build GRCh38
rs777962741 Chr.3: 64539312-64539312 on Build GRCh38
rs953999534 Chr.3: 64539517-64539517 on Build GRCh38
rs945079921 Chr.3: 64539387-64539387 on Build GRCh38
rs903166615 Chr.3: 64539255-64539255 on Build GRCh38
rs35016041 Chr.3: 64539555-64539555 on Build GRCh38
rs1008183281 Chr.3: 64539313-64539313 on Build GRCh38
rs145825100 Chr.3: 64539630-64539630 on Build GRCh38
rs773904324 Chr.3: 64539195-64539195 on Build GRCh38
rs913527021 Chr.3: 64539596-64539596 on Build GRCh38
rs1039233129 Chr.3: 64539505-64539505 on Build GRCh38
rs775740037 Chr.3: 64539258-64539258 on Build GRCh38
rs769458833 Chr.3: 64539242-64539242 on Build GRCh38
rs148016958 Chr.3: 64539221-64539221 on Build GRCh38
rs187760371 Chr.3: 64539206-64539206 on Build GRCh38
rs903639102 Chr.3: 64539447-64539447 on Build GRCh38
rs748187996 Chr.3: 64539224-64539224 on Build GRCh38
rs781418799 Chr.3: 64539339-64539339 on Build GRCh38
rs755581538 Chr.3: 64539220-64539220 on Build GRCh38
rs75431606 Chr.3: 64539470-64539470 on Build GRCh38
rs201534044 Chr.3: 64539191-64539191 on Build GRCh38
rs74412571 Chr.3: 64539508-64539508 on Build GRCh38
rs753248523 Chr.3: 64539298-64539298 on Build GRCh38
rs890707659 Chr.3: 64539293-64539293 on Build GRCh38
rs761165505 Chr.3: 64539263-64539263 on Build GRCh38
rs141665428 Chr.3: 64539254-64539254 on Build GRCh38
rs546873377 Chr.3: 64539236-64539236 on Build GRCh38
rs538643980 Chr.3: 64539625-64539625 on Build GRCh38
rs760443576 Chr.3: 64539273-64539273 on Build GRCh38
rs768535965 Chr.3: 64539257-64539257 on Build GRCh38
rs113145901 Chr.3: 64539564-64539564 on Build GRCh38
rs192728999 Chr.3: 64539578-64539578 on Build GRCh38
rs759998812 Chr.3: 64539281-64539281 on Build GRCh38
rs11130969 Chr.3: 64539501-64539501 on Build GRCh38
rs994560899 Chr.3: 64539371-64539371 on Build GRCh38
rs751722484 Chr.3: 64539324-64539324 on Build GRCh38
rs750257065 Chr.3: 64539395-64539397 on Build GRCh38
rs959277638 Chr.3: 64539418-64539427 on Build GRCh38
rs771094186 Chr.3: 64539265-64539265 on Build GRCh38
rs757552705 Chr.3: 64539336-64539336 on Build GRCh38
rs766825030 Chr.3: 64539216-64539216 on Build GRCh38
rs777787758 Chr.3: 64539234-64539234 on Build GRCh38
rs896322393 Chr.3: 64539475-64539475 on Build GRCh38
rs766820473 Chr.3: 64539276-64539276 on Build GRCh38
rs758536387 Chr.3: 64539232-64539232 on Build GRCh38
rs775344841 Chr.3: 64539190-64539190 on Build GRCh38
rs775326790 Chr.3: 64539253-64539253 on Build GRCh38
rs144202116 Chr.3: 64539462-64539462 on Build GRCh38
rs1043354123 Chr.3: 64539402-64539402 on Build GRCh38
rs989378489 Chr.3: 64539565-64539565 on Build GRCh38
rs146166559 Chr.3: 64539283-64539283 on Build GRCh38
rs924688007 Chr.3: 64539483-64539483 on Build GRCh38
rs1035388018 Chr.3: 64539410-64539410 on Build GRCh38
rs568648768 Chr.3: 64539482-64539482 on Build GRCh38
rs140082715 Chr.3: 64539527-64539527 on Build GRCh38
rs549271779 Chr.3: 64539349-64539349 on Build GRCh38
rs11130968 Chr.3: 64539495-64539495 on Build GRCh38
rs756378012 Chr.3: 64539213-64539213 on Build GRCh38
rs143072536 Chr.3: 64539266-64539266 on Build GRCh38
rs372261210 Chr.3: 64539311-64539311 on Build GRCh38
rs774306492 Chr.3: 64539259-64539259 on Build GRCh38
rs978840586 Chr.3: 64539468-64539468 on Build GRCh38
rs1026004862 Chr.3: 64539383-64539383 on Build GRCh38
rs985580594 Chr.3: 64539524-64539524 on Build GRCh38
rs1051770575 Chr.3: 64539492-64539492 on Build GRCh38
rs745606413 Chr.3: 64539180-64539180 on Build GRCh38
rs571347574 Chr.3: 64539619-64539619 on Build GRCh38
rs533506204 Chr.3: 64539509-64539509 on Build GRCh38
rs761410587 Chr.3: 64539198-64539198 on Build GRCh38
rs1020080281 Chr.3: 64539541-64539541 on Build GRCh38
rs991066722 Chr.3: 64539439-64539439 on Build GRCh38
rs937786094 Chr.3: 64539457-64539457 on Build GRCh38
rs934694207 Chr.3: 64539488-64539488 on Build GRCh38
rs767040189 Chr.3: 64539199-64539199 on Build GRCh38
rs764770242 Chr.3: 64539328-64539329 on Build GRCh38
rs781457154 Chr.3: 64539373-64539373 on Build GRCh38
rs950391558 Chr.3: 64539385-64539387 on Build GRCh38
rs947624883 Chr.3: 64539626-64539626 on Build GRCh38
rs769633848 Chr.3: 64539184-64539184 on Build GRCh38
rs1013802304 Chr.3: 64539479-64539479 on Build GRCh38
rs375448415 Chr.3: 64539545-64539545 on Build GRCh38

View Full Product Details