Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119730082-119730600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAATS1
Entrez Gene ID
89876
Gene Name
MYCBP associated and testis expressed 1
Gene Aliases
AAT1, AAT1alpha, C3orf15, CaM-IP2, SPATA26
UniGene
Hs.341906
Gene Chromosome Location
Chr.3: 119703022-119767102 on Build GRCh38
Transcripts
NM_001320316.1 NM_001320317.1 NM_001320318.1 NM_033364.3 XM_011513277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667671 c.950C>T p.A317V Chr.3: 119730309-119730309 on Build GRCh38
7161821 c.921G>A p.Q307Q Chr.3: 119730280-119730280 on Build GRCh38
1418048 c.984delA p.M330fs*16 Chr.3: 119730343-119730343 on Build GRCh38
3120163 c.907C>T p.R303C Chr.3: 119730266-119730266 on Build GRCh38
6163069 c.952C>G p.R318G Chr.3: 119730311-119730311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766008419 Chr.3: 119730239-119730239 on Build GRCh38
rs79707393 Chr.3: 119730453-119730453 on Build GRCh38
rs1046711388 Chr.3: 119730320-119730320 on Build GRCh38
rs984109906 Chr.3: 119730163-119730163 on Build GRCh38
rs959523740 Chr.3: 119730399-119730399 on Build GRCh38
rs771411833 Chr.3: 119730405-119730405 on Build GRCh38
rs762388235 Chr.3: 119730351-119730351 on Build GRCh38
rs766835433 Chr.3: 119730132-119730138 on Build GRCh38
rs9819218 Chr.3: 119730318-119730318 on Build GRCh38
rs780183569 Chr.3: 119730277-119730277 on Build GRCh38
rs776455619 Chr.3: 119730246-119730246 on Build GRCh38
rs543920270 Chr.3: 119730156-119730156 on Build GRCh38
rs755231831 Chr.3: 119730389-119730389 on Build GRCh38
rs934195549 Chr.3: 119730237-119730237 on Build GRCh38
rs758439972 Chr.3: 119730394-119730394 on Build GRCh38
rs61742290 Chr.3: 119730312-119730312 on Build GRCh38
rs540120371 Chr.3: 119730434-119730434 on Build GRCh38
rs762434808 Chr.3: 119730382-119730382 on Build GRCh38
rs144639559 Chr.3: 119730323-119730323 on Build GRCh38
rs201238294 Chr.3: 119730234-119730234 on Build GRCh38
rs191001867 Chr.3: 119730366-119730366 on Build GRCh38
rs761545403 Chr.3: 119730238-119730238 on Build GRCh38
rs755035891 Chr.3: 119730293-119730293 on Build GRCh38
rs563121539 Chr.3: 119730294-119730294 on Build GRCh38
rs758073767 Chr.3: 119730170-119730170 on Build GRCh38
rs764857021 Chr.3: 119730248-119730248 on Build GRCh38
rs749524313 Chr.3: 119730291-119730291 on Build GRCh38
rs780274317 Chr.3: 119730196-119730196 on Build GRCh38
rs912739624 Chr.3: 119730491-119730491 on Build GRCh38
rs756404380 Chr.3: 119730194-119730194 on Build GRCh38
rs139356773 Chr.3: 119730185-119730185 on Build GRCh38
rs771432001 Chr.3: 119730314-119730314 on Build GRCh38
rs148476947 Chr.3: 119730362-119730362 on Build GRCh38
rs566994345 Chr.3: 119730343-119730343 on Build GRCh38
rs747488291 Chr.3: 119730400-119730400 on Build GRCh38
rs772780380 Chr.3: 119730379-119730380 on Build GRCh38
rs778275774 Chr.3: 119730398-119730398 on Build GRCh38
rs746043398 Chr.3: 119730189-119730189 on Build GRCh38
rs956971945 Chr.3: 119730460-119730460 on Build GRCh38
rs757029797 Chr.3: 119730273-119730273 on Build GRCh38
rs146078151 Chr.3: 119730267-119730267 on Build GRCh38
rs748746264 Chr.3: 119730229-119730229 on Build GRCh38
rs759052987 Chr.3: 119730247-119730247 on Build GRCh38
rs570280663 Chr.3: 119730504-119730504 on Build GRCh38
rs145277027 Chr.3: 119730363-119730363 on Build GRCh38
rs759515471 Chr.3: 119730397-119730397 on Build GRCh38
rs765897772 Chr.3: 119730383-119730386 on Build GRCh38
rs972686553 Chr.3: 119730514-119730514 on Build GRCh38
rs764049216 Chr.3: 119730358-119730358 on Build GRCh38
rs267599558 Chr.3: 119730333-119730333 on Build GRCh38
rs752774281 Chr.3: 119730390-119730390 on Build GRCh38
rs745490770 Chr.3: 119730406-119730406 on Build GRCh38
rs563743919 Chr.3: 119730573-119730573 on Build GRCh38
rs959705771 Chr.3: 119730109-119730109 on Build GRCh38
rs776943512 Chr.3: 119730216-119730216 on Build GRCh38
rs142876723 Chr.3: 119730233-119730233 on Build GRCh38
rs1001401541 Chr.3: 119730370-119730370 on Build GRCh38
rs1002765738 Chr.3: 119730340-119730348 on Build GRCh38
rs574107929 Chr.3: 119730213-119730213 on Build GRCh38
rs942825636 Chr.3: 119730545-119730545 on Build GRCh38
rs544044407 Chr.3: 119730508-119730508 on Build GRCh38
rs751594237 Chr.3: 119730392-119730393 on Build GRCh38
rs891461461 Chr.3: 119730347-119730347 on Build GRCh38
rs533246019 Chr.3: 119730261-119730261 on Build GRCh38
rs374317339 Chr.3: 119730311-119730311 on Build GRCh38
rs111294292 Chr.3: 119730367-119730367 on Build GRCh38
rs774780509 Chr.3: 119730217-119730217 on Build GRCh38
rs758399473 Chr.3: 119730266-119730266 on Build GRCh38
rs371576235 Chr.3: 119730419-119730419 on Build GRCh38
rs759220288 Chr.3: 119730188-119730188 on Build GRCh38
rs961363150 Chr.3: 119730487-119730487 on Build GRCh38
rs566283831 Chr.3: 119730412-119730412 on Build GRCh38
rs769268601 Chr.3: 119730308-119730311 on Build GRCh38

View Full Product Details