Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 40812454-40812959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBB2
Entrez Gene ID
323
Gene Name
amyloid beta precursor protein binding family B member 2
Gene Aliases
FE65L, FE65L1
UniGene
Hs.479602
Gene Chromosome Location
Chr.4: 40810027-41216714 on Build GRCh38
Transcripts
NM_001166050.1 NM_001166051.1 NM_001166052.1 NM_001166053.1 NM_001166054.1 NM_004307.1 NM_173075.4 XM_005248101.3 XM_006714005.3 XM_006714007.3 XM_006714008.3 XM_006714009.3 XM_006714010.3 XM_006714011.3 XM_006714012.3 XM_011513686.2 XM_011513687.2 XM_017008142.1 XM_017008143.1 XM_017008144.1 XM_017008145.1 XM_017008146.1 XM_017008147.1 XM_017008148.1 XM_017008149.1 XM_017008150.1 XM_017008151.1 XM_017008152.1 XM_017008153.1 XM_017008154.1 XM_017008155.1 XM_017008156.1 XM_017008157.1 XM_017008158.1

SNP Detail

SNP ID Chromosome Location
rs561355532 Chr.4: 40812495-40812495 on Build GRCh38
rs1015131444 Chr.4: 40812633-40812633 on Build GRCh38
rs990725537 Chr.4: 40812685-40812685 on Build GRCh38
rs779668571 Chr.4: 40812490-40812490 on Build GRCh38
rs1053380385 Chr.4: 40812560-40812560 on Build GRCh38
rs10537665 Chr.4: 40812625-40812627 on Build GRCh38
rs551599509 Chr.4: 40812787-40812787 on Build GRCh38
rs528847829 Chr.4: 40812606-40812606 on Build GRCh38
rs559161456 Chr.4: 40812711-40812711 on Build GRCh38
rs141944589 Chr.4: 40812712-40812712 on Build GRCh38
rs376507857 Chr.4: 40812915-40812915 on Build GRCh38
rs145809150 Chr.4: 40812735-40812735 on Build GRCh38
rs1051132012 Chr.4: 40812589-40812589 on Build GRCh38
rs1041925084 Chr.4: 40812793-40812793 on Build GRCh38
rs1008889025 Chr.4: 40812489-40812489 on Build GRCh38
rs190415143 Chr.4: 40812790-40812790 on Build GRCh38
rs890604255 Chr.4: 40812604-40812604 on Build GRCh38
rs768652067 Chr.4: 40812555-40812555 on Build GRCh38
rs1027619405 Chr.4: 40812665-40812665 on Build GRCh38
rs796749215 Chr.4: 40812612-40812614 on Build GRCh38
rs201193426 Chr.4: 40812615-40812617 on Build GRCh38
rs989792248 Chr.4: 40812747-40812747 on Build GRCh38
rs923678364 Chr.4: 40812542-40812542 on Build GRCh38
rs1023994415 Chr.4: 40812688-40812688 on Build GRCh38
rs893462792 Chr.4: 40812561-40812561 on Build GRCh38
rs7665366 Chr.4: 40812672-40812672 on Build GRCh38
rs923813543 Chr.4: 40812767-40812767 on Build GRCh38
rs761399973 Chr.4: 40812770-40812770 on Build GRCh38
rs150066610 Chr.4: 40812583-40812583 on Build GRCh38
rs958968581 Chr.4: 40812680-40812680 on Build GRCh38
rs891287614 Chr.4: 40812865-40812865 on Build GRCh38
rs397783017 Chr.4: 40812628-40812630 on Build GRCh38
rs1046109285 Chr.4: 40812538-40812538 on Build GRCh38
rs542864172 Chr.4: 40812477-40812477 on Build GRCh38
rs970645557 Chr.4: 40812703-40812703 on Build GRCh38
rs914884176 Chr.4: 40812886-40812886 on Build GRCh38
rs866141011 Chr.4: 40812742-40812742 on Build GRCh38
rs1039943971 Chr.4: 40812922-40812922 on Build GRCh38
rs145422241 Chr.4: 40812658-40812658 on Build GRCh38
rs373305052 Chr.4: 40812628-40812628 on Build GRCh38
rs185650444 Chr.4: 40812514-40812514 on Build GRCh38
rs7671375 Chr.4: 40812813-40812813 on Build GRCh38
rs1015163715 Chr.4: 40812614-40812614 on Build GRCh38
rs995436997 Chr.4: 40812562-40812563 on Build GRCh38
rs547087355 Chr.4: 40812858-40812858 on Build GRCh38
rs776658520 Chr.4: 40812842-40812843 on Build GRCh38
rs147796690 Chr.4: 40812748-40812748 on Build GRCh38
rs139391619 Chr.4: 40812522-40812522 on Build GRCh38
rs897904424 Chr.4: 40812653-40812653 on Build GRCh38
rs952658199 Chr.4: 40812486-40812486 on Build GRCh38
rs180987723 Chr.4: 40812503-40812503 on Build GRCh38

View Full Product Details