Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 52800993-52801502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA1
Entrez Gene ID
3672
Gene Name
integrin subunit alpha 1
Gene Aliases
CD49a, VLA1
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52788302-52953655 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Gene Symbol
PELO
Entrez Gene ID
53918
Gene Name
pelota homolog (Drosophila)
Gene Aliases
CGI-17, PRO1770
UniGene
Hs.644352
Gene Chromosome Location
Chr.5: 52787940-52802617 on Build GRCh38
Transcripts
NM_181501.1 NM_015946.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6868346 c.696G>A p.L232L Chr.5: 52801090-52801090 on Build GRCh38
7146488 c.639G>A p.V213V Chr.5: 52801033-52801033 on Build GRCh38
1132152 c.622G>T p.A208S Chr.5: 52801016-52801016 on Build GRCh38
3920156 c.606A>T p.V202V Chr.5: 52801000-52801000 on Build GRCh38
6865146 c.644A>C p.E215A Chr.5: 52801038-52801038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112612194 Chr.5: 52801094-52801094 on Build GRCh38
rs371879238 Chr.5: 52801377-52801377 on Build GRCh38
rs769701825 Chr.5: 52801157-52801157 on Build GRCh38
rs113870011 Chr.5: 52801321-52801321 on Build GRCh38
rs139977042 Chr.5: 52801050-52801050 on Build GRCh38
rs752511091 Chr.5: 52801049-52801049 on Build GRCh38
rs746484504 Chr.5: 52801191-52801191 on Build GRCh38
rs772915083 Chr.5: 52801428-52801429 on Build GRCh38
rs780875619 Chr.5: 52801453-52801453 on Build GRCh38
rs927508067 Chr.5: 52801449-52801449 on Build GRCh38
rs375508745 Chr.5: 52801368-52801368 on Build GRCh38
rs749226074 Chr.5: 52801473-52801473 on Build GRCh38
rs145721878 Chr.5: 52801420-52801420 on Build GRCh38
rs369963149 Chr.5: 52801408-52801408 on Build GRCh38
rs775396002 Chr.5: 52801464-52801464 on Build GRCh38
rs181398202 Chr.5: 52801128-52801128 on Build GRCh38
rs999826440 Chr.5: 52801219-52801219 on Build GRCh38
rs747792288 Chr.5: 52801021-52801022 on Build GRCh38
rs780755447 Chr.5: 52801105-52801105 on Build GRCh38
rs756301643 Chr.5: 52801415-52801415 on Build GRCh38
rs937616881 Chr.5: 52801096-52801096 on Build GRCh38
rs764192082 Chr.5: 52801052-52801052 on Build GRCh38
rs2059202 Chr.5: 52801349-52801349 on Build GRCh38
rs770280664 Chr.5: 52801129-52801129 on Build GRCh38
rs750916684 Chr.5: 52801093-52801093 on Build GRCh38
rs779341584 Chr.5: 52801441-52801441 on Build GRCh38
rs749638968 Chr.5: 52801106-52801106 on Build GRCh38
rs186460209 Chr.5: 52801311-52801311 on Build GRCh38
rs769363762 Chr.5: 52801461-52801461 on Build GRCh38
rs769466197 Chr.5: 52801168-52801185 on Build GRCh38
rs964981464 Chr.5: 52801318-52801318 on Build GRCh38
rs746603199 Chr.5: 52801448-52801448 on Build GRCh38
rs1010256338 Chr.5: 52801187-52801187 on Build GRCh38
rs764010359 Chr.5: 52801367-52801367 on Build GRCh38
rs1054158932 Chr.5: 52801255-52801255 on Build GRCh38
rs891212006 Chr.5: 52801200-52801200 on Build GRCh38
rs554896969 Chr.5: 52801083-52801083 on Build GRCh38
rs199703397 Chr.5: 52801371-52801371 on Build GRCh38
rs112573303 Chr.5: 52801277-52801277 on Build GRCh38
rs775449436 Chr.5: 52801359-52801359 on Build GRCh38
rs372365242 Chr.5: 52801143-52801143 on Build GRCh38
rs776236761 Chr.5: 52801134-52801134 on Build GRCh38
rs983095941 Chr.5: 52801376-52801376 on Build GRCh38
rs755238457 Chr.5: 52801440-52801440 on Build GRCh38
rs756957091 Chr.5: 52801450-52801450 on Build GRCh38
rs745354927 Chr.5: 52801457-52801457 on Build GRCh38
rs369308865 Chr.5: 52801393-52801393 on Build GRCh38
rs1025645893 Chr.5: 52801388-52801388 on Build GRCh38
rs757223920 Chr.5: 52801081-52801081 on Build GRCh38
rs770413741 Chr.5: 52801232-52801232 on Build GRCh38
rs145206065 Chr.5: 52801190-52801190 on Build GRCh38
rs990374199 Chr.5: 52801451-52801451 on Build GRCh38
rs75328051 Chr.5: 52801110-52801110 on Build GRCh38
rs1499280 Chr.5: 52801055-52801055 on Build GRCh38
rs202151315 Chr.5: 52801425-52801425 on Build GRCh38
rs765143574 Chr.5: 52801463-52801463 on Build GRCh38
rs762735875 Chr.5: 52801362-52801362 on Build GRCh38
rs375504463 Chr.5: 52801380-52801380 on Build GRCh38
rs777276798 Chr.5: 52801114-52801114 on Build GRCh38

View Full Product Details