Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132483376-132483874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF1
Entrez Gene ID
3659
Gene Name
interferon regulatory factor 1
Gene Aliases
IRF-1, MAR
UniGene
Hs.436061
Gene Chromosome Location
Chr.5: 132481609-132490789 on Build GRCh38
Transcripts
NM_002198.2 XM_011543378.2 XM_011543379.2

SNP Detail

SNP ID Chromosome Location
rs969564021 Chr.5: 132483485-132483485 on Build GRCh38
rs1036312271 Chr.5: 132483441-132483441 on Build GRCh38
rs35642338 Chr.5: 132483505-132483505 on Build GRCh38
rs940040669 Chr.5: 132483661-132483661 on Build GRCh38
rs17848427 Chr.5: 132483809-132483809 on Build GRCh38
rs1023440444 Chr.5: 132483481-132483481 on Build GRCh38
rs35178118 Chr.5: 132483439-132483439 on Build GRCh38
rs1048474095 Chr.5: 132483610-132483610 on Build GRCh38
rs950801236 Chr.5: 132483812-132483812 on Build GRCh38
rs146531199 Chr.5: 132483497-132483497 on Build GRCh38
rs754578243 Chr.5: 132483790-132483790 on Build GRCh38
rs549029062 Chr.5: 132483699-132483699 on Build GRCh38
rs904993156 Chr.5: 132483817-132483817 on Build GRCh38
rs1017677417 Chr.5: 132483446-132483446 on Build GRCh38
rs749126418 Chr.5: 132483614-132483614 on Build GRCh38
rs987764231 Chr.5: 132483524-132483524 on Build GRCh38
rs34850771 Chr.5: 132483523-132483523 on Build GRCh38
rs146224230 Chr.5: 132483504-132483504 on Build GRCh38
rs17848431 Chr.5: 132483442-132483442 on Build GRCh38
rs565931795 Chr.5: 132483727-132483727 on Build GRCh38
rs839 Chr.5: 132483434-132483434 on Build GRCh38
rs1051490325 Chr.5: 132483682-132483682 on Build GRCh38
rs902448715 Chr.5: 132483410-132483410 on Build GRCh38
rs867771153 Chr.5: 132483640-132483640 on Build GRCh38
rs570902330 Chr.5: 132483822-132483822 on Build GRCh38
rs190321767 Chr.5: 132483800-132483800 on Build GRCh38
rs1043631618 Chr.5: 132483400-132483400 on Build GRCh38
rs999736369 Chr.5: 132483433-132483433 on Build GRCh38
rs935242397 Chr.5: 132483677-132483677 on Build GRCh38
rs17848429 Chr.5: 132483507-132483507 on Build GRCh38
rs1027840973 Chr.5: 132483501-132483501 on Build GRCh38
rs908617780 Chr.5: 132483631-132483631 on Build GRCh38
rs923628861 Chr.5: 132483667-132483667 on Build GRCh38
rs890953848 Chr.5: 132483717-132483717 on Build GRCh38
rs533948726 Chr.5: 132483835-132483835 on Build GRCh38
rs529034915 Chr.5: 132483658-132483658 on Build GRCh38
rs747840820 Chr.5: 132483810-132483810 on Build GRCh38
rs550287123 Chr.5: 132483539-132483539 on Build GRCh38
rs1007143644 Chr.5: 132483729-132483729 on Build GRCh38
rs17848428 Chr.5: 132483570-132483570 on Build GRCh38
rs921418414 Chr.5: 132483576-132483576 on Build GRCh38
rs931391299 Chr.5: 132483604-132483604 on Build GRCh38
rs975999136 Chr.5: 132483575-132483575 on Build GRCh38
rs1013858459 Chr.5: 132483467-132483467 on Build GRCh38

View Full Product Details