Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 7289443-7289946 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSR1
Entrez Gene ID
6745
Gene Name
signal sequence receptor subunit 1
Gene Aliases
TRAPA
UniGene
Hs.114033
Gene Chromosome Location
Chr.6: 7281143-7313314 on Build GRCh38
Transcripts
NM_001292008.1 NM_003144.4 NR_120448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6790824 c.826C>T p.R276W Chr.6: 7289899-7289899 on Build GRCh38
238245 c.843A>G p.S281S Chr.6: 7289882-7289882 on Build GRCh38
1081382 c.794-6T>C p.? Chr.6: 7289937-7289937 on Build GRCh38
285376 c.803C>T p.S268L Chr.6: 7289922-7289922 on Build GRCh38
4625840 c.821G>T p.R274M Chr.6: 7289904-7289904 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs897140581 Chr.6: 7289589-7289589 on Build GRCh38
rs531982208 Chr.6: 7289672-7289672 on Build GRCh38
rs925957114 Chr.6: 7289701-7289701 on Build GRCh38
rs995622475 Chr.6: 7289590-7289590 on Build GRCh38
rs138203075 Chr.6: 7289894-7289894 on Build GRCh38
rs995507917 Chr.6: 7289867-7289867 on Build GRCh38
rs140935765 Chr.6: 7289644-7289644 on Build GRCh38
rs200254862 Chr.6: 7289729-7289729 on Build GRCh38
rs537748481 Chr.6: 7289898-7289898 on Build GRCh38
rs960832832 Chr.6: 7289661-7289661 on Build GRCh38
rs554248180 Chr.6: 7289774-7289774 on Build GRCh38
rs771023317 Chr.6: 7289756-7289756 on Build GRCh38
rs5874088 Chr.6: 7289515-7289516 on Build GRCh38
rs756020243 Chr.6: 7289876-7289876 on Build GRCh38
rs397841485 Chr.6: 7289727-7289728 on Build GRCh38
rs989065792 Chr.6: 7289639-7289639 on Build GRCh38
rs190008597 Chr.6: 7289781-7289781 on Build GRCh38
rs778502527 Chr.6: 7289899-7289899 on Build GRCh38
rs1049568540 Chr.6: 7289697-7289697 on Build GRCh38
rs368803812 Chr.6: 7289848-7289848 on Build GRCh38
rs200762552 Chr.6: 7289730-7289730 on Build GRCh38
rs932376675 Chr.6: 7289694-7289694 on Build GRCh38
rs979028200 Chr.6: 7289688-7289688 on Build GRCh38
rs774238351 Chr.6: 7289896-7289896 on Build GRCh38
rs201556739 Chr.6: 7289842-7289842 on Build GRCh38
rs547495609 Chr.6: 7289777-7289777 on Build GRCh38
rs1034778699 Chr.6: 7289645-7289645 on Build GRCh38
rs988069241 Chr.6: 7289661-7289661 on Build GRCh38
rs761190531 Chr.6: 7289863-7289863 on Build GRCh38
rs766483867 Chr.6: 7289841-7289841 on Build GRCh38
rs763971926 Chr.6: 7289883-7289883 on Build GRCh38
rs891052762 Chr.6: 7289516-7289516 on Build GRCh38
rs375330919 Chr.6: 7289823-7289823 on Build GRCh38
rs912381267 Chr.6: 7289737-7289737 on Build GRCh38
rs960586363 Chr.6: 7289636-7289636 on Build GRCh38
rs1050681 Chr.6: 7289549-7289549 on Build GRCh38
rs374110199 Chr.6: 7289646-7289650 on Build GRCh38
rs763079454 Chr.6: 7289826-7289826 on Build GRCh38
rs544500015 Chr.6: 7289645-7289649 on Build GRCh38
rs550168036 Chr.6: 7289498-7289498 on Build GRCh38
rs527714242 Chr.6: 7289644-7289645 on Build GRCh38
rs16740 Chr.6: 7289725-7289726 on Build GRCh38
rs890636907 Chr.6: 7289512-7289513 on Build GRCh38
rs564907493 Chr.6: 7289670-7289670 on Build GRCh38
rs943979568 Chr.6: 7289760-7289760 on Build GRCh38
rs745314343 Chr.6: 7289918-7289918 on Build GRCh38
rs770114375 Chr.6: 7289817-7289817 on Build GRCh38
rs76573543 Chr.6: 7289515-7289515 on Build GRCh38
rs932031554 Chr.6: 7289723-7289723 on Build GRCh38
rs144671908 Chr.6: 7289810-7289810 on Build GRCh38
rs531501817 Chr.6: 7289635-7289635 on Build GRCh38
rs773502503 Chr.6: 7289822-7289822 on Build GRCh38
rs746784549 Chr.6: 7289594-7289594 on Build GRCh38
rs201797960 Chr.6: 7289727-7289727 on Build GRCh38
rs530355981 Chr.6: 7289803-7289803 on Build GRCh38
rs3029819 Chr.6: 7289726-7289727 on Build GRCh38
rs925044599 Chr.6: 7289690-7289690 on Build GRCh38
rs970091424 Chr.6: 7289597-7289597 on Build GRCh38
rs146482489 Chr.6: 7289585-7289585 on Build GRCh38
rs1027066660 Chr.6: 7289595-7289595 on Build GRCh38
rs994948631 Chr.6: 7289596-7289596 on Build GRCh38
rs1001710767 Chr.6: 7289612-7289612 on Build GRCh38

View Full Product Details