Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 18225553-18226074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEK
Entrez Gene ID
7913
Gene Name
DEK proto-oncogene
Gene Aliases
D6S231E
UniGene
Hs.484813
Gene Chromosome Location
Chr.6: 18224169-18264568 on Build GRCh38
Transcripts
NM_001134709.1 NM_003472.3 XM_017011273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5619509 c.1117-1G>A p.? Chr.6: 18225731-18225731 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767444504 Chr.6: 18225919-18225919 on Build GRCh38
rs748771823 Chr.6: 18225778-18225778 on Build GRCh38
rs7740259 Chr.6: 18225982-18225982 on Build GRCh38
rs970930974 Chr.6: 18225642-18225642 on Build GRCh38
rs746306851 Chr.6: 18225626-18225626 on Build GRCh38
rs777915374 Chr.6: 18225649-18225650 on Build GRCh38
rs776270651 Chr.6: 18225749-18225751 on Build GRCh38
rs371844223 Chr.6: 18225802-18225802 on Build GRCh38
rs1012803680 Chr.6: 18225589-18225589 on Build GRCh38
rs923833280 Chr.6: 18226005-18226005 on Build GRCh38
rs758760447 Chr.6: 18225753-18225753 on Build GRCh38
rs755041514 Chr.6: 18225746-18225746 on Build GRCh38
rs761525670 Chr.6: 18225703-18225703 on Build GRCh38
rs986400672 Chr.6: 18225596-18225596 on Build GRCh38
rs141592228 Chr.6: 18225913-18225913 on Build GRCh38
rs184447098 Chr.6: 18225744-18225744 on Build GRCh38
rs528315221 Chr.6: 18225948-18225948 on Build GRCh38
rs975388687 Chr.6: 18225658-18225658 on Build GRCh38
rs747567417 Chr.6: 18225762-18225762 on Build GRCh38
rs548358737 Chr.6: 18225606-18225606 on Build GRCh38
rs763353024 Chr.6: 18225800-18225800 on Build GRCh38
rs777082484 Chr.6: 18225776-18225776 on Build GRCh38
rs893861846 Chr.6: 18225827-18225827 on Build GRCh38
rs779022944 Chr.6: 18225751-18225751 on Build GRCh38
rs769355298 Chr.6: 18225677-18225677 on Build GRCh38
rs372245813 Chr.6: 18225694-18225694 on Build GRCh38
rs569862193 Chr.6: 18225717-18225718 on Build GRCh38
rs969051568 Chr.6: 18226017-18226017 on Build GRCh38
rs767276591 Chr.6: 18225714-18225714 on Build GRCh38
rs780078216 Chr.6: 18225681-18225681 on Build GRCh38
rs756105522 Chr.6: 18225716-18225716 on Build GRCh38
rs1045289009 Chr.6: 18225863-18225863 on Build GRCh38
rs532282018 Chr.6: 18225701-18225703 on Build GRCh38
rs117541327 Chr.6: 18225715-18225715 on Build GRCh38
rs906819549 Chr.6: 18225871-18225871 on Build GRCh38
rs761664467 Chr.6: 18225882-18225882 on Build GRCh38
rs533186606 Chr.6: 18225935-18225935 on Build GRCh38
rs935429218 Chr.6: 18225804-18225804 on Build GRCh38
rs951153547 Chr.6: 18225587-18225587 on Build GRCh38
rs935441112 Chr.6: 18225785-18225785 on Build GRCh38
rs1021517843 Chr.6: 18226008-18226008 on Build GRCh38
rs778100671 Chr.6: 18225755-18225755 on Build GRCh38
rs113873428 Chr.6: 18225781-18225781 on Build GRCh38
rs1023777580 Chr.6: 18225858-18225858 on Build GRCh38
rs935366754 Chr.6: 18226042-18226042 on Build GRCh38
rs576113187 Chr.6: 18225818-18225818 on Build GRCh38
rs770633210 Chr.6: 18225741-18225742 on Build GRCh38
rs896530714 Chr.6: 18225835-18225835 on Build GRCh38
rs947200565 Chr.6: 18225740-18225740 on Build GRCh38
rs770312836 Chr.6: 18225650-18225650 on Build GRCh38
rs983498893 Chr.6: 18225909-18225909 on Build GRCh38
rs893900246 Chr.6: 18226044-18226044 on Build GRCh38
rs768857901 Chr.6: 18225704-18225705 on Build GRCh38
rs950400469 Chr.6: 18225886-18225886 on Build GRCh38
rs779526332 Chr.6: 18225712-18225713 on Build GRCh38
rs1035347736 Chr.6: 18225671-18225671 on Build GRCh38
rs963711391 Chr.6: 18225981-18225981 on Build GRCh38
rs1023913391 Chr.6: 18225609-18225609 on Build GRCh38
rs763685943 Chr.6: 18225695-18225695 on Build GRCh38
rs780326393 Chr.6: 18225723-18225723 on Build GRCh38
rs771398092 Chr.6: 18225772-18225772 on Build GRCh38
rs977237519 Chr.6: 18226025-18226025 on Build GRCh38
rs921858437 Chr.6: 18225748-18225748 on Build GRCh38
rs759058993 Chr.6: 18225780-18225788 on Build GRCh38
rs1007970758 Chr.6: 18225926-18225926 on Build GRCh38
rs1054158690 Chr.6: 18226043-18226043 on Build GRCh38
rs964302591 Chr.6: 18225643-18225643 on Build GRCh38
rs148292160 Chr.6: 18225792-18225792 on Build GRCh38
rs981112274 Chr.6: 18225664-18225664 on Build GRCh38
rs1019262524 Chr.6: 18225639-18225639 on Build GRCh38
rs367707011 Chr.6: 18225797-18225797 on Build GRCh38
rs776805647 Chr.6: 18225661-18225661 on Build GRCh38
rs111981497 Chr.6: 18225698-18225698 on Build GRCh38
rs559181253 Chr.6: 18225918-18225918 on Build GRCh38
rs1025781688 Chr.6: 18225908-18225908 on Build GRCh38
rs186963513 Chr.6: 18225880-18225880 on Build GRCh38
rs751175790 Chr.6: 18225700-18225700 on Build GRCh38
rs1019231705 Chr.6: 18225932-18225932 on Build GRCh38
rs987333548 Chr.6: 18226003-18226005 on Build GRCh38
rs370564751 Chr.6: 18225846-18225846 on Build GRCh38
rs1056463228 Chr.6: 18225805-18225805 on Build GRCh38
rs573496050 Chr.6: 18225881-18225881 on Build GRCh38

View Full Product Details