Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26017646-26018142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H1A
Entrez Gene ID
3024
Gene Name
histone cluster 1, H1a
Gene Aliases
H1.1, H1A, H1F1, HIST1
UniGene
Hs.150206
Gene Chromosome Location
Chr.6: 26017032-26017812 on Build GRCh38
Transcripts
NM_005325.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1076670 c.38C>T p.A13V Chr.6: 26017695-26017695 on Build GRCh38
7217946 c.17C>T p.P6L Chr.6: 26017716-26017716 on Build GRCh38
3860876 c.36T>C p.S12S Chr.6: 26017697-26017697 on Build GRCh38
3860877 c.7G>A p.E3K Chr.6: 26017726-26017726 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs574576116 Chr.6: 26017760-26017760 on Build GRCh38
rs899662085 Chr.6: 26017821-26017821 on Build GRCh38
rs577194237 Chr.6: 26017787-26017787 on Build GRCh38
rs754555136 Chr.6: 26017734-26017734 on Build GRCh38
rs889812641 Chr.6: 26018100-26018100 on Build GRCh38
rs773916128 Chr.6: 26017685-26017685 on Build GRCh38
rs200599379 Chr.6: 26017714-26017714 on Build GRCh38
rs886954879 Chr.6: 26018083-26018083 on Build GRCh38
rs777641888 Chr.6: 26017713-26017713 on Build GRCh38
rs769056610 Chr.6: 26017781-26017781 on Build GRCh38
rs181032819 Chr.6: 26017927-26017927 on Build GRCh38
rs560125779 Chr.6: 26018043-26018043 on Build GRCh38
rs535595979 Chr.6: 26017755-26017755 on Build GRCh38
rs879135512 Chr.6: 26017875-26017875 on Build GRCh38
rs201252490 Chr.6: 26017698-26017698 on Build GRCh38
rs887342216 Chr.6: 26017938-26017950 on Build GRCh38
rs189137001 Chr.6: 26017769-26017769 on Build GRCh38
rs150685143 Chr.6: 26017709-26017709 on Build GRCh38
rs199506907 Chr.6: 26017672-26017672 on Build GRCh38
rs940060816 Chr.6: 26018091-26018091 on Build GRCh38
rs374714764 Chr.6: 26017757-26017757 on Build GRCh38
rs923698356 Chr.6: 26018039-26018039 on Build GRCh38
rs961559309 Chr.6: 26017788-26017788 on Build GRCh38
rs1024897717 Chr.6: 26017859-26017859 on Build GRCh38
rs967500990 Chr.6: 26017945-26017945 on Build GRCh38
rs759742491 Chr.6: 26017714-26017715 on Build GRCh38
rs779812387 Chr.6: 26017676-26017676 on Build GRCh38
rs770055445 Chr.6: 26017755-26017764 on Build GRCh38
rs902379794 Chr.6: 26017850-26017850 on Build GRCh38
rs775399089 Chr.6: 26017719-26017719 on Build GRCh38
rs753364452 Chr.6: 26017708-26017708 on Build GRCh38
rs776480210 Chr.6: 26017716-26017716 on Build GRCh38
rs762414961 Chr.6: 26017730-26017730 on Build GRCh38
rs765453206 Chr.6: 26017686-26017686 on Build GRCh38
rs199749 Chr.6: 26017857-26017857 on Build GRCh38
rs191114802 Chr.6: 26018017-26018017 on Build GRCh38
rs367757993 Chr.6: 26017761-26017761 on Build GRCh38
rs545347364 Chr.6: 26017813-26017813 on Build GRCh38
rs765232012 Chr.6: 26017702-26017702 on Build GRCh38
rs777272307 Chr.6: 26017749-26017749 on Build GRCh38
rs935060094 Chr.6: 26018047-26018047 on Build GRCh38
rs961867703 Chr.6: 26017978-26017978 on Build GRCh38
rs201609154 Chr.6: 26017776-26017776 on Build GRCh38
rs562285030 Chr.6: 26017896-26017896 on Build GRCh38
rs62000985 Chr.6: 26017706-26017706 on Build GRCh38
rs866101117 Chr.6: 26017671-26017671 on Build GRCh38
rs771248003 Chr.6: 26017692-26017693 on Build GRCh38
rs748989852 Chr.6: 26017680-26017680 on Build GRCh38
rs1033926498 Chr.6: 26017909-26017909 on Build GRCh38
rs752315798 Chr.6: 26017740-26017740 on Build GRCh38
rs1010627708 Chr.6: 26017724-26017724 on Build GRCh38
rs570082732 Chr.6: 26017721-26017721 on Build GRCh38
rs887170035 Chr.6: 26017834-26017834 on Build GRCh38
rs752330429 Chr.6: 26017766-26017766 on Build GRCh38
rs541398653 Chr.6: 26017788-26017789 on Build GRCh38
rs914239768 Chr.6: 26017678-26017678 on Build GRCh38
rs756731017 Chr.6: 26017786-26017786 on Build GRCh38
rs764290804 Chr.6: 26017722-26017722 on Build GRCh38
rs990078908 Chr.6: 26017938-26017938 on Build GRCh38
rs780823533 Chr.6: 26017754-26017754 on Build GRCh38
rs199610755 Chr.6: 26017712-26017712 on Build GRCh38
rs767988320 Chr.6: 26017732-26017732 on Build GRCh38
rs569963796 Chr.6: 26018051-26018051 on Build GRCh38
rs758117661 Chr.6: 26017767-26017767 on Build GRCh38
rs976840616 Chr.6: 26017675-26017675 on Build GRCh38
rs958218192 Chr.6: 26017917-26017917 on Build GRCh38
rs960955792 Chr.6: 26018003-26018003 on Build GRCh38
rs931169193 Chr.6: 26017824-26017824 on Build GRCh38
rs372168255 Chr.6: 26017752-26017752 on Build GRCh38
rs750850130 Chr.6: 26017733-26017733 on Build GRCh38
rs531403135 Chr.6: 26017867-26017867 on Build GRCh38
rs772096010 Chr.6: 26017694-26017694 on Build GRCh38
rs916272818 Chr.6: 26018018-26018018 on Build GRCh38
rs931266411 Chr.6: 26018024-26018024 on Build GRCh38
rs201733476 Chr.6: 26017764-26017764 on Build GRCh38
rs977204440 Chr.6: 26017950-26017950 on Build GRCh38
rs367920068 Chr.6: 26017705-26017705 on Build GRCh38
rs1050355501 Chr.6: 26018094-26018094 on Build GRCh38
rs1028743465 Chr.6: 26017890-26017890 on Build GRCh38
rs752049023 Chr.6: 26017763-26017763 on Build GRCh38
rs752053479 Chr.6: 26017729-26017729 on Build GRCh38
rs757150864 Chr.6: 26017772-26017772 on Build GRCh38
rs750626867 Chr.6: 26017695-26017695 on Build GRCh38
rs975315544 Chr.6: 26018008-26018008 on Build GRCh38
rs370833756 Chr.6: 26017696-26017696 on Build GRCh38
rs912191402 Chr.6: 26017980-26017980 on Build GRCh38
rs747736182 Chr.6: 26017711-26017711 on Build GRCh38
rs757849608 Chr.6: 26017746-26017746 on Build GRCh38
rs148095398 Chr.6: 26017703-26017703 on Build GRCh38
rs1048678003 Chr.6: 26018052-26018052 on Build GRCh38
rs146301529 Chr.6: 26017674-26017674 on Build GRCh38
rs368674789 Chr.6: 26017762-26017762 on Build GRCh38
rs541593621 Chr.6: 26017877-26017877 on Build GRCh38
rs776702168 Chr.6: 26017706-26017706 on Build GRCh38
rs749517330 Chr.6: 26017779-26017779 on Build GRCh38
rs1043482589 Chr.6: 26017840-26017840 on Build GRCh38
rs548100862 Chr.6: 26017982-26017982 on Build GRCh38
rs778590180 Chr.6: 26017735-26017735 on Build GRCh38
rs1017006208 Chr.6: 26017956-26017956 on Build GRCh38
rs1048458729 Chr.6: 26017833-26017833 on Build GRCh38
rs752139590 Chr.6: 26017704-26017704 on Build GRCh38
rs767993113 Chr.6: 26017681-26017681 on Build GRCh38
rs144780956 Chr.6: 26017718-26017718 on Build GRCh38
rs999304103 Chr.6: 26017856-26017856 on Build GRCh38
rs946397125 Chr.6: 26018020-26018020 on Build GRCh38
rs746948404 Chr.6: 26017751-26017751 on Build GRCh38
rs142577528 Chr.6: 26017810-26017810 on Build GRCh38
rs139988498 Chr.6: 26017717-26017717 on Build GRCh38
rs748023674 Chr.6: 26017688-26017688 on Build GRCh38
rs765417575 Chr.6: 26018016-26018016 on Build GRCh38
rs1043896929 Chr.6: 26018027-26018027 on Build GRCh38
rs781686670 Chr.6: 26017775-26017775 on Build GRCh38
rs1028249824 Chr.6: 26017773-26017773 on Build GRCh38
rs748198269 Chr.6: 26017783-26017783 on Build GRCh38
rs777856269 Chr.6: 26017684-26017685 on Build GRCh38
rs770649989 Chr.6: 26017715-26017715 on Build GRCh38
rs187114919 Chr.6: 26018001-26018001 on Build GRCh38
rs751115728 Chr.6: 26017770-26017770 on Build GRCh38
rs778265738 Chr.6: 26017670-26017670 on Build GRCh38
rs747072384 Chr.6: 26017690-26017691 on Build GRCh38
rs928183660 Chr.6: 26017789-26017789 on Build GRCh38
rs778264839 Chr.6: 26017710-26017710 on Build GRCh38
rs553694004 Chr.6: 26017809-26017809 on Build GRCh38
rs1040189934 Chr.6: 26018076-26018076 on Build GRCh38
rs776779168 Chr.6: 26017701-26017701 on Build GRCh38
rs755878945 Chr.6: 26017777-26017777 on Build GRCh38
rs774542607 Chr.6: 26017782-26017782 on Build GRCh38
rs1004647356 Chr.6: 26017736-26017736 on Build GRCh38
rs1021141563 Chr.6: 26017944-26017944 on Build GRCh38
rs769505738 Chr.6: 26017759-26017759 on Build GRCh38

View Full Product Details