Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 105078713-105079224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIN28B
Entrez Gene ID
389421
Gene Name
lin-28 homolog B
Gene Aliases
CSDD2
UniGene
Hs.23616
Gene Chromosome Location
Chr.6: 104936999-105083332 on Build GRCh38
Transcripts
NM_001004317.3 XM_006715477.2 XM_011535818.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5812417 c.695T>A p.I232K Chr.6: 105078725-105078725 on Build GRCh38
3619148 c.721G>C p.G241R Chr.6: 105078751-105078751 on Build GRCh38
1071705 c.728C>G p.S243* Chr.6: 105078758-105078758 on Build GRCh38
5812863 c.696A>T p.I232I Chr.6: 105078726-105078726 on Build GRCh38
4557789 c.740G>A p.R247K Chr.6: 105078770-105078770 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015427540 Chr.6: 105079113-105079113 on Build GRCh38
rs979850723 Chr.6: 105078929-105078929 on Build GRCh38
rs1026921076 Chr.6: 105079169-105079169 on Build GRCh38
rs761848852 Chr.6: 105078833-105078833 on Build GRCh38
rs947444213 Chr.6: 105079006-105079006 on Build GRCh38
rs961281320 Chr.6: 105078893-105078893 on Build GRCh38
rs144813272 Chr.6: 105078815-105078815 on Build GRCh38
rs556937735 Chr.6: 105078943-105078943 on Build GRCh38
rs996264928 Chr.6: 105078760-105078760 on Build GRCh38
rs771652110 Chr.6: 105078813-105078813 on Build GRCh38
rs1008022798 Chr.6: 105079111-105079111 on Build GRCh38
rs1043321685 Chr.6: 105079002-105079002 on Build GRCh38
rs745471808 Chr.6: 105078799-105078799 on Build GRCh38
rs1040564683 Chr.6: 105078936-105078936 on Build GRCh38
rs1022529509 Chr.6: 105078762-105078762 on Build GRCh38
rs918993026 Chr.6: 105078897-105078897 on Build GRCh38
rs754658515 Chr.6: 105078940-105078940 on Build GRCh38
rs781575321 Chr.6: 105078737-105078737 on Build GRCh38
rs1034899559 Chr.6: 105079132-105079132 on Build GRCh38
rs969237980 Chr.6: 105078780-105078780 on Build GRCh38
rs559844386 Chr.6: 105078988-105078988 on Build GRCh38
rs905023234 Chr.6: 105079060-105079060 on Build GRCh38
rs1002111592 Chr.6: 105078786-105078786 on Build GRCh38
rs185731640 Chr.6: 105079124-105079124 on Build GRCh38
rs1044517867 Chr.6: 105079018-105079018 on Build GRCh38
rs144393333 Chr.6: 105078753-105078753 on Build GRCh38
rs960320258 Chr.6: 105078867-105078867 on Build GRCh38
rs181093638 Chr.6: 105078935-105078935 on Build GRCh38
rs778405403 Chr.6: 105078798-105078798 on Build GRCh38
rs893159016 Chr.6: 105078956-105078956 on Build GRCh38
rs201277574 Chr.6: 105078741-105078741 on Build GRCh38
rs1051828581 Chr.6: 105079076-105079076 on Build GRCh38
rs749856642 Chr.6: 105078790-105078790 on Build GRCh38
rs1053547946 Chr.6: 105078953-105078953 on Build GRCh38
rs140486596 Chr.6: 105078924-105078924 on Build GRCh38
rs780775290 Chr.6: 105079038-105079038 on Build GRCh38
rs565863642 Chr.6: 105078768-105078768 on Build GRCh38
rs373789186 Chr.6: 105078791-105078791 on Build GRCh38
rs76951639 Chr.6: 105078899-105078899 on Build GRCh38
rs564137829 Chr.6: 105079183-105079183 on Build GRCh38
rs573952996 Chr.6: 105078955-105078955 on Build GRCh38
rs768285067 Chr.6: 105078830-105078830 on Build GRCh38
rs527963795 Chr.6: 105078918-105078918 on Build GRCh38
rs947634473 Chr.6: 105078998-105078998 on Build GRCh38
rs776704923 Chr.6: 105078823-105078823 on Build GRCh38
rs890545399 Chr.6: 105079081-105079081 on Build GRCh38
rs757706123 Chr.6: 105078884-105078884 on Build GRCh38
rs760365016 Chr.6: 105078816-105078816 on Build GRCh38
rs768554739 Chr.6: 105078817-105078817 on Build GRCh38
rs545309015 Chr.6: 105079159-105079159 on Build GRCh38
rs543293627 Chr.6: 105078981-105078981 on Build GRCh38
rs969720082 Chr.6: 105079187-105079187 on Build GRCh38
rs996598886 Chr.6: 105079069-105079069 on Build GRCh38

View Full Product Details