Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 113938704-113939205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDAC2
Entrez Gene ID
3066
Gene Name
histone deacetylase 2
Gene Aliases
HD2, RPD3, YAF1
UniGene
Hs.3352
Gene Chromosome Location
Chr.6: 113936156-113971195 on Build GRCh38
Transcripts
NM_001527.3 NR_033441.1 NR_073443.1

SNP Detail

SNP ID Chromosome Location
rs944652764 Chr.6: 113938861-113938861 on Build GRCh38
rs187754099 Chr.6: 113938964-113938964 on Build GRCh38
rs539367701 Chr.6: 113938997-113938997 on Build GRCh38
rs545213994 Chr.6: 113938808-113938811 on Build GRCh38
rs990580176 Chr.6: 113938854-113938854 on Build GRCh38
rs972460385 Chr.6: 113939134-113939134 on Build GRCh38
rs537845464 Chr.6: 113938958-113938958 on Build GRCh38
rs984677266 Chr.6: 113939150-113939150 on Build GRCh38
rs183465135 Chr.6: 113938866-113938866 on Build GRCh38
rs879259137 Chr.6: 113939040-113939040 on Build GRCh38
rs921834698 Chr.6: 113939131-113939131 on Build GRCh38
rs921777324 Chr.6: 113938881-113938881 on Build GRCh38
rs1010940545 Chr.6: 113938774-113938777 on Build GRCh38
rs911882860 Chr.6: 113938813-113938813 on Build GRCh38
rs757722949 Chr.6: 113939087-113939087 on Build GRCh38
rs886231321 Chr.6: 113938983-113938983 on Build GRCh38
rs930462245 Chr.6: 113938888-113938888 on Build GRCh38
rs781667803 Chr.6: 113939152-113939152 on Build GRCh38
rs1049291065 Chr.6: 113938924-113938924 on Build GRCh38
rs1014533900 Chr.6: 113939021-113939021 on Build GRCh38
rs752383780 Chr.6: 113938962-113938962 on Build GRCh38
rs554048298 Chr.6: 113939033-113939033 on Build GRCh38
rs752627413 Chr.6: 113939031-113939032 on Build GRCh38
rs755739205 Chr.6: 113939026-113939026 on Build GRCh38
rs564977093 Chr.6: 113939163-113939163 on Build GRCh38
rs956237007 Chr.6: 113938775-113938775 on Build GRCh38
rs767283476 Chr.6: 113938791-113938791 on Build GRCh38
rs527381933 Chr.6: 113938731-113938731 on Build GRCh38
rs988810642 Chr.6: 113938778-113938778 on Build GRCh38
rs185392904 Chr.6: 113939085-113939085 on Build GRCh38
rs572632992 Chr.6: 113939034-113939034 on Build GRCh38
rs571963058 Chr.6: 113938996-113938996 on Build GRCh38
rs959498995 Chr.6: 113938810-113938810 on Build GRCh38
rs753434710 Chr.6: 113939047-113939047 on Build GRCh38
rs893865845 Chr.6: 113939022-113939022 on Build GRCh38
rs751977514 Chr.6: 113939016-113939016 on Build GRCh38
rs1039564219 Chr.6: 113938760-113938760 on Build GRCh38
rs1056172522 Chr.6: 113939000-113939000 on Build GRCh38
rs543419898 Chr.6: 113939098-113939098 on Build GRCh38
rs890622657 Chr.6: 113938742-113938742 on Build GRCh38
rs1003462040 Chr.6: 113938782-113938783 on Build GRCh38
rs192467124 Chr.6: 113939056-113939056 on Build GRCh38
rs139238022 Chr.6: 113939078-113939078 on Build GRCh38
rs987738801 Chr.6: 113939183-113939183 on Build GRCh38
rs754748807 Chr.6: 113938889-113938890 on Build GRCh38
rs1025230137 Chr.6: 113939153-113939153 on Build GRCh38
rs974642843 Chr.6: 113939119-113939119 on Build GRCh38
rs1039104317 Chr.6: 113939009-113939009 on Build GRCh38
rs930555270 Chr.6: 113939135-113939135 on Build GRCh38
rs962341862 Chr.6: 113938865-113938870 on Build GRCh38
rs893793684 Chr.6: 113938761-113938763 on Build GRCh38
rs894962107 Chr.6: 113939083-113939083 on Build GRCh38
rs747506873 Chr.6: 113939169-113939169 on Build GRCh38
rs549078054 Chr.6: 113938751-113938751 on Build GRCh38
rs918271736 Chr.6: 113938879-113938880 on Build GRCh38
rs929522357 Chr.6: 113938883-113938883 on Build GRCh38
rs67365672 Chr.6: 113938976-113938976 on Build GRCh38
rs1033063456 Chr.6: 113938743-113938743 on Build GRCh38
rs965992476 Chr.6: 113939118-113939118 on Build GRCh38
rs955076787 Chr.6: 113939162-113939162 on Build GRCh38
rs896136860 Chr.6: 113939002-113939002 on Build GRCh38

View Full Product Details