Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74512752-74513260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2IRD1
Entrez Gene ID
9569
Gene Name
GTF2I repeat domain containing 1
Gene Aliases
BEN, CREAM1, GTF3, MUSTRD1, RBAP2, WBS, WBSCR11, WBSCR12, hMusTRD1alpha1
UniGene
Hs.647056
Gene Chromosome Location
Chr.7: 74453790-74603070 on Build GRCh38
Transcripts
NM_001199207.1 NM_005685.3 NM_016328.2 XM_006716182.3 XM_006716183.3 XM_011516713.1 XM_017012802.1 XM_017012803.1 XM_017012804.1 XM_017012805.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1736574 c.180G>C p.E60D Chr.7: 74512886-74512886 on Build GRCh38
4689167 c.160T>A p.C54S Chr.7: 74512866-74512866 on Build GRCh38
3087771 c.174C>T p.H58H Chr.7: 74512880-74512880 on Build GRCh38
3833220 c.207G>A p.K69K Chr.7: 74512913-74512913 on Build GRCh38
3087772 c.212G>A p.R71K Chr.7: 74512918-74512918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373158285 Chr.7: 74512838-74512838 on Build GRCh38
rs536671718 Chr.7: 74512932-74512932 on Build GRCh38
rs530504323 Chr.7: 74513124-74513124 on Build GRCh38
rs146892455 Chr.7: 74513077-74513077 on Build GRCh38
rs782115517 Chr.7: 74512784-74512784 on Build GRCh38
rs541822874 Chr.7: 74513070-74513070 on Build GRCh38
rs782360413 Chr.7: 74512890-74512890 on Build GRCh38
rs529723061 Chr.7: 74513066-74513066 on Build GRCh38
rs782302127 Chr.7: 74513012-74513012 on Build GRCh38
rs372776217 Chr.7: 74512908-74512908 on Build GRCh38
rs1041584574 Chr.7: 74513201-74513201 on Build GRCh38
rs782455709 Chr.7: 74512871-74512871 on Build GRCh38
rs781926381 Chr.7: 74512899-74512899 on Build GRCh38
rs782217840 Chr.7: 74513004-74513004 on Build GRCh38
rs781937968 Chr.7: 74513014-74513014 on Build GRCh38
rs781842043 Chr.7: 74513128-74513128 on Build GRCh38
rs781884512 Chr.7: 74512810-74512810 on Build GRCh38
rs981252134 Chr.7: 74513038-74513038 on Build GRCh38
rs782108422 Chr.7: 74512803-74512803 on Build GRCh38
rs199718773 Chr.7: 74512790-74512790 on Build GRCh38
rs782216607 Chr.7: 74512881-74512881 on Build GRCh38
rs782498759 Chr.7: 74512990-74512990 on Build GRCh38
rs1014600081 Chr.7: 74513028-74513028 on Build GRCh38
rs372797882 Chr.7: 74512780-74512780 on Build GRCh38
rs782299708 Chr.7: 74512875-74512875 on Build GRCh38
rs962006537 Chr.7: 74513030-74513030 on Build GRCh38
rs374266849 Chr.7: 74513018-74513018 on Build GRCh38
rs552971097 Chr.7: 74513024-74513024 on Build GRCh38
rs781826443 Chr.7: 74512795-74512795 on Build GRCh38
rs377230003 Chr.7: 74512783-74512783 on Build GRCh38
rs191451503 Chr.7: 74512936-74512936 on Build GRCh38
rs576371797 Chr.7: 74512991-74512991 on Build GRCh38
rs377354300 Chr.7: 74512961-74512961 on Build GRCh38
rs782740685 Chr.7: 74512978-74512978 on Build GRCh38
rs559491469 Chr.7: 74513025-74513025 on Build GRCh38
rs781866989 Chr.7: 74512837-74512837 on Build GRCh38
rs782708500 Chr.7: 74512804-74512804 on Build GRCh38
rs77717166 Chr.7: 74513005-74513005 on Build GRCh38
rs376904675 Chr.7: 74512896-74512896 on Build GRCh38
rs782002458 Chr.7: 74512880-74512880 on Build GRCh38
rs991296219 Chr.7: 74513088-74513088 on Build GRCh38
rs145072521 Chr.7: 74512879-74512879 on Build GRCh38
rs782667823 Chr.7: 74512872-74512872 on Build GRCh38
rs782546936 Chr.7: 74512817-74512817 on Build GRCh38
rs782776639 Chr.7: 74513088-74513089 on Build GRCh38
rs565790840 Chr.7: 74512853-74512853 on Build GRCh38
rs545837941 Chr.7: 74512772-74512772 on Build GRCh38
rs781793598 Chr.7: 74512979-74512979 on Build GRCh38
rs376848806 Chr.7: 74512999-74512999 on Build GRCh38
rs368969542 Chr.7: 74512789-74512789 on Build GRCh38
rs202134524 Chr.7: 74513015-74513015 on Build GRCh38
rs782573993 Chr.7: 74513001-74513001 on Build GRCh38
rs782762263 Chr.7: 74512998-74512999 on Build GRCh38
rs10268071 Chr.7: 74513235-74513235 on Build GRCh38
rs782623656 Chr.7: 74512846-74512846 on Build GRCh38
rs370667130 Chr.7: 74512998-74512998 on Build GRCh38
rs781870139 Chr.7: 74512992-74512992 on Build GRCh38
rs782759290 Chr.7: 74512823-74512823 on Build GRCh38
rs536029138 Chr.7: 74513000-74513000 on Build GRCh38
rs782409971 Chr.7: 74513013-74513013 on Build GRCh38
rs77802272 Chr.7: 74513125-74513125 on Build GRCh38
rs568587089 Chr.7: 74512997-74512997 on Build GRCh38

View Full Product Details