Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34372504-34373006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1161
Entrez Gene ID
57462
Gene Name
KIAA1161
Gene Aliases
NET37
UniGene
Hs.522083
Gene Chromosome Location
Chr.9: 34366666-34376952 on Build GRCh38
Transcripts
NM_020702.4 XM_011517966.2 XM_017014930.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6693746 c.103C>T p.L35L Chr.9: 34372739-34372739 on Build GRCh38
3745976 c.92T>C p.L31P Chr.9: 34372750-34372750 on Build GRCh38
5166863 c.232C>T p.R78C Chr.9: 34372610-34372610 on Build GRCh38
6229566 c.146G>A p.R49H Chr.9: 34372696-34372696 on Build GRCh38
4635733 c.308C>T p.T103M Chr.9: 34372534-34372534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371798446 Chr.9: 34372566-34372566 on Build GRCh38
rs368703737 Chr.9: 34372724-34372724 on Build GRCh38
rs780289213 Chr.9: 34372626-34372626 on Build GRCh38
rs755964295 Chr.9: 34372916-34372916 on Build GRCh38
rs913324472 Chr.9: 34372538-34372538 on Build GRCh38
rs201509421 Chr.9: 34372897-34372897 on Build GRCh38
rs4879781 Chr.9: 34372785-34372785 on Build GRCh38
rs778863651 Chr.9: 34372768-34372768 on Build GRCh38
rs1006112382 Chr.9: 34372873-34372873 on Build GRCh38
rs201988029 Chr.9: 34372815-34372815 on Build GRCh38
rs201924624 Chr.9: 34372540-34372540 on Build GRCh38
rs773884528 Chr.9: 34372817-34372817 on Build GRCh38
rs933181820 Chr.9: 34372531-34372531 on Build GRCh38
rs376703595 Chr.9: 34372776-34372776 on Build GRCh38
rs1028015538 Chr.9: 34372689-34372689 on Build GRCh38
rs770621275 Chr.9: 34372721-34372721 on Build GRCh38
rs764625887 Chr.9: 34372671-34372671 on Build GRCh38
rs375513654 Chr.9: 34372569-34372569 on Build GRCh38
rs369886696 Chr.9: 34372760-34372760 on Build GRCh38
rs1054215226 Chr.9: 34372651-34372651 on Build GRCh38
rs757132223 Chr.9: 34372853-34372853 on Build GRCh38
rs777247097 Chr.9: 34372947-34372947 on Build GRCh38
rs775660830 Chr.9: 34372971-34372971 on Build GRCh38
rs760451348 Chr.9: 34372719-34372719 on Build GRCh38
rs770219500 Chr.9: 34372953-34372953 on Build GRCh38
rs762234509 Chr.9: 34372740-34372740 on Build GRCh38
rs752197766 Chr.9: 34372737-34372737 on Build GRCh38
rs748981258 Chr.9: 34372921-34372921 on Build GRCh38
rs754187377 Chr.9: 34372823-34372823 on Build GRCh38
rs761863582 Chr.9: 34372977-34372977 on Build GRCh38
rs775173304 Chr.9: 34372863-34372863 on Build GRCh38
rs775762093 Chr.9: 34372595-34372596 on Build GRCh38
rs758842962 Chr.9: 34372545-34372545 on Build GRCh38
rs745662864 Chr.9: 34372859-34372859 on Build GRCh38
rs987291444 Chr.9: 34372533-34372533 on Build GRCh38
rs764119512 Chr.9: 34372584-34372584 on Build GRCh38
rs531504090 Chr.9: 34372745-34372745 on Build GRCh38
rs764679804 Chr.9: 34372734-34372734 on Build GRCh38
rs751700839 Chr.9: 34372975-34372975 on Build GRCh38
rs748847692 Chr.9: 34372864-34372864 on Build GRCh38
rs760248333 Chr.9: 34372659-34372659 on Build GRCh38
rs777677025 Chr.9: 34372848-34372848 on Build GRCh38
rs773937627 Chr.9: 34372865-34372865 on Build GRCh38
rs757738711 Chr.9: 34372675-34372675 on Build GRCh38
rs376242927 Chr.9: 34372629-34372629 on Build GRCh38
rs1043458328 Chr.9: 34372550-34372550 on Build GRCh38
rs755192320 Chr.9: 34372767-34372767 on Build GRCh38
rs367819702 Chr.9: 34372886-34372886 on Build GRCh38
rs774934407 Chr.9: 34372801-34372801 on Build GRCh38
rs770087033 Chr.9: 34372625-34372625 on Build GRCh38
rs745536220 Chr.9: 34372792-34372792 on Build GRCh38
rs967608770 Chr.9: 34372763-34372763 on Build GRCh38
rs748048431 Chr.9: 34372712-34372712 on Build GRCh38
rs750107522 Chr.9: 34372967-34372967 on Build GRCh38
rs202207332 Chr.9: 34372939-34372939 on Build GRCh38
rs373554419 Chr.9: 34372624-34372624 on Build GRCh38
rs781767374 Chr.9: 34372599-34372599 on Build GRCh38
rs756514041 Chr.9: 34372753-34372753 on Build GRCh38
rs577461044 Chr.9: 34372894-34372894 on Build GRCh38
rs1026796632 Chr.9: 34372955-34372955 on Build GRCh38
rs751113708 Chr.9: 34372891-34372895 on Build GRCh38
rs766863116 Chr.9: 34372818-34372818 on Build GRCh38
rs776364497 Chr.9: 34372791-34372791 on Build GRCh38
rs767235933 Chr.9: 34372593-34372593 on Build GRCh38
rs755543919 Chr.9: 34372534-34372534 on Build GRCh38
rs938009163 Chr.9: 34372597-34372597 on Build GRCh38
rs752041698 Chr.9: 34372673-34372673 on Build GRCh38
rs375608013 Chr.9: 34372901-34372901 on Build GRCh38
rs761766271 Chr.9: 34372650-34372650 on Build GRCh38
rs74600562 Chr.9: 34372581-34372581 on Build GRCh38
rs751961772 Chr.9: 34372580-34372580 on Build GRCh38
rs374091744 Chr.9: 34372769-34372769 on Build GRCh38
rs759149442 Chr.9: 34372729-34372729 on Build GRCh38
rs373750261 Chr.9: 34372877-34372877 on Build GRCh38
rs372489726 Chr.9: 34372938-34372938 on Build GRCh38
rs200089454 Chr.9: 34372727-34372727 on Build GRCh38
rs755222208 Chr.9: 34372825-34372825 on Build GRCh38
rs748675255 Chr.9: 34372541-34372541 on Build GRCh38
rs769393546 Chr.9: 34372862-34372862 on Build GRCh38
rs746848230 Chr.9: 34372777-34372777 on Build GRCh38
rs754132155 Chr.9: 34372761-34372761 on Build GRCh38
rs552439888 Chr.9: 34372617-34372617 on Build GRCh38
rs769425565 Chr.9: 34372796-34372796 on Build GRCh38
rs926014393 Chr.9: 34372771-34372771 on Build GRCh38
rs926477066 Chr.9: 34372550-34372570 on Build GRCh38
rs959427122 Chr.9: 34372967-34372967 on Build GRCh38
rs780971805 Chr.9: 34372855-34372855 on Build GRCh38
rs898888467 Chr.9: 34372914-34372914 on Build GRCh38
rs768125948 Chr.9: 34372928-34372928 on Build GRCh38
rs749415302 Chr.9: 34372706-34372706 on Build GRCh38
rs752769476 Chr.9: 34372844-34372844 on Build GRCh38
rs963035672 Chr.9: 34372708-34372708 on Build GRCh38
rs954603951 Chr.9: 34372746-34372746 on Build GRCh38
rs759922282 Chr.9: 34372880-34372880 on Build GRCh38
rs995784404 Chr.9: 34372710-34372710 on Build GRCh38
rs750073561 Chr.9: 34372594-34372594 on Build GRCh38
rs763244109 Chr.9: 34372972-34372972 on Build GRCh38
rs771977253 Chr.9: 34372716-34372716 on Build GRCh38
rs767136106 Chr.9: 34372956-34372959 on Build GRCh38
rs753406540 Chr.9: 34372666-34372666 on Build GRCh38
rs758473364 Chr.9: 34372847-34372847 on Build GRCh38
rs771784841 Chr.9: 34372653-34372653 on Build GRCh38
rs995815782 Chr.9: 34372920-34372920 on Build GRCh38
rs761266634 Chr.9: 34372874-34372874 on Build GRCh38
rs766017522 Chr.9: 34372663-34372663 on Build GRCh38
rs746003043 Chr.9: 34372571-34372571 on Build GRCh38
rs767060731 Chr.9: 34372526-34372526 on Build GRCh38
rs371635711 Chr.9: 34372592-34372592 on Build GRCh38
rs757256895 Chr.9: 34372905-34372905 on Build GRCh38
rs925983305 Chr.9: 34372576-34372576 on Build GRCh38
rs749573561 Chr.9: 34372912-34372912 on Build GRCh38
rs761312002 Chr.9: 34372903-34372904 on Build GRCh38
rs768884848 Chr.9: 34372707-34372707 on Build GRCh38
rs1042653946 Chr.9: 34372787-34372787 on Build GRCh38
rs746972260 Chr.9: 34372850-34372850 on Build GRCh38
rs750735700 Chr.9: 34372696-34372696 on Build GRCh38
rs752996895 Chr.9: 34372895-34372895 on Build GRCh38
rs61738936 Chr.9: 34372618-34372618 on Build GRCh38
rs763478820 Chr.9: 34372603-34372604 on Build GRCh38
rs759024749 Chr.9: 34372669-34372669 on Build GRCh38
rs375180119 Chr.9: 34372958-34372958 on Build GRCh38
rs756459031 Chr.9: 34372697-34372697 on Build GRCh38
rs4879782 Chr.9: 34372875-34372875 on Build GRCh38
rs755742995 Chr.9: 34372595-34372595 on Build GRCh38
rs866383781 Chr.9: 34372808-34372808 on Build GRCh38
rs767541549 Chr.9: 34372980-34372980 on Build GRCh38
rs753277260 Chr.9: 34372606-34372606 on Build GRCh38
rs1047756929 Chr.9: 34372834-34372834 on Build GRCh38
rs978893619 Chr.9: 34372765-34372765 on Build GRCh38
rs1004671508 Chr.9: 34372691-34372691 on Build GRCh38
rs553850134 Chr.9: 34372906-34372906 on Build GRCh38
rs779498767 Chr.9: 34372535-34372535 on Build GRCh38
rs746104060 Chr.9: 34372619-34372619 on Build GRCh38
rs764531821 Chr.9: 34372751-34372751 on Build GRCh38
rs372773584 Chr.9: 34372807-34372807 on Build GRCh38
rs774344202 Chr.9: 34372587-34372587 on Build GRCh38
rs575445815 Chr.9: 34372529-34372529 on Build GRCh38
rs778117056 Chr.9: 34372562-34372562 on Build GRCh38
rs762499379 Chr.9: 34372813-34372813 on Build GRCh38
rs377177253 Chr.9: 34372752-34372752 on Build GRCh38
rs765483979 Chr.9: 34372841-34372841 on Build GRCh38
rs772946228 Chr.9: 34372717-34372717 on Build GRCh38
rs781098241 Chr.9: 34372911-34372911 on Build GRCh38
rs758971652 Chr.9: 34372607-34372607 on Build GRCh38
rs2297776 Chr.9: 34372933-34372933 on Build GRCh38
rs765884956 Chr.9: 34372602-34372602 on Build GRCh38
rs774120280 Chr.9: 34372633-34372633 on Build GRCh38
rs770029104 Chr.9: 34372572-34372572 on Build GRCh38
rs781425533 Chr.9: 34372677-34372677 on Build GRCh38
rs780397910 Chr.9: 34372698-34372698 on Build GRCh38
rs986613484 Chr.9: 34372556-34372556 on Build GRCh38
rs749995055 Chr.9: 34372530-34372530 on Build GRCh38

View Full Product Details