Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228100643-228101151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf35
Entrez Gene ID
79169
Gene Name
chromosome 1 open reading frame 35
Gene Aliases
MMTAG2
UniGene
Hs.445952
Gene Chromosome Location
Chr.1: 228100727-228103462 on Build GRCh38
Transcripts
NM_024319.3 NR_130975.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3789576 c.784G>A p.E262K Chr.1: 228101139-228101139 on Build GRCh38
6615259 c.791G>A p.*264* Chr.1: 228101132-228101132 on Build GRCh38
7102938 c.779G>T p.G260V Chr.1: 228101144-228101144 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1009767038 Chr.1: 228101071-228101071 on Build GRCh38
rs935859486 Chr.1: 228100699-228100699 on Build GRCh38
rs1047973535 Chr.1: 228101061-228101061 on Build GRCh38
rs368630825 Chr.1: 228101096-228101096 on Build GRCh38
rs960779649 Chr.1: 228101129-228101129 on Build GRCh38
rs754634528 Chr.1: 228101098-228101098 on Build GRCh38
rs868247229 Chr.1: 228100851-228100851 on Build GRCh38
rs746438370 Chr.1: 228101120-228101120 on Build GRCh38
rs927229687 Chr.1: 228100667-228100667 on Build GRCh38
rs778651554 Chr.1: 228101099-228101099 on Build GRCh38
rs187255808 Chr.1: 228101108-228101108 on Build GRCh38
rs538677940 Chr.1: 228100900-228100900 on Build GRCh38
rs534502054 Chr.1: 228100979-228100979 on Build GRCh38
rs1034101375 Chr.1: 228101118-228101118 on Build GRCh38
rs554831435 Chr.1: 228101000-228101000 on Build GRCh38
rs776881190 Chr.1: 228101083-228101086 on Build GRCh38
rs747764343 Chr.1: 228101101-228101101 on Build GRCh38
rs937254334 Chr.1: 228100670-228100670 on Build GRCh38
rs116824237 Chr.1: 228101031-228101031 on Build GRCh38
rs775100243 Chr.1: 228100702-228100703 on Build GRCh38
rs887933051 Chr.1: 228100814-228100814 on Build GRCh38
rs892597913 Chr.1: 228100844-228100844 on Build GRCh38
rs552914777 Chr.1: 228101068-228101068 on Build GRCh38
rs922951871 Chr.1: 228101042-228101042 on Build GRCh38
rs763316938 Chr.1: 228100971-228100978 on Build GRCh38
rs993086346 Chr.1: 228100701-228100701 on Build GRCh38
rs958884855 Chr.1: 228100957-228100957 on Build GRCh38
rs989837796 Chr.1: 228100704-228100704 on Build GRCh38
rs187213062 Chr.1: 228100747-228100747 on Build GRCh38
rs920369456 Chr.1: 228101021-228101021 on Build GRCh38
rs768848617 Chr.1: 228100703-228100703 on Build GRCh38
rs888061097 Chr.1: 228101062-228101062 on Build GRCh38
rs898914369 Chr.1: 228100770-228100770 on Build GRCh38
rs1032027298 Chr.1: 228100931-228100931 on Build GRCh38
rs761975832 Chr.1: 228100854-228100857 on Build GRCh38
rs537352729 Chr.1: 228101033-228101033 on Build GRCh38
rs1019538700 Chr.1: 228100836-228100836 on Build GRCh38
rs190705272 Chr.1: 228100883-228100883 on Build GRCh38
rs975819601 Chr.1: 228101034-228101034 on Build GRCh38
rs917163348 Chr.1: 228100980-228100981 on Build GRCh38
rs1036112465 Chr.1: 228100769-228100769 on Build GRCh38
rs1033756873 Chr.1: 228100845-228100845 on Build GRCh38
rs929547552 Chr.1: 228100780-228100780 on Build GRCh38
rs773053841 Chr.1: 228100745-228100745 on Build GRCh38
rs924440192 Chr.1: 228100665-228100665 on Build GRCh38
rs1039978308 Chr.1: 228100764-228100764 on Build GRCh38
rs940565284 Chr.1: 228100985-228100985 on Build GRCh38
rs376487721 Chr.1: 228101115-228101115 on Build GRCh38
rs557347090 Chr.1: 228101050-228101050 on Build GRCh38
rs34368735 Chr.1: 228100883-228100884 on Build GRCh38
rs977351939 Chr.1: 228100916-228100916 on Build GRCh38
rs551981323 Chr.1: 228100958-228100958 on Build GRCh38
rs1048481756 Chr.1: 228100786-228100786 on Build GRCh38
rs895199503 Chr.1: 228100872-228100872 on Build GRCh38
rs992721212 Chr.1: 228100961-228100961 on Build GRCh38
rs1033660833 Chr.1: 228100932-228100932 on Build GRCh38
rs1006434176 Chr.1: 228100830-228100830 on Build GRCh38
rs35143288 Chr.1: 228100896-228100897 on Build GRCh38
rs150982482 Chr.1: 228100674-228100674 on Build GRCh38
rs943029030 Chr.1: 228100751-228100751 on Build GRCh38
rs1050076481 Chr.1: 228100805-228100808 on Build GRCh38
rs569558357 Chr.1: 228100897-228100897 on Build GRCh38
rs182958918 Chr.1: 228100964-228100964 on Build GRCh38
rs901525818 Chr.1: 228100767-228100767 on Build GRCh38

View Full Product Details