Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14774570-14775066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM107B
Entrez Gene ID
83641
Gene Name
family with sequence similarity 107 member B
Gene Aliases
C10orf45
UniGene
Hs.446315
Gene Chromosome Location
Chr.10: 14518557-14774897 on Build GRCh38
Transcripts
NM_001282695.1 NM_031453.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7366937 c.68C>T p.P23L Chr.10: 14774596-14774596 on Build GRCh38
6550697 c.36G>A p.L12L Chr.10: 14774628-14774628 on Build GRCh38
683849 c.76G>C p.A26P Chr.10: 14774588-14774588 on Build GRCh38
3436122 c.87C>T p.A29A Chr.10: 14774577-14774577 on Build GRCh38
3935003 c.84C>T p.L28L Chr.10: 14774580-14774580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772871111 Chr.10: 14774782-14774782 on Build GRCh38
rs766653127 Chr.10: 14774857-14774857 on Build GRCh38
rs775065335 Chr.10: 14774612-14774612 on Build GRCh38
rs925931825 Chr.10: 14774825-14774825 on Build GRCh38
rs113468940 Chr.10: 14774781-14774781 on Build GRCh38
rs368339716 Chr.10: 14774703-14774703 on Build GRCh38
rs987361795 Chr.10: 14774842-14774842 on Build GRCh38
rs745457700 Chr.10: 14774606-14774606 on Build GRCh38
rs751459149 Chr.10: 14774670-14774670 on Build GRCh38
rs540805331 Chr.10: 14774717-14774717 on Build GRCh38
rs756271027 Chr.10: 14774910-14774911 on Build GRCh38
rs929704609 Chr.10: 14774903-14774903 on Build GRCh38
rs1015936071 Chr.10: 14774760-14774760 on Build GRCh38
rs552244278 Chr.10: 14774883-14774883 on Build GRCh38
rs777763218 Chr.10: 14774662-14774662 on Build GRCh38
rs770691389 Chr.10: 14774609-14774610 on Build GRCh38
rs913049823 Chr.10: 14774870-14774870 on Build GRCh38
rs371031031 Chr.10: 14774673-14774673 on Build GRCh38
rs759772122 Chr.10: 14774655-14774655 on Build GRCh38
rs1034627355 Chr.10: 14774689-14774689 on Build GRCh38
rs554193671 Chr.10: 14774960-14774960 on Build GRCh38
rs1024437261 Chr.10: 14775043-14775043 on Build GRCh38
rs951698231 Chr.10: 14774721-14774721 on Build GRCh38
rs780996235 Chr.10: 14774594-14774594 on Build GRCh38
rs921710100 Chr.10: 14774827-14774827 on Build GRCh38
rs984748778 Chr.10: 14774723-14774723 on Build GRCh38
rs766277521 Chr.10: 14774949-14774949 on Build GRCh38
rs189073414 Chr.10: 14774974-14774974 on Build GRCh38
rs758473055 Chr.10: 14774661-14774661 on Build GRCh38
rs911162151 Chr.10: 14774849-14774849 on Build GRCh38
rs762522141 Chr.10: 14774616-14774616 on Build GRCh38
rs895927039 Chr.10: 14774992-14774992 on Build GRCh38
rs61842992 Chr.10: 14774820-14774820 on Build GRCh38
rs752927067 Chr.10: 14774658-14774658 on Build GRCh38
rs114855063 Chr.10: 14774778-14774778 on Build GRCh38
rs761519489 Chr.10: 14774676-14774676 on Build GRCh38
rs963017510 Chr.10: 14774763-14774763 on Build GRCh38
rs933039906 Chr.10: 14774828-14774828 on Build GRCh38
rs765482128 Chr.10: 14774657-14774657 on Build GRCh38
rs532143252 Chr.10: 14774864-14774864 on Build GRCh38
rs753381990 Chr.10: 14774989-14774989 on Build GRCh38
rs779729169 Chr.10: 14774688-14774688 on Build GRCh38
rs1014750241 Chr.10: 14775001-14775001 on Build GRCh38
rs561109040 Chr.10: 14774719-14774719 on Build GRCh38
rs768095213 Chr.10: 14774618-14774618 on Build GRCh38
rs773750659 Chr.10: 14774619-14774619 on Build GRCh38
rs569654987 Chr.10: 14774848-14774848 on Build GRCh38
rs12240920 Chr.10: 14774884-14774884 on Build GRCh38
rs1014928045 Chr.10: 14775005-14775005 on Build GRCh38
rs867730934 Chr.10: 14774910-14774910 on Build GRCh38
rs375693237 Chr.10: 14774679-14774679 on Build GRCh38
rs761290949 Chr.10: 14774624-14774624 on Build GRCh38
rs375173369 Chr.10: 14774728-14774728 on Build GRCh38
rs141618876 Chr.10: 14774609-14774609 on Build GRCh38
rs1005243165 Chr.10: 14774991-14774991 on Build GRCh38
rs748945711 Chr.10: 14774696-14774696 on Build GRCh38
rs894760662 Chr.10: 14775002-14775002 on Build GRCh38
rs766805253 Chr.10: 14774638-14774638 on Build GRCh38
rs564827487 Chr.10: 14774641-14774641 on Build GRCh38
rs377251335 Chr.10: 14774803-14774803 on Build GRCh38
rs751487333 Chr.10: 14774814-14774815 on Build GRCh38
rs769546122 Chr.10: 14774680-14774680 on Build GRCh38
rs760655666 Chr.10: 14774948-14774948 on Build GRCh38
rs774025133 Chr.10: 14774706-14774706 on Build GRCh38
rs767415539 Chr.10: 14774747-14774747 on Build GRCh38

View Full Product Details