Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 26216163-26216656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAD2
Entrez Gene ID
2572
Gene Name
glutamate decarboxylase 2
Gene Aliases
GAD65
UniGene
Hs.231829
Gene Chromosome Location
Chr.10: 26216307-26304562 on Build GRCh38
Transcripts
NM_000818.2 NM_001134366.1

SNP Detail

SNP ID Chromosome Location
rs940929934 Chr.10: 26216554-26216554 on Build GRCh38
rs968442450 Chr.10: 26216232-26216232 on Build GRCh38
rs568851386 Chr.10: 26216340-26216340 on Build GRCh38
rs1034066259 Chr.10: 26216478-26216478 on Build GRCh38
rs537907139 Chr.10: 26216390-26216390 on Build GRCh38
rs547984088 Chr.10: 26216399-26216399 on Build GRCh38
rs995783945 Chr.10: 26216243-26216243 on Build GRCh38
rs1050505245 Chr.10: 26216365-26216365 on Build GRCh38
rs1020109169 Chr.10: 26216413-26216413 on Build GRCh38
rs374467453 Chr.10: 26216294-26216295 on Build GRCh38
rs536855933 Chr.10: 26216560-26216560 on Build GRCh38
rs1049819175 Chr.10: 26216373-26216373 on Build GRCh38
rs897085546 Chr.10: 26216437-26216437 on Build GRCh38
rs889203607 Chr.10: 26216632-26216632 on Build GRCh38
rs1015472326 Chr.10: 26216221-26216221 on Build GRCh38
rs889207771 Chr.10: 26216382-26216382 on Build GRCh38
rs551800915 Chr.10: 26216325-26216325 on Build GRCh38
rs959688953 Chr.10: 26216592-26216592 on Build GRCh38
rs970239196 Chr.10: 26216248-26216248 on Build GRCh38
rs143662552 Chr.10: 26216597-26216597 on Build GRCh38
rs916931188 Chr.10: 26216535-26216535 on Build GRCh38
rs193208856 Chr.10: 26216401-26216401 on Build GRCh38
rs905711454 Chr.10: 26216515-26216515 on Build GRCh38
rs995336689 Chr.10: 26216458-26216458 on Build GRCh38
rs1021189246 Chr.10: 26216634-26216634 on Build GRCh38
rs1051035751 Chr.10: 26216613-26216613 on Build GRCh38
rs567820076 Chr.10: 26216416-26216416 on Build GRCh38
rs945563625 Chr.10: 26216324-26216324 on Build GRCh38
rs1009946970 Chr.10: 26216626-26216626 on Build GRCh38
rs746362677 Chr.10: 26216250-26216250 on Build GRCh38
rs959699062 Chr.10: 26216283-26216283 on Build GRCh38
rs531959007 Chr.10: 26216309-26216309 on Build GRCh38
rs187620304 Chr.10: 26216265-26216265 on Build GRCh38
rs932700331 Chr.10: 26216354-26216354 on Build GRCh38
rs891092169 Chr.10: 26216384-26216384 on Build GRCh38
rs930832752 Chr.10: 26216351-26216351 on Build GRCh38
rs896978649 Chr.10: 26216234-26216234 on Build GRCh38
rs1035449305 Chr.10: 26216576-26216576 on Build GRCh38
rs1026715233 Chr.10: 26216245-26216245 on Build GRCh38
rs551109315 Chr.10: 26216498-26216498 on Build GRCh38
rs914193104 Chr.10: 26216321-26216321 on Build GRCh38
rs917508336 Chr.10: 26216300-26216300 on Build GRCh38
rs2236418 Chr.10: 26216567-26216567 on Build GRCh38
rs2236417 Chr.10: 26216285-26216285 on Build GRCh38
rs1036905514 Chr.10: 26216417-26216417 on Build GRCh38
rs1035329395 Chr.10: 26216281-26216281 on Build GRCh38
rs1027203380 Chr.10: 26216500-26216500 on Build GRCh38
rs758725088 Chr.10: 26216195-26216195 on Build GRCh38
rs992501244 Chr.10: 26216528-26216528 on Build GRCh38
rs1065239 Chr.10: 26216186-26216186 on Build GRCh38
rs539023340 Chr.10: 26216616-26216616 on Build GRCh38
rs901081457 Chr.10: 26216409-26216409 on Build GRCh38

View Full Product Details