Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83263728-83264230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BNC1
Entrez Gene ID
646
Gene Name
basonuclin 1
Gene Aliases
BNC, BSN1, HsT19447
UniGene
Hs.459153
Gene Chromosome Location
Chr.15: 83255884-83284716 on Build GRCh38
Transcripts
NM_001301206.1 NM_001717.3 XM_011521893.1 XM_011521894.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5451375 c.1301G>T p.S434I Chr.15: 83263950-83263950 on Build GRCh38
7418677 c.1190G>T p.S397I Chr.15: 83264061-83264061 on Build GRCh38
77344 c.1171G>A p.G391R Chr.15: 83264080-83264080 on Build GRCh38
1195658 c.1133C>T p.A378V Chr.15: 83264118-83264118 on Build GRCh38
5524644 c.1383G>A p.E461E Chr.15: 83263868-83263868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376472151 Chr.15: 83263754-83263754 on Build GRCh38
rs763147958 Chr.15: 83263957-83263957 on Build GRCh38
rs765598976 Chr.15: 83263987-83263987 on Build GRCh38
rs529712761 Chr.15: 83264054-83264054 on Build GRCh38
rs117340847 Chr.15: 83264156-83264156 on Build GRCh38
rs953634804 Chr.15: 83264183-83264183 on Build GRCh38
rs774487854 Chr.15: 83264207-83264207 on Build GRCh38
rs377144673 Chr.15: 83264188-83264188 on Build GRCh38
rs887437830 Chr.15: 83264051-83264051 on Build GRCh38
rs151194434 Chr.15: 83264180-83264180 on Build GRCh38
rs748702632 Chr.15: 83263893-83263893 on Build GRCh38
rs370870891 Chr.15: 83263760-83263760 on Build GRCh38
rs747572517 Chr.15: 83263920-83263920 on Build GRCh38
rs141436914 Chr.15: 83263772-83263772 on Build GRCh38
rs753840595 Chr.15: 83263813-83263813 on Build GRCh38
rs560685127 Chr.15: 83263970-83263970 on Build GRCh38
rs760326173 Chr.15: 83264084-83264084 on Build GRCh38
rs773630599 Chr.15: 83263851-83263851 on Build GRCh38
rs375359195 Chr.15: 83263986-83263986 on Build GRCh38
rs765511004 Chr.15: 83263940-83263940 on Build GRCh38
rs996599294 Chr.15: 83263934-83263934 on Build GRCh38
rs766627325 Chr.15: 83263811-83263811 on Build GRCh38
rs768158582 Chr.15: 83263778-83263778 on Build GRCh38
rs1015095182 Chr.15: 83264121-83264121 on Build GRCh38
rs766175957 Chr.15: 83263999-83263999 on Build GRCh38
rs376158536 Chr.15: 83263913-83263913 on Build GRCh38
rs761264158 Chr.15: 83263809-83263809 on Build GRCh38
rs372084119 Chr.15: 83264182-83264182 on Build GRCh38
rs866447022 Chr.15: 83263922-83263922 on Build GRCh38
rs781606013 Chr.15: 83264164-83264164 on Build GRCh38
rs751394257 Chr.15: 83263843-83263843 on Build GRCh38
rs140462892 Chr.15: 83264102-83264102 on Build GRCh38
rs754381422 Chr.15: 83264016-83264016 on Build GRCh38
rs777746729 Chr.15: 83263949-83263949 on Build GRCh38
rs775524755 Chr.15: 83264059-83264059 on Build GRCh38
rs866661520 Chr.15: 83263906-83263906 on Build GRCh38
rs750184269 Chr.15: 83263974-83263974 on Build GRCh38
rs766342529 Chr.15: 83264088-83264088 on Build GRCh38
rs1008843271 Chr.15: 83264022-83264022 on Build GRCh38
rs142187743 Chr.15: 83263900-83263900 on Build GRCh38
rs914541066 Chr.15: 83263850-83263850 on Build GRCh38
rs759486222 Chr.15: 83263938-83263938 on Build GRCh38
rs896417613 Chr.15: 83264024-83264024 on Build GRCh38
rs768877017 Chr.15: 83264204-83264204 on Build GRCh38
rs765338696 Chr.15: 83263836-83263836 on Build GRCh38
rs780959264 Chr.15: 83263851-83263851 on Build GRCh38
rs961596009 Chr.15: 83263753-83263753 on Build GRCh38
rs774807331 Chr.15: 83263781-83263781 on Build GRCh38
rs1056199786 Chr.15: 83264015-83264015 on Build GRCh38
rs758155135 Chr.15: 83264124-83264124 on Build GRCh38
rs200109505 Chr.15: 83264147-83264147 on Build GRCh38
rs779651453 Chr.15: 83263878-83263878 on Build GRCh38
rs756978872 Chr.15: 83263847-83263847 on Build GRCh38
rs759909793 Chr.15: 83263774-83263774 on Build GRCh38
rs762216781 Chr.15: 83263783-83263783 on Build GRCh38
rs757220105 Chr.15: 83263761-83263761 on Build GRCh38
rs372547308 Chr.15: 83263763-83263763 on Build GRCh38
rs144958076 Chr.15: 83263857-83263857 on Build GRCh38
rs200106261 Chr.15: 83264083-83264083 on Build GRCh38
rs771310823 Chr.15: 83263927-83263927 on Build GRCh38
rs775640292 Chr.15: 83263950-83263950 on Build GRCh38
rs151242276 Chr.15: 83264056-83264056 on Build GRCh38
rs747392774 Chr.15: 83264150-83264150 on Build GRCh38
rs1049984323 Chr.15: 83263971-83263971 on Build GRCh38
rs779416407 Chr.15: 83263773-83263773 on Build GRCh38
rs753399637 Chr.15: 83264008-83264008 on Build GRCh38
rs768374759 Chr.15: 83263895-83263895 on Build GRCh38
rs758105907 Chr.15: 83264032-83264032 on Build GRCh38
rs79108047 Chr.15: 83264100-83264100 on Build GRCh38
rs771492990 Chr.15: 83264037-83264037 on Build GRCh38
rs749313982 Chr.15: 83264194-83264194 on Build GRCh38
rs759842765 Chr.15: 83263824-83263824 on Build GRCh38
rs747870739 Chr.15: 83264021-83264021 on Build GRCh38
rs986178330 Chr.15: 83263825-83263825 on Build GRCh38
rs752227490 Chr.15: 83264119-83264119 on Build GRCh38
rs776803527 Chr.15: 83263932-83263932 on Build GRCh38
rs750417445 Chr.15: 83263855-83263855 on Build GRCh38
rs773008885 Chr.15: 83264078-83264078 on Build GRCh38
rs773790587 Chr.15: 83263797-83263797 on Build GRCh38
rs772722549 Chr.15: 83263787-83263787 on Build GRCh38
rs778323025 Chr.15: 83264033-83264033 on Build GRCh38
rs778484158 Chr.15: 83264017-83264017 on Build GRCh38
rs372171219 Chr.15: 83264095-83264095 on Build GRCh38
rs763202839 Chr.15: 83264062-83264062 on Build GRCh38
rs995686903 Chr.15: 83264181-83264181 on Build GRCh38
rs377611889 Chr.15: 83263914-83263914 on Build GRCh38
rs142462257 Chr.15: 83264208-83264208 on Build GRCh38
rs758458944 Chr.15: 83263807-83263807 on Build GRCh38
rs923518968 Chr.15: 83263791-83263791 on Build GRCh38
rs747339395 Chr.15: 83264036-83264036 on Build GRCh38
rs757834469 Chr.15: 83264172-83264172 on Build GRCh38
rs764706422 Chr.15: 83264117-83264117 on Build GRCh38
rs764255918 Chr.15: 83264066-83264066 on Build GRCh38
rs745385289 Chr.15: 83263764-83263764 on Build GRCh38
rs778369152 Chr.15: 83263896-83263896 on Build GRCh38

View Full Product Details