Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8585368-8585862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS10
Entrez Gene ID
81794
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 10
Gene Aliases
ADAM-TS10, ADAMTS-10, WMS, WMS1
UniGene
Hs.657508
Gene Chromosome Location
Chr.19: 8580240-8610735 on Build GRCh38
Transcripts
NM_001282352.1 NM_030957.3 XM_006722917.3 XM_017027338.1 XM_017027339.1 XM_017027340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
84478 c.2793G>A p.P931P Chr.19: 8585528-8585528 on Build GRCh38
3541675 c.2724G>A p.S908S Chr.19: 8585597-8585597 on Build GRCh38
6500366 c.2744T>A p.V915D Chr.19: 8585577-8585577 on Build GRCh38
2730575 c.2701G>A p.D901N Chr.19: 8585620-8585620 on Build GRCh38
4409349 c.2772C>T p.D924D Chr.19: 8585549-8585549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781814365 Chr.19: 8585689-8585689 on Build GRCh38
rs782101044 Chr.19: 8585419-8585419 on Build GRCh38
rs782300330 Chr.19: 8585544-8585544 on Build GRCh38
rs782448026 Chr.19: 8585500-8585500 on Build GRCh38
rs1036966878 Chr.19: 8585699-8585699 on Build GRCh38
rs531216020 Chr.19: 8585668-8585668 on Build GRCh38
rs956979801 Chr.19: 8585826-8585826 on Build GRCh38
rs782408737 Chr.19: 8585548-8585548 on Build GRCh38
rs73501568 Chr.19: 8585697-8585697 on Build GRCh38
rs782603993 Chr.19: 8585511-8585511 on Build GRCh38
rs957232017 Chr.19: 8585415-8585415 on Build GRCh38
rs1000784492 Chr.19: 8585820-8585820 on Build GRCh38
rs782673030 Chr.19: 8585677-8585677 on Build GRCh38
rs370280196 Chr.19: 8585630-8585630 on Build GRCh38
rs73922160 Chr.19: 8585767-8585767 on Build GRCh38
rs373459011 Chr.19: 8585597-8585597 on Build GRCh38
rs781947411 Chr.19: 8585549-8585549 on Build GRCh38
rs782733796 Chr.19: 8585424-8585424 on Build GRCh38
rs782323821 Chr.19: 8585699-8585700 on Build GRCh38
rs782804969 Chr.19: 8585456-8585456 on Build GRCh38
rs782269517 Chr.19: 8585694-8585694 on Build GRCh38
rs368617099 Chr.19: 8585754-8585754 on Build GRCh38
rs115358734 Chr.19: 8585728-8585728 on Build GRCh38
rs782064165 Chr.19: 8585600-8585600 on Build GRCh38
rs781925765 Chr.19: 8585700-8585700 on Build GRCh38
rs1032448166 Chr.19: 8585825-8585825 on Build GRCh38
rs782182584 Chr.19: 8585567-8585567 on Build GRCh38
rs782377043 Chr.19: 8585598-8585598 on Build GRCh38
rs781953736 Chr.19: 8585413-8585413 on Build GRCh38
rs905212549 Chr.19: 8585808-8585808 on Build GRCh38
rs782102342 Chr.19: 8585564-8585564 on Build GRCh38
rs375664205 Chr.19: 8585609-8585609 on Build GRCh38
rs781952434 Chr.19: 8585593-8585593 on Build GRCh38
rs782785534 Chr.19: 8585633-8585633 on Build GRCh38
rs782202660 Chr.19: 8585541-8585541 on Build GRCh38
rs367808475 Chr.19: 8585445-8585445 on Build GRCh38
rs782360001 Chr.19: 8585579-8585579 on Build GRCh38
rs202158691 Chr.19: 8585476-8585476 on Build GRCh38
rs781874034 Chr.19: 8585436-8585436 on Build GRCh38
rs781853974 Chr.19: 8585648-8585648 on Build GRCh38
rs781838030 Chr.19: 8585611-8585611 on Build GRCh38
rs562193028 Chr.19: 8585479-8585479 on Build GRCh38
rs73501570 Chr.19: 8585783-8585783 on Build GRCh38

View Full Product Details