Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8586016-8586520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS10
Entrez Gene ID
81794
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 10
Gene Aliases
ADAM-TS10, ADAMTS-10, WMS, WMS1
UniGene
Hs.657508
Gene Chromosome Location
Chr.19: 8580240-8610735 on Build GRCh38
Transcripts
NM_001282352.1 NM_030957.3 XM_006722917.3 XM_017027338.1 XM_017027339.1 XM_017027340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7098640 c.2494G>A p.A832T Chr.19: 8586380-8586380 on Build GRCh38
2730577 c.2546C>T p.A849V Chr.19: 8586236-8586236 on Build GRCh38
1680594 c.2611A>G p.K871E Chr.19: 8586171-8586171 on Build GRCh38
1613116 c.2418C>T p.T806T Chr.19: 8586456-8586456 on Build GRCh38
5032713 c.2461C>T p.R821C Chr.19: 8586413-8586413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782279191 Chr.19: 8586130-8586130 on Build GRCh38
rs781849936 Chr.19: 8586223-8586223 on Build GRCh38
rs782362045 Chr.19: 8586281-8586281 on Build GRCh38
rs940460213 Chr.19: 8586041-8586041 on Build GRCh38
rs782820188 Chr.19: 8586100-8586100 on Build GRCh38
rs10420313 Chr.19: 8586451-8586451 on Build GRCh38
rs375076433 Chr.19: 8586312-8586312 on Build GRCh38
rs782566449 Chr.19: 8586258-8586258 on Build GRCh38
rs1019686797 Chr.19: 8586243-8586243 on Build GRCh38
rs781798823 Chr.19: 8586452-8586452 on Build GRCh38
rs139754132 Chr.19: 8586046-8586046 on Build GRCh38
rs781857217 Chr.19: 8586236-8586236 on Build GRCh38
rs782210820 Chr.19: 8586090-8586090 on Build GRCh38
rs782257731 Chr.19: 8586279-8586279 on Build GRCh38
rs376308911 Chr.19: 8586368-8586368 on Build GRCh38
rs553593479 Chr.19: 8586071-8586071 on Build GRCh38
rs782600400 Chr.19: 8586275-8586275 on Build GRCh38
rs782336542 Chr.19: 8586486-8586486 on Build GRCh38
rs782335136 Chr.19: 8586163-8586163 on Build GRCh38
rs781982290 Chr.19: 8586330-8586330 on Build GRCh38
rs917628457 Chr.19: 8586410-8586410 on Build GRCh38
rs782625211 Chr.19: 8586335-8586335 on Build GRCh38
rs965874905 Chr.19: 8586264-8586264 on Build GRCh38
rs782416288 Chr.19: 8586495-8586495 on Build GRCh38
rs782659603 Chr.19: 8586226-8586226 on Build GRCh38
rs782279692 Chr.19: 8586294-8586294 on Build GRCh38
rs782540299 Chr.19: 8586337-8586337 on Build GRCh38
rs781849329 Chr.19: 8586428-8586428 on Build GRCh38
rs782298163 Chr.19: 8586461-8586461 on Build GRCh38
rs370557753 Chr.19: 8586135-8586135 on Build GRCh38
rs10418929 Chr.19: 8586198-8586198 on Build GRCh38
rs782777003 Chr.19: 8586417-8586417 on Build GRCh38
rs782330611 Chr.19: 8586305-8586305 on Build GRCh38
rs782000281 Chr.19: 8586265-8586265 on Build GRCh38
rs1010103176 Chr.19: 8586211-8586211 on Build GRCh38
rs782704779 Chr.19: 8586405-8586405 on Build GRCh38
rs782465453 Chr.19: 8586225-8586225 on Build GRCh38
rs949522594 Chr.19: 8586053-8586053 on Build GRCh38
rs782638735 Chr.19: 8586459-8586459 on Build GRCh38
rs200003456 Chr.19: 8586202-8586202 on Build GRCh38
rs782181683 Chr.19: 8586346-8586346 on Build GRCh38
rs545953920 Chr.19: 8586453-8586453 on Build GRCh38
rs782113320 Chr.19: 8586313-8586313 on Build GRCh38
rs782428779 Chr.19: 8586331-8586331 on Build GRCh38
rs782582642 Chr.19: 8586344-8586344 on Build GRCh38
rs368291203 Chr.19: 8586315-8586315 on Build GRCh38
rs374989871 Chr.19: 8586485-8586485 on Build GRCh38
rs781957043 Chr.19: 8586379-8586379 on Build GRCh38
rs781821428 Chr.19: 8586333-8586333 on Build GRCh38
rs782351669 Chr.19: 8586104-8586104 on Build GRCh38
rs782547236 Chr.19: 8586433-8586433 on Build GRCh38
rs61757472 Chr.19: 8586465-8586465 on Build GRCh38
rs377236393 Chr.19: 8586078-8586078 on Build GRCh38
rs782146436 Chr.19: 8586401-8586401 on Build GRCh38
rs782021502 Chr.19: 8586395-8586396 on Build GRCh38
rs781964363 Chr.19: 8586408-8586408 on Build GRCh38
rs561610880 Chr.19: 8586398-8586398 on Build GRCh38
rs1045293806 Chr.19: 8586062-8586062 on Build GRCh38
rs886054712 Chr.19: 8586441-8586441 on Build GRCh38
rs782744351 Chr.19: 8586332-8586332 on Build GRCh38
rs781831056 Chr.19: 8586334-8586334 on Build GRCh38
rs782081097 Chr.19: 8586412-8586412 on Build GRCh38
rs886041104 Chr.19: 8586353-8586353 on Build GRCh38
rs782390421 Chr.19: 8586092-8586092 on Build GRCh38
rs782265662 Chr.19: 8586396-8586396 on Build GRCh38
rs782361251 Chr.19: 8586266-8586266 on Build GRCh38
rs781988933 Chr.19: 8586193-8586193 on Build GRCh38
rs781800851 Chr.19: 8586329-8586329 on Build GRCh38
rs200370843 Chr.19: 8586168-8586168 on Build GRCh38
rs782110185 Chr.19: 8586491-8586491 on Build GRCh38
rs575961068 Chr.19: 8586291-8586291 on Build GRCh38
rs151228380 Chr.19: 8586397-8586397 on Build GRCh38
rs575071506 Chr.19: 8586400-8586400 on Build GRCh38
rs576529214 Chr.19: 8586175-8586175 on Build GRCh38
rs782117156 Chr.19: 8586321-8586321 on Build GRCh38
rs782002497 Chr.19: 8586487-8586487 on Build GRCh38
rs782804635 Chr.19: 8586325-8586325 on Build GRCh38
rs782073089 Chr.19: 8586199-8586199 on Build GRCh38
rs782180979 Chr.19: 8586265-8586265 on Build GRCh38
rs782796850 Chr.19: 8586184-8586184 on Build GRCh38
rs1036110508 Chr.19: 8586045-8586045 on Build GRCh38
rs376876724 Chr.19: 8586455-8586455 on Build GRCh38
rs369328663 Chr.19: 8586483-8586483 on Build GRCh38
rs1054223094 Chr.19: 8586192-8586192 on Build GRCh38
rs782633137 Chr.19: 8586454-8586454 on Build GRCh38
rs782583269 Chr.19: 8586260-8586260 on Build GRCh38
rs782520560 Chr.19: 8586456-8586456 on Build GRCh38
rs949575241 Chr.19: 8586432-8586432 on Build GRCh38

View Full Product Details